Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92797364-92797909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA5
Entrez Gene ID
9950
Gene Name
golgin A5
Gene Aliases
GOLIM5, RFG5, ret-II
UniGene
Hs.104320
Gene Chromosome Location
Chr.14: 92794231-92839963 on Build GRCh38
Transcripts
NM_005113.3 XM_011537420.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4795107 c.259G>A p.G87R Chr.14: 92797696-92797696 on Build GRCh38
7135690 c.310C>T p.P104S Chr.14: 92797747-92797747 on Build GRCh38
3754175 c.200C>G p.A67G Chr.14: 92797637-92797637 on Build GRCh38
5989324 c.239G>T p.G80V Chr.14: 92797676-92797676 on Build GRCh38
1323330 c.438G>C p.K146N Chr.14: 92797875-92797875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768992142 Chr.14: 92797558-92797558 on Build GRCh38
rs773139024 Chr.14: 92797426-92797426 on Build GRCh38
rs144267601 Chr.14: 92797500-92797500 on Build GRCh38
rs919784155 Chr.14: 92797698-92797698 on Build GRCh38
rs34532135 Chr.14: 92797800-92797801 on Build GRCh38
rs141963625 Chr.14: 92797845-92797845 on Build GRCh38
rs751560647 Chr.14: 92797862-92797862 on Build GRCh38
rs780072747 Chr.14: 92797771-92797771 on Build GRCh38
rs1033897200 Chr.14: 92797796-92797796 on Build GRCh38
rs757865891 Chr.14: 92797477-92797477 on Build GRCh38
rs775132831 Chr.14: 92797707-92797707 on Build GRCh38
rs760147246 Chr.14: 92797516-92797518 on Build GRCh38
rs748184546 Chr.14: 92797654-92797654 on Build GRCh38
rs1026520489 Chr.14: 92797781-92797781 on Build GRCh38
rs764349513 Chr.14: 92797843-92797843 on Build GRCh38
rs764953832 Chr.14: 92797597-92797597 on Build GRCh38
rs904063112 Chr.14: 92797785-92797785 on Build GRCh38
rs779797527 Chr.14: 92797881-92797881 on Build GRCh38
rs760604591 Chr.14: 92797428-92797428 on Build GRCh38
rs757380809 Chr.14: 92797880-92797880 on Build GRCh38
rs201328765 Chr.14: 92797529-92797529 on Build GRCh38
rs376581530 Chr.14: 92797410-92797410 on Build GRCh38
rs891447266 Chr.14: 92797754-92797754 on Build GRCh38
rs749255638 Chr.14: 92797885-92797885 on Build GRCh38
rs151097986 Chr.14: 92797713-92797713 on Build GRCh38
rs149196374 Chr.14: 92797838-92797838 on Build GRCh38
rs765848702 Chr.14: 92797431-92797431 on Build GRCh38
rs17128572 Chr.14: 92797637-92797637 on Build GRCh38
rs768384721 Chr.14: 92797710-92797710 on Build GRCh38
rs760276060 Chr.14: 92797816-92797816 on Build GRCh38
rs563117542 Chr.14: 92797542-92797542 on Build GRCh38
rs768088371 Chr.14: 92797613-92797613 on Build GRCh38
rs568247621 Chr.14: 92797702-92797702 on Build GRCh38
rs201517262 Chr.14: 92797751-92797751 on Build GRCh38
rs772158192 Chr.14: 92797664-92797664 on Build GRCh38
rs146787240 Chr.14: 92797602-92797602 on Build GRCh38
rs774082205 Chr.14: 92797775-92797775 on Build GRCh38
rs376035342 Chr.14: 92797884-92797884 on Build GRCh38
rs76861952 Chr.14: 92797824-92797824 on Build GRCh38
rs150328141 Chr.14: 92797758-92797758 on Build GRCh38
rs149593701 Chr.14: 92797455-92797455 on Build GRCh38
rs201501111 Chr.14: 92797487-92797487 on Build GRCh38
rs770111862 Chr.14: 92797393-92797393 on Build GRCh38
rs768635009 Chr.14: 92797416-92797416 on Build GRCh38
rs767143480 Chr.14: 92797716-92797716 on Build GRCh38
rs372390722 Chr.14: 92797703-92797703 on Build GRCh38
rs994114647 Chr.14: 92797763-92797763 on Build GRCh38
rs1052875058 Chr.14: 92797474-92797474 on Build GRCh38
rs749341040 Chr.14: 92797415-92797415 on Build GRCh38
rs761442474 Chr.14: 92797611-92797611 on Build GRCh38
rs140027628 Chr.14: 92797533-92797533 on Build GRCh38
rs577055972 Chr.14: 92797817-92797817 on Build GRCh38
rs762853769 Chr.14: 92797709-92797709 on Build GRCh38
rs1009748315 Chr.14: 92797527-92797527 on Build GRCh38
rs368970852 Chr.14: 92797687-92797687 on Build GRCh38
rs766365457 Chr.14: 92797432-92797432 on Build GRCh38
rs1002449161 Chr.14: 92797541-92797541 on Build GRCh38
rs781774988 Chr.14: 92797510-92797510 on Build GRCh38
rs759273994 Chr.14: 92797596-92797596 on Build GRCh38
rs777823905 Chr.14: 92797765-92797765 on Build GRCh38
rs548691389 Chr.14: 92797469-92797469 on Build GRCh38
rs375398139 Chr.14: 92797587-92797587 on Build GRCh38
rs750023400 Chr.14: 92797737-92797737 on Build GRCh38
rs748272820 Chr.14: 92797575-92797575 on Build GRCh38
rs772578887 Chr.14: 92797799-92797799 on Build GRCh38
rs566215762 Chr.14: 92797608-92797608 on Build GRCh38
rs761556497 Chr.14: 92797792-92797792 on Build GRCh38
rs141029204 Chr.14: 92797714-92797714 on Build GRCh38
rs753126089 Chr.14: 92797842-92797842 on Build GRCh38
rs776576975 Chr.14: 92797589-92797589 on Build GRCh38
rs769596121 Chr.14: 92797768-92797768 on Build GRCh38
rs756438734 Chr.14: 92797620-92797620 on Build GRCh38
rs111348610 Chr.14: 92797619-92797619 on Build GRCh38
rs758404420 Chr.14: 92797761-92797761 on Build GRCh38
rs747171055 Chr.14: 92797767-92797767 on Build GRCh38
rs866299587 Chr.14: 92797673-92797673 on Build GRCh38
rs763863424 Chr.14: 92797604-92797604 on Build GRCh38
rs749480282 Chr.14: 92797556-92797556 on Build GRCh38
rs554607146 Chr.14: 92797649-92797649 on Build GRCh38
rs768396335 Chr.14: 92797772-92797772 on Build GRCh38
rs752021322 Chr.14: 92797470-92797472 on Build GRCh38
rs761077825 Chr.14: 92797742-92797742 on Build GRCh38
rs759759384 Chr.14: 92797832-92797832 on Build GRCh38
rs143536819 Chr.14: 92797563-92797563 on Build GRCh38
rs754288879 Chr.14: 92797627-92797627 on Build GRCh38
rs531917271 Chr.14: 92797546-92797546 on Build GRCh38
rs775140712 Chr.14: 92797599-92797599 on Build GRCh38
rs752311515 Chr.14: 92797464-92797464 on Build GRCh38
rs780281785 Chr.14: 92797622-92797622 on Build GRCh38
rs752927689 Chr.14: 92797760-92797760 on Build GRCh38
rs369533656 Chr.14: 92797434-92797434 on Build GRCh38
rs145331070 Chr.14: 92797794-92797794 on Build GRCh38
rs537022332 Chr.14: 92797744-92797744 on Build GRCh38
rs768007764 Chr.14: 92797554-92797555 on Build GRCh38

View Full Product Details