Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94768556-94769055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSC
Entrez Gene ID
145258
Gene Name
goosecoid homeobox
Gene Aliases
SAMS
UniGene
Hs.440438
Gene Chromosome Location
Chr.14: 94768223-94770162 on Build GRCh38
Transcripts
NM_173849.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7080538 c.668C>T p.S223L Chr.14: 94768597-94768597 on Build GRCh38
6554663 c.537C>A p.F179L Chr.14: 94769036-94769036 on Build GRCh38
1371839 c.601G>A p.E201K Chr.14: 94768972-94768972 on Build GRCh38
7370439 c.662C>A p.S221Y Chr.14: 94768603-94768603 on Build GRCh38
4053280 c.599G>A p.R200H Chr.14: 94768974-94768974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376810813 Chr.14: 94768967-94768967 on Build GRCh38
rs552542938 Chr.14: 94768708-94768708 on Build GRCh38
rs977790118 Chr.14: 94768789-94768789 on Build GRCh38
rs930059252 Chr.14: 94768895-94768895 on Build GRCh38
rs369844550 Chr.14: 94769014-94769014 on Build GRCh38
rs746712934 Chr.14: 94769018-94769018 on Build GRCh38
rs771684088 Chr.14: 94768632-94768632 on Build GRCh38
rs746400904 Chr.14: 94768933-94768933 on Build GRCh38
rs772749531 Chr.14: 94768655-94768655 on Build GRCh38
rs774777528 Chr.14: 94768974-94768974 on Build GRCh38
rs918674218 Chr.14: 94768882-94768882 on Build GRCh38
rs148850972 Chr.14: 94768700-94768700 on Build GRCh38
rs760130394 Chr.14: 94768662-94768662 on Build GRCh38
rs987926061 Chr.14: 94768717-94768717 on Build GRCh38
rs368679056 Chr.14: 94768692-94768692 on Build GRCh38
rs112647490 Chr.14: 94768715-94768715 on Build GRCh38
rs1038970223 Chr.14: 94768976-94768976 on Build GRCh38
rs757336569 Chr.14: 94768696-94768696 on Build GRCh38
rs978954333 Chr.14: 94768763-94768763 on Build GRCh38
rs369428563 Chr.14: 94768750-94768750 on Build GRCh38
rs778616169 Chr.14: 94768913-94768913 on Build GRCh38
rs550060776 Chr.14: 94768909-94768909 on Build GRCh38
rs1006129717 Chr.14: 94768718-94768718 on Build GRCh38
rs117555620 Chr.14: 94769021-94769021 on Build GRCh38
rs770405219 Chr.14: 94768929-94768929 on Build GRCh38
rs758070634 Chr.14: 94768925-94768925 on Build GRCh38
rs1048343956 Chr.14: 94768710-94768710 on Build GRCh38
rs540045819 Chr.14: 94768694-94768694 on Build GRCh38
rs997328488 Chr.14: 94768742-94768742 on Build GRCh38
rs925527728 Chr.14: 94768614-94768614 on Build GRCh38
rs539446199 Chr.14: 94768871-94768871 on Build GRCh38
rs554215427 Chr.14: 94768841-94768841 on Build GRCh38
rs776182634 Chr.14: 94768932-94768932 on Build GRCh38
rs1046514698 Chr.14: 94768876-94768876 on Build GRCh38
rs995293182 Chr.14: 94768865-94768865 on Build GRCh38
rs201517530 Chr.14: 94768689-94768689 on Build GRCh38
rs750487062 Chr.14: 94769002-94769002 on Build GRCh38
rs765173607 Chr.14: 94769006-94769006 on Build GRCh38
rs543676956 Chr.14: 94769000-94769000 on Build GRCh38
rs763348360 Chr.14: 94768962-94768962 on Build GRCh38
rs924963398 Chr.14: 94768792-94768792 on Build GRCh38
rs777349659 Chr.14: 94768926-94768926 on Build GRCh38
rs756139979 Chr.14: 94768578-94768578 on Build GRCh38
rs895300812 Chr.14: 94768672-94768672 on Build GRCh38
rs778759721 Chr.14: 94768612-94768612 on Build GRCh38
rs746507132 Chr.14: 94768943-94768943 on Build GRCh38
rs758123549 Chr.14: 94769009-94769009 on Build GRCh38
rs543612146 Chr.14: 94768580-94768580 on Build GRCh38
rs535597760 Chr.14: 94768830-94768830 on Build GRCh38
rs779718984 Chr.14: 94768684-94768684 on Build GRCh38
rs527260651 Chr.14: 94768704-94768704 on Build GRCh38
rs777531321 Chr.14: 94769015-94769015 on Build GRCh38
rs1027869502 Chr.14: 94768697-94768697 on Build GRCh38
rs562169265 Chr.14: 94769008-94769008 on Build GRCh38
rs1050624639 Chr.14: 94768797-94768797 on Build GRCh38
rs3905049 Chr.14: 94768772-94768772 on Build GRCh38
rs746527118 Chr.14: 94768927-94768927 on Build GRCh38
rs1048910239 Chr.14: 94768950-94768950 on Build GRCh38
rs768820007 Chr.14: 94768737-94768737 on Build GRCh38
rs911917048 Chr.14: 94768840-94768840 on Build GRCh38
rs768406079 Chr.14: 94768607-94768607 on Build GRCh38
rs573555823 Chr.14: 94768785-94768797 on Build GRCh38
rs202126439 Chr.14: 94768678-94768678 on Build GRCh38
rs759411614 Chr.14: 94768862-94768862 on Build GRCh38
rs913643736 Chr.14: 94768758-94768758 on Build GRCh38
rs1017884816 Chr.14: 94768733-94768733 on Build GRCh38
rs781332306 Chr.14: 94768698-94768698 on Build GRCh38
rs772741806 Chr.14: 94768661-94768661 on Build GRCh38
rs771441119 Chr.14: 94768666-94768666 on Build GRCh38
rs749109231 Chr.14: 94768593-94768593 on Build GRCh38
rs770293346 Chr.14: 94768951-94768951 on Build GRCh38
rs968168467 Chr.14: 94768762-94768762 on Build GRCh38
rs994136241 Chr.14: 94768707-94768707 on Build GRCh38
rs762116917 Chr.14: 94768988-94768988 on Build GRCh38
rs137930313 Chr.14: 94768860-94768860 on Build GRCh38
rs568211957 Chr.14: 94768777-94768777 on Build GRCh38
rs1034930631 Chr.14: 94768916-94768916 on Build GRCh38
rs141452513 Chr.14: 94768633-94768634 on Build GRCh38
rs72698873 Chr.14: 94768691-94768691 on Build GRCh38

View Full Product Details