Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104171584-104172100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF26A
Entrez Gene ID
26153
Gene Name
kinesin family member 26A
Gene Aliases
-
UniGene
Hs.134970
Gene Chromosome Location
Chr.14: 104138723-104180898 on Build GRCh38
Transcripts
NM_015656.1 XM_011536641.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5381928 c.1219G>A p.D407N Chr.14: 104171828-104171828 on Build GRCh38
1172188 c.1255C>T p.R419W Chr.14: 104171864-104171864 on Build GRCh38
4696067 c.1126C>A p.L376M Chr.14: 104171735-104171735 on Build GRCh38
5882462 c.1141G>A p.A381T Chr.14: 104171750-104171750 on Build GRCh38
4759693 c.1217A>G p.Y406C Chr.14: 104171826-104171826 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867221643 Chr.14: 104171722-104171722 on Build GRCh38
rs887201925 Chr.14: 104172077-104172077 on Build GRCh38
rs368488344 Chr.14: 104171735-104171735 on Build GRCh38
rs981410115 Chr.14: 104171983-104171983 on Build GRCh38
rs567988905 Chr.14: 104171681-104171681 on Build GRCh38
rs1036250425 Chr.14: 104171606-104171606 on Build GRCh38
rs558530686 Chr.14: 104172014-104172014 on Build GRCh38
rs757706475 Chr.14: 104171836-104171836 on Build GRCh38
rs1056200641 Chr.14: 104171789-104171789 on Build GRCh38
rs752153614 Chr.14: 104171929-104171929 on Build GRCh38
rs778311000 Chr.14: 104171706-104171706 on Build GRCh38
rs567153015 Chr.14: 104171873-104171873 on Build GRCh38
rs368528336 Chr.14: 104171776-104171776 on Build GRCh38
rs762902271 Chr.14: 104171753-104171753 on Build GRCh38
rs373030254 Chr.14: 104172001-104172001 on Build GRCh38
rs943345202 Chr.14: 104172016-104172016 on Build GRCh38
rs772514163 Chr.14: 104171705-104171705 on Build GRCh38
rs930574836 Chr.14: 104172005-104172005 on Build GRCh38
rs1007424844 Chr.14: 104171636-104171636 on Build GRCh38
rs553031786 Chr.14: 104171949-104171949 on Build GRCh38
rs182606134 Chr.14: 104171802-104171802 on Build GRCh38
rs767393449 Chr.14: 104171861-104171861 on Build GRCh38
rs910552262 Chr.14: 104172015-104172015 on Build GRCh38
rs199943086 Chr.14: 104171854-104171854 on Build GRCh38
rs761561312 Chr.14: 104171859-104171859 on Build GRCh38
rs748397600 Chr.14: 104171748-104171748 on Build GRCh38
rs748713149 Chr.14: 104171702-104171702 on Build GRCh38
rs748095234 Chr.14: 104171626-104171626 on Build GRCh38
rs778226969 Chr.14: 104171805-104171805 on Build GRCh38
rs867298848 Chr.14: 104171745-104171745 on Build GRCh38
rs924916339 Chr.14: 104172049-104172049 on Build GRCh38
rs372972875 Chr.14: 104171865-104171865 on Build GRCh38
rs147965835 Chr.14: 104171793-104171793 on Build GRCh38
rs769390689 Chr.14: 104171750-104171750 on Build GRCh38
rs563884589 Chr.14: 104171653-104171653 on Build GRCh38
rs371918508 Chr.14: 104171772-104171772 on Build GRCh38
rs537157716 Chr.14: 104171847-104171847 on Build GRCh38
rs377586593 Chr.14: 104171788-104171788 on Build GRCh38
rs910175086 Chr.14: 104171771-104171771 on Build GRCh38
rs919204847 Chr.14: 104171979-104171979 on Build GRCh38
rs374676074 Chr.14: 104171677-104171677 on Build GRCh38
rs866780027 Chr.14: 104171769-104171769 on Build GRCh38
rs761651322 Chr.14: 104171773-104171773 on Build GRCh38
rs572386592 Chr.14: 104172074-104172074 on Build GRCh38
rs747366268 Chr.14: 104171833-104171833 on Build GRCh38
rs779256682 Chr.14: 104171698-104171698 on Build GRCh38
rs202138946 Chr.14: 104171911-104171911 on Build GRCh38
rs958366981 Chr.14: 104171920-104171920 on Build GRCh38
rs566052004 Chr.14: 104171685-104171685 on Build GRCh38
rs76319525 Chr.14: 104172023-104172023 on Build GRCh38
rs978827823 Chr.14: 104171770-104171770 on Build GRCh38
rs943312499 Chr.14: 104172058-104172058 on Build GRCh38
rs901928673 Chr.14: 104172024-104172024 on Build GRCh38
rs962997464 Chr.14: 104171957-104171957 on Build GRCh38
rs1033901188 Chr.14: 104172050-104172050 on Build GRCh38
rs375444859 Chr.14: 104171774-104171774 on Build GRCh38
rs1012585072 Chr.14: 104172052-104172052 on Build GRCh38
rs543989813 Chr.14: 104171652-104171652 on Build GRCh38
rs1017821037 Chr.14: 104171639-104171639 on Build GRCh38
rs971883690 Chr.14: 104171970-104171970 on Build GRCh38
rs1019550351 Chr.14: 104171747-104171747 on Build GRCh38
rs752838716 Chr.14: 104171877-104171877 on Build GRCh38
rs945072321 Chr.14: 104172033-104172033 on Build GRCh38
rs979094042 Chr.14: 104172034-104172034 on Build GRCh38
rs549442781 Chr.14: 104171684-104171684 on Build GRCh38
rs917098933 Chr.14: 104171785-104171785 on Build GRCh38
rs913685337 Chr.14: 104172026-104172026 on Build GRCh38
rs749162945 Chr.14: 104171844-104171844 on Build GRCh38
rs201201891 Chr.14: 104171843-104171843 on Build GRCh38
rs201446180 Chr.14: 104171950-104171950 on Build GRCh38
rs753212733 Chr.14: 104171801-104171801 on Build GRCh38
rs778596142 Chr.14: 104172068-104172068 on Build GRCh38
rs999889858 Chr.14: 104171878-104171878 on Build GRCh38
rs919173786 Chr.14: 104171817-104171817 on Build GRCh38
rs370698809 Chr.14: 104171711-104171711 on Build GRCh38
rs962433023 Chr.14: 104171864-104171864 on Build GRCh38
rs544343535 Chr.14: 104172070-104172070 on Build GRCh38
rs758610431 Chr.14: 104171888-104171888 on Build GRCh38
rs1039391045 Chr.14: 104172066-104172066 on Build GRCh38
rs1043403248 Chr.14: 104171828-104171828 on Build GRCh38
rs369583853 Chr.14: 104171945-104171945 on Build GRCh38
rs763840180 Chr.14: 104171759-104171759 on Build GRCh38
rs374018343 Chr.14: 104171956-104171956 on Build GRCh38
rs376110702 Chr.14: 104171868-104171868 on Build GRCh38
rs758977098 Chr.14: 104171804-104171804 on Build GRCh38
rs375034353 Chr.14: 104171840-104171840 on Build GRCh38
rs1047546933 Chr.14: 104172076-104172076 on Build GRCh38
rs780678262 Chr.14: 104171839-104171839 on Build GRCh38
rs1009689919 Chr.14: 104171860-104171860 on Build GRCh38
rs756331527 Chr.14: 104171676-104171676 on Build GRCh38
rs776403392 Chr.14: 104171719-104171719 on Build GRCh38
rs750596905 Chr.14: 104171673-104171673 on Build GRCh38
rs1006522098 Chr.14: 104171842-104171842 on Build GRCh38
rs903995132 Chr.14: 104172044-104172044 on Build GRCh38

View Full Product Details