Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63608532-63609040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL22
Entrez Gene ID
283807
Gene Name
F-box and leucine rich repeat protein 22
Gene Aliases
Fbl22
UniGene
Hs.631163
Gene Chromosome Location
Chr.15: 63596225-63608898 on Build GRCh38
Transcripts
XM_017022086.1 NM_003922.3 XM_017022699.1 XM_017022700.1 XM_017022701.1 XM_017022702.1 XM_017022703.1 XM_017022704.1 XM_017022705.1

Gene Symbol
HERC1
Entrez Gene ID
8925
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase family member 1
Gene Aliases
p532, p619
UniGene
Hs.210385
Gene Chromosome Location
Chr.15: 63608618-63833964 on Build GRCh38
Transcripts
XM_017022086.1 NM_003922.3 XM_017022699.1 XM_017022700.1 XM_017022701.1 XM_017022702.1 XM_017022703.1 XM_017022704.1 XM_017022705.1

SNP Detail

SNP ID Chromosome Location
rs190428949 Chr.15: 63608705-63608705 on Build GRCh38
rs540723417 Chr.15: 63608647-63608647 on Build GRCh38
rs1014340776 Chr.15: 63608913-63608913 on Build GRCh38
rs777741365 Chr.15: 63608960-63608960 on Build GRCh38
rs546426400 Chr.15: 63608691-63608691 on Build GRCh38
rs1030874893 Chr.15: 63608979-63608979 on Build GRCh38
rs749110220 Chr.15: 63608993-63608993 on Build GRCh38
rs35287299 Chr.15: 63608935-63608936 on Build GRCh38
rs566205111 Chr.15: 63608983-63608983 on Build GRCh38
rs1046332812 Chr.15: 63608848-63608848 on Build GRCh38
rs1006332402 Chr.15: 63608946-63608946 on Build GRCh38
rs182252541 Chr.15: 63608758-63608758 on Build GRCh38
rs952300467 Chr.15: 63608702-63608702 on Build GRCh38
rs908041772 Chr.15: 63609007-63609007 on Build GRCh38
rs749334127 Chr.15: 63608996-63608996 on Build GRCh38
rs149515322 Chr.15: 63608648-63608648 on Build GRCh38
rs990519404 Chr.15: 63608781-63608781 on Build GRCh38
rs920881881 Chr.15: 63608699-63608699 on Build GRCh38
rs75894300 Chr.15: 63608566-63608566 on Build GRCh38
rs1032159270 Chr.15: 63608622-63608622 on Build GRCh38
rs1006280106 Chr.15: 63608591-63608591 on Build GRCh38
rs1026700887 Chr.15: 63608561-63608561 on Build GRCh38
rs1040651232 Chr.15: 63608767-63608767 on Build GRCh38
rs1028228512 Chr.15: 63608611-63608611 on Build GRCh38
rs967006209 Chr.15: 63608981-63608981 on Build GRCh38
rs922261910 Chr.15: 63608769-63608769 on Build GRCh38
rs970081660 Chr.15: 63608654-63608654 on Build GRCh38
rs964856619 Chr.15: 63608623-63608623 on Build GRCh38
rs932358415 Chr.15: 63608805-63608805 on Build GRCh38
rs992486817 Chr.15: 63608791-63608791 on Build GRCh38
rs747852140 Chr.15: 63608861-63608861 on Build GRCh38
rs916441080 Chr.15: 63608836-63608836 on Build GRCh38
rs572540370 Chr.15: 63608683-63608683 on Build GRCh38
rs1050090189 Chr.15: 63608831-63608832 on Build GRCh38
rs144013522 Chr.15: 63608716-63608716 on Build GRCh38
rs967329094 Chr.15: 63608688-63608688 on Build GRCh38
rs1017699492 Chr.15: 63608959-63608959 on Build GRCh38
rs774257878 Chr.15: 63608923-63608926 on Build GRCh38
rs997690604 Chr.15: 63608974-63608974 on Build GRCh38
rs1032037660 Chr.15: 63609012-63609012 on Build GRCh38
rs904351104 Chr.15: 63608613-63608613 on Build GRCh38
rs867797432 Chr.15: 63608870-63608870 on Build GRCh38
rs1131647 Chr.15: 63608914-63608914 on Build GRCh38
rs139175515 Chr.15: 63608912-63608912 on Build GRCh38
rs901056487 Chr.15: 63608574-63608574 on Build GRCh38
rs936448646 Chr.15: 63608763-63608763 on Build GRCh38
rs1017815564 Chr.15: 63608615-63608615 on Build GRCh38
rs949219488 Chr.15: 63608842-63608842 on Build GRCh38
rs561716616 Chr.15: 63608877-63608877 on Build GRCh38
rs1028294782 Chr.15: 63608973-63608973 on Build GRCh38
rs929845018 Chr.15: 63608884-63608884 on Build GRCh38
rs532979088 Chr.15: 63609000-63609000 on Build GRCh38
rs116143018 Chr.15: 63608628-63608628 on Build GRCh38

View Full Product Details