Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63615690-63616202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERC1
Entrez Gene ID
8925
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase family member 1
Gene Aliases
p532, p619
UniGene
Hs.210385
Gene Chromosome Location
Chr.15: 63608618-63833964 on Build GRCh38
Transcripts
NM_003922.3 XM_017022699.1 XM_017022700.1 XM_017022701.1 XM_017022702.1 XM_017022703.1 XM_017022704.1 XM_017022705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
963700 c.14071C>T p.R4691* Chr.15: 63615791-63615791 on Build GRCh38
4396707 c.14033C>T p.S4678F Chr.15: 63615829-63615829 on Build GRCh38
1209544 c.13942-2A>G p.? Chr.15: 63615922-63615922 on Build GRCh38
3502973 c.14001C>T p.I4667I Chr.15: 63615861-63615861 on Build GRCh38
963703 c.13967G>T p.G4656V Chr.15: 63615895-63615895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764097509 Chr.15: 63615884-63615884 on Build GRCh38
rs939066403 Chr.15: 63616038-63616038 on Build GRCh38
rs914928129 Chr.15: 63615984-63615984 on Build GRCh38
rs375787266 Chr.15: 63615994-63615994 on Build GRCh38
rs192231642 Chr.15: 63615987-63615987 on Build GRCh38
rs901991690 Chr.15: 63615975-63615975 on Build GRCh38
rs935613149 Chr.15: 63615970-63615970 on Build GRCh38
rs371576830 Chr.15: 63615866-63615866 on Build GRCh38
rs369089188 Chr.15: 63616018-63616018 on Build GRCh38
rs59408197 Chr.15: 63616034-63616034 on Build GRCh38
rs759972144 Chr.15: 63615750-63615750 on Build GRCh38
rs563489663 Chr.15: 63615872-63615872 on Build GRCh38
rs75713882 Chr.15: 63616003-63616003 on Build GRCh38
rs531973049 Chr.15: 63615993-63615993 on Build GRCh38
rs771212463 Chr.15: 63615745-63615745 on Build GRCh38
rs747759485 Chr.15: 63615851-63615851 on Build GRCh38
rs760873384 Chr.15: 63615765-63615765 on Build GRCh38
rs182587471 Chr.15: 63615937-63615937 on Build GRCh38
rs778579502 Chr.15: 63615800-63615800 on Build GRCh38
rs537704865 Chr.15: 63616080-63616080 on Build GRCh38
rs746917734 Chr.15: 63615820-63615820 on Build GRCh38
rs1005511199 Chr.15: 63616076-63616076 on Build GRCh38
rs374359281 Chr.15: 63615760-63615760 on Build GRCh38
rs886934746 Chr.15: 63616071-63616071 on Build GRCh38
rs909215431 Chr.15: 63616151-63616151 on Build GRCh38
rs188404892 Chr.15: 63615971-63615971 on Build GRCh38
rs753230662 Chr.15: 63615764-63615764 on Build GRCh38
rs909436268 Chr.15: 63615902-63615902 on Build GRCh38
rs750623082 Chr.15: 63615927-63615927 on Build GRCh38
rs899713244 Chr.15: 63616125-63616125 on Build GRCh38
rs780984939 Chr.15: 63615948-63615948 on Build GRCh38
rs755553890 Chr.15: 63615964-63615964 on Build GRCh38
rs746580382 Chr.15: 63615727-63615727 on Build GRCh38
rs184664455 Chr.15: 63616142-63616142 on Build GRCh38
rs541476504 Chr.15: 63615721-63615721 on Build GRCh38
rs772982803 Chr.15: 63615859-63615859 on Build GRCh38
rs761797214 Chr.15: 63615924-63615924 on Build GRCh38
rs758643728 Chr.15: 63615839-63615839 on Build GRCh38
rs776176315 Chr.15: 63615868-63615868 on Build GRCh38
rs774788018 Chr.15: 63615749-63615749 on Build GRCh38
rs368591079 Chr.15: 63615804-63615804 on Build GRCh38
rs972713616 Chr.15: 63616173-63616173 on Build GRCh38
rs764197624 Chr.15: 63615774-63615774 on Build GRCh38
rs747748584 Chr.15: 63615952-63615952 on Build GRCh38
rs567210985 Chr.15: 63615932-63615932 on Build GRCh38
rs988777440 Chr.15: 63615789-63615789 on Build GRCh38
rs971611676 Chr.15: 63616062-63616062 on Build GRCh38
rs981317539 Chr.15: 63616104-63616104 on Build GRCh38
rs576252314 Chr.15: 63616052-63616052 on Build GRCh38
rs762055261 Chr.15: 63615738-63615738 on Build GRCh38
rs1046961484 Chr.15: 63616048-63616048 on Build GRCh38
rs1006527313 Chr.15: 63616022-63616022 on Build GRCh38
rs371842557 Chr.15: 63616141-63616141 on Build GRCh38
rs188736640 Chr.15: 63616168-63616168 on Build GRCh38
rs78824147 Chr.15: 63616129-63616129 on Build GRCh38
rs951302383 Chr.15: 63615791-63615791 on Build GRCh38
rs367720452 Chr.15: 63615856-63615856 on Build GRCh38
rs376171992 Chr.15: 63615894-63615894 on Build GRCh38
rs754876718 Chr.15: 63615945-63615945 on Build GRCh38
rs189997840 Chr.15: 63615903-63615903 on Build GRCh38
rs551636516 Chr.15: 63615778-63615778 on Build GRCh38
rs372357333 Chr.15: 63615842-63615842 on Build GRCh38
rs757446033 Chr.15: 63615790-63615790 on Build GRCh38
rs200200988 Chr.15: 63615734-63615734 on Build GRCh38
rs374739089 Chr.15: 63615850-63615850 on Build GRCh38
rs750707071 Chr.15: 63615824-63615824 on Build GRCh38
rs780016545 Chr.15: 63615814-63615814 on Build GRCh38
rs954815711 Chr.15: 63616149-63616149 on Build GRCh38
rs954763810 Chr.15: 63615920-63615920 on Build GRCh38
rs760023312 Chr.15: 63615861-63615861 on Build GRCh38
rs750286687 Chr.15: 63615801-63615801 on Build GRCh38
rs546183135 Chr.15: 63615977-63615983 on Build GRCh38
rs1016854697 Chr.15: 63616121-63616121 on Build GRCh38

View Full Product Details