Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68302300-68302843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA11
Entrez Gene ID
22801
Gene Name
integrin subunit alpha 11
Gene Aliases
HsT18964
UniGene
Hs.436416
Gene Chromosome Location
Chr.15: 68297433-68432312 on Build GRCh38
Transcripts
NM_001004439.1 XM_005254228.3 XM_011521363.2

SNP Detail

SNP ID Chromosome Location
rs1019275471 Chr.15: 68302667-68302667 on Build GRCh38
rs79251975 Chr.15: 68302353-68302353 on Build GRCh38
rs754580941 Chr.15: 68302391-68302391 on Build GRCh38
rs533383579 Chr.15: 68302417-68302417 on Build GRCh38
rs570777943 Chr.15: 68302528-68302528 on Build GRCh38
rs1010134441 Chr.15: 68302545-68302545 on Build GRCh38
rs139403785 Chr.15: 68302422-68302422 on Build GRCh38
rs540609691 Chr.15: 68302806-68302806 on Build GRCh38
rs571737467 Chr.15: 68302797-68302797 on Build GRCh38
rs905254586 Chr.15: 68302330-68302330 on Build GRCh38
rs74023106 Chr.15: 68302482-68302482 on Build GRCh38
rs963681665 Chr.15: 68302681-68302681 on Build GRCh38
rs56165498 Chr.15: 68302737-68302737 on Build GRCh38
rs1019338949 Chr.15: 68302410-68302410 on Build GRCh38
rs905522442 Chr.15: 68302641-68302641 on Build GRCh38
rs976723426 Chr.15: 68302486-68302486 on Build GRCh38
rs949811570 Chr.15: 68302604-68302604 on Build GRCh38
rs140033923 Chr.15: 68302456-68302457 on Build GRCh38
rs919811558 Chr.15: 68302469-68302469 on Build GRCh38
rs539719564 Chr.15: 68302590-68302590 on Build GRCh38
rs188652997 Chr.15: 68302375-68302375 on Build GRCh38
rs376428924 Chr.15: 68302556-68302556 on Build GRCh38
rs938111084 Chr.15: 68302533-68302533 on Build GRCh38
rs1028448263 Chr.15: 68302742-68302742 on Build GRCh38
rs181081998 Chr.15: 68302483-68302483 on Build GRCh38
rs1049439552 Chr.15: 68302658-68302658 on Build GRCh38
rs934863081 Chr.15: 68302326-68302326 on Build GRCh38
rs1044026918 Chr.15: 68302606-68302606 on Build GRCh38
rs893958324 Chr.15: 68302370-68302370 on Build GRCh38
rs1054276538 Chr.15: 68302537-68302537 on Build GRCh38
rs963733150 Chr.15: 68302446-68302446 on Build GRCh38
rs1042655672 Chr.15: 68302577-68302577 on Build GRCh38
rs996634743 Chr.15: 68302734-68302734 on Build GRCh38
rs1007818465 Chr.15: 68302409-68302409 on Build GRCh38
rs908459212 Chr.15: 68302804-68302804 on Build GRCh38
rs895540448 Chr.15: 68302602-68302602 on Build GRCh38
rs1122247 Chr.15: 68302523-68302523 on Build GRCh38
rs563703512 Chr.15: 68302812-68302812 on Build GRCh38
rs777407131 Chr.15: 68302653-68302653 on Build GRCh38
rs982017592 Chr.15: 68302808-68302808 on Build GRCh38
rs1018806179 Chr.15: 68302717-68302717 on Build GRCh38
rs534146755 Chr.15: 68302544-68302544 on Build GRCh38
rs747457304 Chr.15: 68302455-68302456 on Build GRCh38
rs1053924226 Chr.15: 68302367-68302367 on Build GRCh38
rs926172638 Chr.15: 68302800-68302800 on Build GRCh38
rs183822013 Chr.15: 68302517-68302517 on Build GRCh38
rs570121219 Chr.15: 68302451-68302451 on Build GRCh38
rs10851784 Chr.15: 68302792-68302792 on Build GRCh38
rs965421423 Chr.15: 68302725-68302725 on Build GRCh38
rs149991492 Chr.15: 68302713-68302713 on Build GRCh38
rs1007849514 Chr.15: 68302664-68302664 on Build GRCh38
rs886380695 Chr.15: 68302675-68302675 on Build GRCh38
rs749743910 Chr.15: 68302774-68302774 on Build GRCh38
rs758430513 Chr.15: 68302371-68302371 on Build GRCh38
rs975127263 Chr.15: 68302461-68302461 on Build GRCh38
rs541371963 Chr.15: 68302354-68302354 on Build GRCh38
rs576118179 Chr.15: 68302726-68302726 on Build GRCh38
rs541949925 Chr.15: 68302791-68302791 on Build GRCh38
rs535010965 Chr.15: 68302522-68302522 on Build GRCh38
rs1027139563 Chr.15: 68302749-68302749 on Build GRCh38

View Full Product Details