Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1510518-1511018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT140
Entrez Gene ID
9742
Gene Name
intraflagellar transport 140
Gene Aliases
MZSDS, SRTD9, WDTC2, c305C8.4, c380F5.1, gs114
UniGene
Hs.389438
Gene Chromosome Location
Chr.16: 1510427-1612108 on Build GRCh38
Transcripts
NM_014714.3 XM_006720990.3 XM_006720991.3 XM_006720992.3 XM_011522766.2 XM_011522767.2 XM_017023910.1 XM_017023911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7143636 c.4380C>G p.D1460E Chr.16: 1510953-1510953 on Build GRCh38
368618 c.4352A>G p.D1451G Chr.16: 1510981-1510981 on Build GRCh38
1563238 c.4380C>T p.D1460D Chr.16: 1510953-1510953 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs886051707 Chr.16: 1510786-1510786 on Build GRCh38
rs985796594 Chr.16: 1510892-1510892 on Build GRCh38
rs868011285 Chr.16: 1510825-1510825 on Build GRCh38
rs1003241920 Chr.16: 1510735-1510735 on Build GRCh38
rs909009465 Chr.16: 1510843-1510843 on Build GRCh38
rs758511414 Chr.16: 1510923-1510923 on Build GRCh38
rs755667163 Chr.16: 1510937-1510937 on Build GRCh38
rs1037821178 Chr.16: 1510810-1510810 on Build GRCh38
rs200162770 Chr.16: 1510932-1510932 on Build GRCh38
rs1010515510 Chr.16: 1510857-1510857 on Build GRCh38
rs753261966 Chr.16: 1510997-1510997 on Build GRCh38
rs761282751 Chr.16: 1510898-1510898 on Build GRCh38
rs958587227 Chr.16: 1510827-1510827 on Build GRCh38
rs184405274 Chr.16: 1510709-1510709 on Build GRCh38
rs779634878 Chr.16: 1510995-1510995 on Build GRCh38
rs1053730 Chr.16: 1510885-1510885 on Build GRCh38
rs775384967 Chr.16: 1510968-1510968 on Build GRCh38
rs143732634 Chr.16: 1510541-1510541 on Build GRCh38
rs372303683 Chr.16: 1510826-1510826 on Build GRCh38
rs1004204259 Chr.16: 1510833-1510833 on Build GRCh38
rs940780636 Chr.16: 1510875-1510875 on Build GRCh38
rs1029386853 Chr.16: 1510975-1510975 on Build GRCh38
rs367764759 Chr.16: 1510903-1510903 on Build GRCh38
rs1036775824 Chr.16: 1510711-1510711 on Build GRCh38
rs1001045418 Chr.16: 1510788-1510788 on Build GRCh38
rs1049627167 Chr.16: 1510799-1510799 on Build GRCh38
rs771632573 Chr.16: 1510954-1510954 on Build GRCh38
rs368151122 Chr.16: 1510795-1510795 on Build GRCh38
rs981359201 Chr.16: 1510598-1510598 on Build GRCh38
rs543398853 Chr.16: 1510884-1510885 on Build GRCh38
rs993654959 Chr.16: 1510601-1510601 on Build GRCh38
rs997513782 Chr.16: 1510771-1510771 on Build GRCh38
rs376007738 Chr.16: 1510931-1510931 on Build GRCh38
rs1033770672 Chr.16: 1510757-1510757 on Build GRCh38
rs541891452 Chr.16: 1510831-1510831 on Build GRCh38
rs760889117 Chr.16: 1510966-1510986 on Build GRCh38
rs372243759 Chr.16: 1510933-1510933 on Build GRCh38
rs899346069 Chr.16: 1510586-1510587 on Build GRCh38
rs201065562 Chr.16: 1510952-1510952 on Build GRCh38
rs889089332 Chr.16: 1510542-1510542 on Build GRCh38
rs763899729 Chr.16: 1510927-1510927 on Build GRCh38
rs757221765 Chr.16: 1510693-1510693 on Build GRCh38
rs751635341 Chr.16: 1510607-1510607 on Build GRCh38
rs923331704 Chr.16: 1510884-1510884 on Build GRCh38
rs753109181 Chr.16: 1510955-1510956 on Build GRCh38
rs773668018 Chr.16: 1510895-1510895 on Build GRCh38
rs937199452 Chr.16: 1510673-1510673 on Build GRCh38
rs752974569 Chr.16: 1510921-1510921 on Build GRCh38
rs149678731 Chr.16: 1510881-1510881 on Build GRCh38
rs910575696 Chr.16: 1510802-1510802 on Build GRCh38
rs1048867968 Chr.16: 1510691-1510691 on Build GRCh38
rs776891038 Chr.16: 1510916-1510916 on Build GRCh38
rs750381445 Chr.16: 1510983-1510983 on Build GRCh38
rs112626319 Chr.16: 1510812-1510812 on Build GRCh38
rs763213109 Chr.16: 1510971-1510971 on Build GRCh38
rs545865311 Chr.16: 1510947-1510947 on Build GRCh38
rs768372140 Chr.16: 1510950-1510950 on Build GRCh38
rs573166923 Chr.16: 1510829-1510829 on Build GRCh38
rs933911702 Chr.16: 1510796-1510796 on Build GRCh38
rs971382709 Chr.16: 1510664-1510664 on Build GRCh38
rs113310961 Chr.16: 1510836-1510836 on Build GRCh38
rs927211265 Chr.16: 1510604-1510604 on Build GRCh38
rs760102170 Chr.16: 1510920-1510920 on Build GRCh38
rs556741432 Chr.16: 1510787-1510787 on Build GRCh38
rs886051705 Chr.16: 1510666-1510666 on Build GRCh38
rs61749517 Chr.16: 1510953-1510953 on Build GRCh38
rs901811355 Chr.16: 1510734-1510734 on Build GRCh38
rs142106374 Chr.16: 1510979-1510979 on Build GRCh38
rs971295132 Chr.16: 1510577-1510577 on Build GRCh38
rs1022023304 Chr.16: 1510870-1510870 on Build GRCh38
rs769920958 Chr.16: 1510958-1510958 on Build GRCh38
rs1007651476 Chr.16: 1510554-1510554 on Build GRCh38
rs536823833 Chr.16: 1510761-1510761 on Build GRCh38
rs760987640 Chr.16: 1510791-1510791 on Build GRCh38
rs767835848 Chr.16: 1510950-1510952 on Build GRCh38
rs371193393 Chr.16: 1510928-1510928 on Build GRCh38
rs755962676 Chr.16: 1510994-1510994 on Build GRCh38
rs1032487541 Chr.16: 1510790-1510790 on Build GRCh38
rs958990368 Chr.16: 1510759-1510759 on Build GRCh38
rs528105061 Chr.16: 1510571-1510571 on Build GRCh38
rs767245111 Chr.16: 1510981-1510981 on Build GRCh38
rs144879630 Chr.16: 1510942-1510942 on Build GRCh38
rs548066567 Chr.16: 1510624-1510624 on Build GRCh38
rs761585883 Chr.16: 1510980-1510980 on Build GRCh38
rs1026592633 Chr.16: 1510647-1510647 on Build GRCh38
rs759791391 Chr.16: 1510946-1510946 on Build GRCh38
rs1020444517 Chr.16: 1510806-1510806 on Build GRCh38
rs1050687916 Chr.16: 1510772-1510772 on Build GRCh38
rs553238182 Chr.16: 1510817-1510817 on Build GRCh38
rs886051706 Chr.16: 1510718-1510718 on Build GRCh38
rs777181948 Chr.16: 1510955-1510955 on Build GRCh38

View Full Product Details