Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1794592-1795082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFALS
Entrez Gene ID
3483
Gene Name
insulin like growth factor binding protein acid labile subunit
Gene Aliases
ACLSD, ALS
UniGene
Hs.839
Gene Chromosome Location
Chr.16: 1790413-1794908 on Build GRCh38
Transcripts
NR_027389.1

SNP Detail

SNP ID Chromosome Location
rs918619291 Chr.16: 1794625-1794625 on Build GRCh38
rs3829561 Chr.16: 1794871-1794871 on Build GRCh38
rs1011985957 Chr.16: 1794854-1794854 on Build GRCh38
rs756766594 Chr.16: 1794998-1794998 on Build GRCh38
rs553022054 Chr.16: 1794753-1794753 on Build GRCh38
rs960783535 Chr.16: 1794624-1794624 on Build GRCh38
rs532013617 Chr.16: 1795060-1795060 on Build GRCh38
rs931087538 Chr.16: 1795027-1795027 on Build GRCh38
rs1042716317 Chr.16: 1794812-1794812 on Build GRCh38
rs376488090 Chr.16: 1794927-1794927 on Build GRCh38
rs564313964 Chr.16: 1794752-1794752 on Build GRCh38
rs578049823 Chr.16: 1794870-1794870 on Build GRCh38
rs918829473 Chr.16: 1794772-1794772 on Build GRCh38
rs887027455 Chr.16: 1794727-1794727 on Build GRCh38
rs1038845767 Chr.16: 1794674-1794674 on Build GRCh38
rs938638700 Chr.16: 1794869-1794869 on Build GRCh38
rs933996526 Chr.16: 1794626-1794626 on Build GRCh38
rs994753265 Chr.16: 1795061-1795061 on Build GRCh38
rs3829562 Chr.16: 1794874-1794874 on Build GRCh38
rs911310348 Chr.16: 1794783-1794783 on Build GRCh38
rs946554885 Chr.16: 1794944-1794944 on Build GRCh38
rs36163787 Chr.16: 1794677-1794677 on Build GRCh38
rs2256923 Chr.16: 1794780-1794780 on Build GRCh38
rs893638556 Chr.16: 1794686-1794686 on Build GRCh38
rs772629388 Chr.16: 1794873-1794873 on Build GRCh38
rs973425770 Chr.16: 1794770-1794770 on Build GRCh38
rs1024128198 Chr.16: 1794768-1794768 on Build GRCh38
rs34687459 Chr.16: 1794763-1794763 on Build GRCh38
rs753411042 Chr.16: 1794795-1794796 on Build GRCh38
rs942461563 Chr.16: 1794685-1794685 on Build GRCh38
rs979861885 Chr.16: 1794779-1794779 on Build GRCh38
rs975550353 Chr.16: 1794808-1794808 on Build GRCh38
rs562877274 Chr.16: 1794936-1794936 on Build GRCh38
rs776742870 Chr.16: 1794912-1794912 on Build GRCh38
rs34679887 Chr.16: 1794732-1794732 on Build GRCh38
rs771085360 Chr.16: 1794907-1794907 on Build GRCh38
rs994326170 Chr.16: 1794872-1794872 on Build GRCh38
rs9924224 Chr.16: 1794800-1794800 on Build GRCh38
rs1046703482 Chr.16: 1794726-1794726 on Build GRCh38
rs1003628569 Chr.16: 1794827-1794827 on Build GRCh38
rs36157422 Chr.16: 1794693-1794693 on Build GRCh38
rs886767952 Chr.16: 1794853-1794853 on Build GRCh38
rs765201399 Chr.16: 1794931-1794931 on Build GRCh38
rs894843836 Chr.16: 1795044-1795044 on Build GRCh38
rs943189005 Chr.16: 1794672-1794672 on Build GRCh38
rs200043350 Chr.16: 1794799-1794800 on Build GRCh38
rs955578247 Chr.16: 1794777-1794777 on Build GRCh38
rs946552269 Chr.16: 1794807-1794807 on Build GRCh38
rs549220196 Chr.16: 1794636-1794636 on Build GRCh38
rs916617485 Chr.16: 1794631-1794631 on Build GRCh38
rs1054535633 Chr.16: 1794792-1794792 on Build GRCh38
rs36150258 Chr.16: 1794689-1794689 on Build GRCh38
rs968436779 Chr.16: 1794632-1794632 on Build GRCh38
rs775310441 Chr.16: 1795047-1795047 on Build GRCh38
rs747381073 Chr.16: 1794880-1794880 on Build GRCh38
rs557911808 Chr.16: 1794830-1794830 on Build GRCh38
rs914635913 Chr.16: 1794939-1794939 on Build GRCh38
rs894812736 Chr.16: 1794831-1794831 on Build GRCh38
rs569207341 Chr.16: 1794654-1794654 on Build GRCh38
rs34898949 Chr.16: 1794722-1794722 on Build GRCh38
rs920072184 Chr.16: 1795016-1795016 on Build GRCh38
rs1042189202 Chr.16: 1794963-1794963 on Build GRCh38
rs868676035 Chr.16: 1794767-1794767 on Build GRCh38
rs911258228 Chr.16: 1794923-1794923 on Build GRCh38
rs551936469 Chr.16: 1794718-1794718 on Build GRCh38
rs35158822 Chr.16: 1794822-1794822 on Build GRCh38
rs755607168 Chr.16: 1794846-1794846 on Build GRCh38
rs924203551 Chr.16: 1794642-1794642 on Build GRCh38
rs1035519071 Chr.16: 1794740-1794740 on Build GRCh38
rs1017097288 Chr.16: 1794760-1794760 on Build GRCh38
rs566436425 Chr.16: 1794778-1794778 on Build GRCh38
rs555744435 Chr.16: 1794795-1794795 on Build GRCh38
rs544111874 Chr.16: 1794819-1794819 on Build GRCh38
rs1056303197 Chr.16: 1795015-1795015 on Build GRCh38
rs942799885 Chr.16: 1794787-1794787 on Build GRCh38
rs923858463 Chr.16: 1794976-1794976 on Build GRCh38
rs964264375 Chr.16: 1795000-1795000 on Build GRCh38
rs990256432 Chr.16: 1794932-1794932 on Build GRCh38
rs1020857015 Chr.16: 1794692-1794692 on Build GRCh38
rs529295455 Chr.16: 1794888-1794888 on Build GRCh38
rs779651533 Chr.16: 1794851-1794851 on Build GRCh38
rs35902226 Chr.16: 1794684-1794684 on Build GRCh38
rs1003186149 Chr.16: 1794723-1794723 on Build GRCh38
rs759297325 Chr.16: 1794896-1794896 on Build GRCh38
rs903940893 Chr.16: 1794902-1794902 on Build GRCh38
rs370534474 Chr.16: 1795057-1795057 on Build GRCh38

View Full Product Details