Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3065701-3066149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL32
Entrez Gene ID
9235
Gene Name
interleukin 32
Gene Aliases
IL-32alpha, IL-32beta, IL-32delta, IL-32gamma, NK4, TAIF, TAIFa, TAIFb, TAIFc, TAIFd
UniGene
Hs.943
Gene Chromosome Location
Chr.16: 3065312-3069819 on Build GRCh38
Transcripts
NM_001012631.1 NM_001012632.1 NM_001012633.1 NM_001012634.1 NM_001012635.1 NM_001012636.1 NM_001012718.1 NM_001308078.1 NM_004221.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5384787 c.12G>A p.P4P Chr.16: 3065823-3065823 on Build GRCh38
4405106 c.7T>G p.F3V Chr.16: 3065818-3065818 on Build GRCh38
5384788 c.12G>A p.P4P Chr.16: 3065823-3065823 on Build GRCh38
4405107 c.7T>G p.F3V Chr.16: 3065818-3065818 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755328474 Chr.16: 3066048-3066048 on Build GRCh38
rs575128686 Chr.16: 3065734-3065734 on Build GRCh38
rs137891370 Chr.16: 3065823-3065823 on Build GRCh38
rs770407535 Chr.16: 3065764-3065764 on Build GRCh38
rs749063506 Chr.16: 3065909-3065909 on Build GRCh38
rs372259388 Chr.16: 3065972-3065972 on Build GRCh38
rs947348872 Chr.16: 3065954-3065954 on Build GRCh38
rs1047606153 Chr.16: 3065893-3065893 on Build GRCh38
rs776130655 Chr.16: 3065774-3065774 on Build GRCh38
rs1043369400 Chr.16: 3066055-3066055 on Build GRCh38
rs753743076 Chr.16: 3065795-3065795 on Build GRCh38
rs998918294 Chr.16: 3066075-3066075 on Build GRCh38
rs372765113 Chr.16: 3065875-3065875 on Build GRCh38
rs758429758 Chr.16: 3065740-3065740 on Build GRCh38
rs919948394 Chr.16: 3065880-3065880 on Build GRCh38
rs752875759 Chr.16: 3066082-3066082 on Build GRCh38
rs772781958 Chr.16: 3065975-3065975 on Build GRCh38
rs531934291 Chr.16: 3066114-3066114 on Build GRCh38
rs777891968 Chr.16: 3066098-3066098 on Build GRCh38
rs188483845 Chr.16: 3065885-3065885 on Build GRCh38
rs555658957 Chr.16: 3065961-3065961 on Build GRCh38
rs117411512 Chr.16: 3066072-3066072 on Build GRCh38
rs1035400453 Chr.16: 3066081-3066081 on Build GRCh38
rs532644073 Chr.16: 3065999-3065999 on Build GRCh38
rs754305935 Chr.16: 3066042-3066042 on Build GRCh38
rs375770611 Chr.16: 3066073-3066073 on Build GRCh38
rs761790625 Chr.16: 3065856-3065856 on Build GRCh38
rs202051533 Chr.16: 3065754-3065754 on Build GRCh38
rs149965494 Chr.16: 3065793-3065793 on Build GRCh38
rs771875586 Chr.16: 3065840-3065840 on Build GRCh38
rs932063441 Chr.16: 3065973-3065973 on Build GRCh38
rs745554267 Chr.16: 3065838-3065838 on Build GRCh38
rs899743312 Chr.16: 3065939-3065939 on Build GRCh38
rs181673289 Chr.16: 3065953-3065953 on Build GRCh38
rs149042998 Chr.16: 3065906-3065906 on Build GRCh38
rs537689102 Chr.16: 3066008-3066008 on Build GRCh38
rs988999810 Chr.16: 3066060-3066060 on Build GRCh38
rs373647835 Chr.16: 3065767-3065767 on Build GRCh38
rs527798842 Chr.16: 3065944-3065944 on Build GRCh38
rs368745485 Chr.16: 3065863-3065863 on Build GRCh38
rs376408189 Chr.16: 3065876-3065876 on Build GRCh38
rs199922289 Chr.16: 3065833-3065833 on Build GRCh38
rs531970849 Chr.16: 3065905-3065905 on Build GRCh38
rs916773449 Chr.16: 3066004-3066004 on Build GRCh38
rs758575799 Chr.16: 3066087-3066087 on Build GRCh38
rs1043095113 Chr.16: 3065959-3065959 on Build GRCh38
rs776494993 Chr.16: 3065853-3065853 on Build GRCh38
rs960468983 Chr.16: 3066051-3066051 on Build GRCh38
rs1023777782 Chr.16: 3066093-3066093 on Build GRCh38
rs992323924 Chr.16: 3065985-3065985 on Build GRCh38
rs773909642 Chr.16: 3065963-3065963 on Build GRCh38
rs531419740 Chr.16: 3066020-3066020 on Build GRCh38
rs1038413238 Chr.16: 3065728-3065728 on Build GRCh38
rs547445097 Chr.16: 3065945-3065945 on Build GRCh38
rs775458799 Chr.16: 3065777-3065777 on Build GRCh38
rs367665595 Chr.16: 3065877-3065877 on Build GRCh38
rs368483493 Chr.16: 3065899-3065899 on Build GRCh38
rs992376544 Chr.16: 3065848-3065848 on Build GRCh38
rs755307247 Chr.16: 3065822-3065822 on Build GRCh38
rs371234723 Chr.16: 3065745-3065745 on Build GRCh38
rs750696132 Chr.16: 3065806-3065806 on Build GRCh38
rs73490422 Chr.16: 3065842-3065842 on Build GRCh38
rs754293568 Chr.16: 3066021-3066021 on Build GRCh38
rs895107973 Chr.16: 3065720-3065720 on Build GRCh38
rs969613859 Chr.16: 3066005-3066005 on Build GRCh38
rs375695095 Chr.16: 3065775-3065775 on Build GRCh38
rs761481973 Chr.16: 3065964-3065964 on Build GRCh38
rs114106725 Chr.16: 3065946-3065946 on Build GRCh38
rs898577356 Chr.16: 3065731-3065731 on Build GRCh38
rs540346009 Chr.16: 3066052-3066052 on Build GRCh38
rs968969381 Chr.16: 3066068-3066068 on Build GRCh38
rs571812592 Chr.16: 3065907-3065907 on Build GRCh38
rs553455522 Chr.16: 3066061-3066061 on Build GRCh38
rs557539086 Chr.16: 3066010-3066010 on Build GRCh38
rs924109828 Chr.16: 3065836-3065836 on Build GRCh38
rs1047093359 Chr.16: 3066012-3066012 on Build GRCh38
rs563884988 Chr.16: 3065776-3065776 on Build GRCh38
rs929967913 Chr.16: 3066011-3066011 on Build GRCh38
rs758420856 Chr.16: 3065867-3065867 on Build GRCh38
rs200820637 Chr.16: 3065808-3065808 on Build GRCh38
rs773942215 Chr.16: 3065769-3065769 on Build GRCh38
rs983172650 Chr.16: 3066119-3066119 on Build GRCh38
rs1039499986 Chr.16: 3066040-3066040 on Build GRCh38
rs1008091384 Chr.16: 3065986-3065986 on Build GRCh38
rs200892861 Chr.16: 3065809-3065809 on Build GRCh38
rs967466770 Chr.16: 3065951-3065951 on Build GRCh38
rs948042856 Chr.16: 3065725-3065725 on Build GRCh38
rs760499463 Chr.16: 3065784-3065784 on Build GRCh38
rs912595207 Chr.16: 3066120-3066120 on Build GRCh38
rs563245891 Chr.16: 3066099-3066099 on Build GRCh38
rs890964492 Chr.16: 3065760-3065760 on Build GRCh38
rs777851387 Chr.16: 3065785-3065785 on Build GRCh38
rs951702901 Chr.16: 3066116-3066116 on Build GRCh38
rs890946955 Chr.16: 3065976-3065976 on Build GRCh38
rs377133786 Chr.16: 3065797-3065797 on Build GRCh38
rs944097604 Chr.16: 3066122-3066122 on Build GRCh38
rs765349497 Chr.16: 3065804-3065804 on Build GRCh38
rs374370998 Chr.16: 3065843-3065843 on Build GRCh38
rs561942890 Chr.16: 3066115-3066115 on Build GRCh38
rs1011426292 Chr.16: 3066054-3066054 on Build GRCh38
rs770405203 Chr.16: 3065824-3065824 on Build GRCh38
rs899689322 Chr.16: 3066045-3066045 on Build GRCh38

View Full Product Details