Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29992068-29992576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIRIP3
Entrez Gene ID
8479
Gene Name
HIRA interacting protein 3
Gene Aliases
-
UniGene
Hs.567370
Gene Chromosome Location
Chr.16: 29992321-29996096 on Build GRCh38
Transcripts
NM_001197323.1 NM_003609.4 NM_004783.3 XM_011545984.1 XM_011545985.1

Gene Symbol
TAOK2
Entrez Gene ID
9344
Gene Name
TAO kinase 2
Gene Aliases
MAP3K17, PSK, PSK1, PSK1-BETA, TAO1, TAO2
UniGene
Hs.291623
Gene Chromosome Location
Chr.16: 29973641-29992261 on Build GRCh38
Transcripts
NM_001197323.1 NM_003609.4 NM_004783.3 XM_011545984.1 XM_011545985.1

SNP Detail

SNP ID Chromosome Location
rs933947568 Chr.16: 29992237-29992237 on Build GRCh38
rs67430411 Chr.16: 29992452-29992453 on Build GRCh38
rs773797269 Chr.16: 29992433-29992433 on Build GRCh38
rs909598045 Chr.16: 29992150-29992150 on Build GRCh38
rs749283906 Chr.16: 29992164-29992164 on Build GRCh38
rs896797675 Chr.16: 29992136-29992136 on Build GRCh38
rs570739170 Chr.16: 29992523-29992523 on Build GRCh38
rs1006397241 Chr.16: 29992198-29992198 on Build GRCh38
rs1006344812 Chr.16: 29992551-29992551 on Build GRCh38
rs1043553361 Chr.16: 29992451-29992451 on Build GRCh38
rs935455185 Chr.16: 29992463-29992463 on Build GRCh38
rs972877961 Chr.16: 29992393-29992393 on Build GRCh38
rs933648437 Chr.16: 29992404-29992404 on Build GRCh38
rs1043839810 Chr.16: 29992318-29992318 on Build GRCh38
rs1016654609 Chr.16: 29992374-29992374 on Build GRCh38
rs1036791669 Chr.16: 29992128-29992128 on Build GRCh38
rs903909388 Chr.16: 29992323-29992323 on Build GRCh38
rs112533759 Chr.16: 29992435-29992435 on Build GRCh38
rs953561564 Chr.16: 29992391-29992391 on Build GRCh38
rs115551876 Chr.16: 29992378-29992378 on Build GRCh38
rs914374722 Chr.16: 29992422-29992422 on Build GRCh38
rs576355841 Chr.16: 29992107-29992107 on Build GRCh38
rs887952060 Chr.16: 29992183-29992183 on Build GRCh38
rs77712644 Chr.16: 29992450-29992451 on Build GRCh38
rs938642781 Chr.16: 29992472-29992472 on Build GRCh38
rs969533427 Chr.16: 29992366-29992366 on Build GRCh38
rs551930203 Chr.16: 29992490-29992490 on Build GRCh38
rs984891048 Chr.16: 29992371-29992371 on Build GRCh38
rs754947890 Chr.16: 29992363-29992366 on Build GRCh38
rs972398495 Chr.16: 29992173-29992173 on Build GRCh38
rs182926596 Chr.16: 29992266-29992266 on Build GRCh38
rs186208574 Chr.16: 29992342-29992342 on Build GRCh38
rs1023735602 Chr.16: 29992175-29992175 on Build GRCh38
rs1015123788 Chr.16: 29992247-29992247 on Build GRCh38
rs918519366 Chr.16: 29992184-29992184 on Build GRCh38
rs773516026 Chr.16: 29992483-29992483 on Build GRCh38
rs1048141901 Chr.16: 29992533-29992533 on Build GRCh38
rs924503277 Chr.16: 29992439-29992439 on Build GRCh38
rs1013869471 Chr.16: 29992337-29992337 on Build GRCh38
rs374980627 Chr.16: 29992286-29992286 on Build GRCh38
rs896463050 Chr.16: 29992335-29992335 on Build GRCh38
rs1031023238 Chr.16: 29992334-29992334 on Build GRCh38
rs948437200 Chr.16: 29992524-29992524 on Build GRCh38
rs986738611 Chr.16: 29992411-29992411 on Build GRCh38
rs143947811 Chr.16: 29992397-29992397 on Build GRCh38
rs1057172340 Chr.16: 29992473-29992473 on Build GRCh38
rs867067741 Chr.16: 29992390-29992390 on Build GRCh38
rs867180076 Chr.16: 29992534-29992534 on Build GRCh38
rs878891173 Chr.16: 29992091-29992091 on Build GRCh38
rs373316363 Chr.16: 29992129-29992129 on Build GRCh38
rs889663212 Chr.16: 29992273-29992273 on Build GRCh38
rs896526476 Chr.16: 29992505-29992505 on Build GRCh38
rs58217768 Chr.16: 29992451-29992452 on Build GRCh38
rs36047629 Chr.16: 29992166-29992167 on Build GRCh38
rs990372260 Chr.16: 29992525-29992526 on Build GRCh38
rs942471385 Chr.16: 29992292-29992292 on Build GRCh38
rs868098809 Chr.16: 29992111-29992111 on Build GRCh38
rs559850663 Chr.16: 29992408-29992408 on Build GRCh38
rs999542650 Chr.16: 29992330-29992330 on Build GRCh38
rs903668877 Chr.16: 29992457-29992461 on Build GRCh38
rs868538552 Chr.16: 29992321-29992321 on Build GRCh38

View Full Product Details