Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29993338-29993833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIRIP3
Entrez Gene ID
8479
Gene Name
HIRA interacting protein 3
Gene Aliases
-
UniGene
Hs.567370
Gene Chromosome Location
Chr.16: 29992321-29996096 on Build GRCh38
Transcripts
NM_001197323.1 NM_003609.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
703327 c.1537C>A p.P513T Chr.16: 29993341-29993341 on Build GRCh38
1478777 c.1404G>A p.M468I Chr.16: 29993644-29993644 on Build GRCh38
5980961 c.1270C>G p.P424A Chr.16: 29993778-29993778 on Build GRCh38
7120648 c.1261G>A p.E421K Chr.16: 29993787-29993787 on Build GRCh38
1640458 c.1507+8G>A p.? Chr.16: 29993451-29993451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547533863 Chr.16: 29993756-29993756 on Build GRCh38
rs763754084 Chr.16: 29993553-29993553 on Build GRCh38
rs904812761 Chr.16: 29993533-29993533 on Build GRCh38
rs773442313 Chr.16: 29993517-29993517 on Build GRCh38
rs376218176 Chr.16: 29993630-29993630 on Build GRCh38
rs767816944 Chr.16: 29993752-29993752 on Build GRCh38
rs910690040 Chr.16: 29993722-29993722 on Build GRCh38
rs754457784 Chr.16: 29993461-29993461 on Build GRCh38
rs771077091 Chr.16: 29993684-29993684 on Build GRCh38
rs758814365 Chr.16: 29993575-29993575 on Build GRCh38
rs774202117 Chr.16: 29993627-29993627 on Build GRCh38
rs753859926 Chr.16: 29993757-29993757 on Build GRCh38
rs771879251 Chr.16: 29993777-29993777 on Build GRCh38
rs201909868 Chr.16: 29993609-29993609 on Build GRCh38
rs762154116 Chr.16: 29993436-29993436 on Build GRCh38
rs752941065 Chr.16: 29993796-29993796 on Build GRCh38
rs754510811 Chr.16: 29993362-29993362 on Build GRCh38
rs368827418 Chr.16: 29993433-29993433 on Build GRCh38
rs926094276 Chr.16: 29993772-29993772 on Build GRCh38
rs753526967 Chr.16: 29993646-29993646 on Build GRCh38
rs971973543 Chr.16: 29993591-29993591 on Build GRCh38
rs768820692 Chr.16: 29993417-29993417 on Build GRCh38
rs770702994 Chr.16: 29993624-29993624 on Build GRCh38
rs768604334 Chr.16: 29993493-29993493 on Build GRCh38
rs759910294 Chr.16: 29993748-29993748 on Build GRCh38
rs753238284 Chr.16: 29993753-29993753 on Build GRCh38
rs141766249 Chr.16: 29993654-29993654 on Build GRCh38
rs772638811 Chr.16: 29993388-29993388 on Build GRCh38
rs370424441 Chr.16: 29993367-29993367 on Build GRCh38
rs372423732 Chr.16: 29993444-29993444 on Build GRCh38
rs138542330 Chr.16: 29993785-29993785 on Build GRCh38
rs773934939 Chr.16: 29993541-29993541 on Build GRCh38
rs564027311 Chr.16: 29993653-29993653 on Build GRCh38
rs760508276 Chr.16: 29993723-29993724 on Build GRCh38
rs748206727 Chr.16: 29993607-29993607 on Build GRCh38
rs77645102 Chr.16: 29993475-29993475 on Build GRCh38
rs752275269 Chr.16: 29993627-29993634 on Build GRCh38
rs1014071878 Chr.16: 29993389-29993389 on Build GRCh38
rs35431046 Chr.16: 29993479-29993479 on Build GRCh38
rs553327917 Chr.16: 29993586-29993586 on Build GRCh38
rs950741997 Chr.16: 29993698-29993698 on Build GRCh38
rs373495037 Chr.16: 29993532-29993532 on Build GRCh38
rs1032448674 Chr.16: 29993531-29993531 on Build GRCh38
rs562013780 Chr.16: 29993721-29993721 on Build GRCh38
rs772587729 Chr.16: 29993482-29993482 on Build GRCh38
rs1039199342 Chr.16: 29993767-29993767 on Build GRCh38
rs776153731 Chr.16: 29993396-29993396 on Build GRCh38
rs777297444 Chr.16: 29993478-29993478 on Build GRCh38
rs774022148 Chr.16: 29993483-29993483 on Build GRCh38
rs746117392 Chr.16: 29993776-29993776 on Build GRCh38
rs749565027 Chr.16: 29993622-29993622 on Build GRCh38
rs758400018 Chr.16: 29993358-29993358 on Build GRCh38
rs375105900 Chr.16: 29993500-29993500 on Build GRCh38
rs199805442 Chr.16: 29993793-29993793 on Build GRCh38
rs769465348 Chr.16: 29993789-29993789 on Build GRCh38
rs961018748 Chr.16: 29993467-29993467 on Build GRCh38
rs933762950 Chr.16: 29993476-29993476 on Build GRCh38
rs775467858 Chr.16: 29993714-29993714 on Build GRCh38
rs779390363 Chr.16: 29993373-29993373 on Build GRCh38
rs764984091 Chr.16: 29993652-29993652 on Build GRCh38
rs747568307 Chr.16: 29993492-29993492 on Build GRCh38
rs201419451 Chr.16: 29993775-29993775 on Build GRCh38
rs751524757 Chr.16: 29993453-29993453 on Build GRCh38
rs746824815 Chr.16: 29993779-29993779 on Build GRCh38
rs761364612 Chr.16: 29993786-29993786 on Build GRCh38
rs769997072 Chr.16: 29993510-29993510 on Build GRCh38
rs557702327 Chr.16: 29993439-29993439 on Build GRCh38
rs759256952 Chr.16: 29993628-29993628 on Build GRCh38
rs767923220 Chr.16: 29993795-29993795 on Build GRCh38
rs199954887 Chr.16: 29993588-29993588 on Build GRCh38
rs201856578 Chr.16: 29993405-29993405 on Build GRCh38
rs750071446 Chr.16: 29993747-29993747 on Build GRCh38
rs529203258 Chr.16: 29993729-29993729 on Build GRCh38
rs375975982 Chr.16: 29993584-29993584 on Build GRCh38
rs764930949 Chr.16: 29993745-29993745 on Build GRCh38
rs372708663 Chr.16: 29993570-29993570 on Build GRCh38
rs746370797 Chr.16: 29993384-29993384 on Build GRCh38
rs756685108 Chr.16: 29993754-29993754 on Build GRCh38
rs766975064 Chr.16: 29993578-29993579 on Build GRCh38
rs775313320 Chr.16: 29993631-29993631 on Build GRCh38
rs762692953 Chr.16: 29993525-29993525 on Build GRCh38
rs772894641 Chr.16: 29993792-29993792 on Build GRCh38
rs372116326 Chr.16: 29993685-29993685 on Build GRCh38
rs760235629 Chr.16: 29993696-29993696 on Build GRCh38
rs533418828 Chr.16: 29993778-29993778 on Build GRCh38
rs369576645 Chr.16: 29993552-29993552 on Build GRCh38
rs960442778 Chr.16: 29993369-29993369 on Build GRCh38
rs978966585 Chr.16: 29993730-29993730 on Build GRCh38
rs1052011587 Chr.16: 29993451-29993451 on Build GRCh38
rs368208784 Chr.16: 29993667-29993667 on Build GRCh38
rs773387396 Chr.16: 29993614-29993614 on Build GRCh38
rs763736799 Chr.16: 29993645-29993645 on Build GRCh38
rs745629063 Chr.16: 29993709-29993709 on Build GRCh38
rs1016518433 Chr.16: 29993463-29993463 on Build GRCh38
rs372425223 Chr.16: 29993670-29993670 on Build GRCh38
rs201341216 Chr.16: 29993528-29993528 on Build GRCh38
rs1022756514 Chr.16: 29993619-29993619 on Build GRCh38
rs371440185 Chr.16: 29993598-29993598 on Build GRCh38
rs78703232 Chr.16: 29993669-29993669 on Build GRCh38
rs376150507 Chr.16: 29993534-29993534 on Build GRCh38
rs955128048 Chr.16: 29993694-29993694 on Build GRCh38
rs752187292 Chr.16: 29993550-29993550 on Build GRCh38
rs770555869 Chr.16: 29993421-29993424 on Build GRCh38
rs766259332 Chr.16: 29993447-29993447 on Build GRCh38
rs1135662 Chr.16: 29993689-29993689 on Build GRCh38
rs760192451 Chr.16: 29993634-29993634 on Build GRCh38
rs150030843 Chr.16: 29993498-29993498 on Build GRCh38
rs759191947 Chr.16: 29993559-29993560 on Build GRCh38
rs764939319 Chr.16: 29993437-29993437 on Build GRCh38
rs757352507 Chr.16: 29993774-29993774 on Build GRCh38
rs747871037 Chr.16: 29993368-29993368 on Build GRCh38
rs543484635 Chr.16: 29993713-29993713 on Build GRCh38
rs367587459 Chr.16: 29993361-29993361 on Build GRCh38
rs139374551 Chr.16: 29993468-29993468 on Build GRCh38
rs766935679 Chr.16: 29993542-29993542 on Build GRCh38

View Full Product Details