Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 54284341-54284837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRX3
Entrez Gene ID
79191
Gene Name
iroquois homeobox 3
Gene Aliases
IRX-1, IRXB1
UniGene
Hs.499205
Gene Chromosome Location
Chr.16: 54283300-54286772 on Build GRCh38
Transcripts
NM_024336.2 XM_005256139.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5798375 c.1243G>C p.G415R Chr.16: 54284638-54284638 on Build GRCh38
6216043 c.1101C>T p.P367P Chr.16: 54284780-54284780 on Build GRCh38
1189276 c.1309G>T p.G437* Chr.16: 54284572-54284572 on Build GRCh38
4972596 c.1139C>T p.P380L Chr.16: 54284742-54284742 on Build GRCh38
6053354 c.1179_1181delCGC p.A394delA Chr.16: 54284700-54284702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370099799 Chr.16: 54284367-54284367 on Build GRCh38
rs952323094 Chr.16: 54284426-54284426 on Build GRCh38
rs753493182 Chr.16: 54284624-54284624 on Build GRCh38
rs562871048 Chr.16: 54284476-54284476 on Build GRCh38
rs770113994 Chr.16: 54284771-54284771 on Build GRCh38
rs901003833 Chr.16: 54284793-54284793 on Build GRCh38
rs545518698 Chr.16: 54284597-54284597 on Build GRCh38
rs771808886 Chr.16: 54284646-54284646 on Build GRCh38
rs1053670976 Chr.16: 54284528-54284528 on Build GRCh38
rs866894525 Chr.16: 54284530-54284530 on Build GRCh38
rs762366659 Chr.16: 54284813-54284813 on Build GRCh38
rs763440889 Chr.16: 54284782-54284782 on Build GRCh38
rs764486984 Chr.16: 54284787-54284787 on Build GRCh38
rs962269374 Chr.16: 54284445-54284445 on Build GRCh38
rs753077390 Chr.16: 54284609-54284609 on Build GRCh38
rs980825274 Chr.16: 54284716-54284716 on Build GRCh38
rs909459742 Chr.16: 54284440-54284440 on Build GRCh38
rs936636946 Chr.16: 54284506-54284506 on Build GRCh38
rs778668814 Chr.16: 54284363-54284363 on Build GRCh38
rs900976990 Chr.16: 54284549-54284549 on Build GRCh38
rs540383621 Chr.16: 54284728-54284728 on Build GRCh38
rs748903656 Chr.16: 54284360-54284360 on Build GRCh38
rs945298586 Chr.16: 54284756-54284756 on Build GRCh38
rs770669069 Chr.16: 54284699-54284700 on Build GRCh38
rs949520204 Chr.16: 54284469-54284469 on Build GRCh38
rs892370899 Chr.16: 54284539-54284539 on Build GRCh38
rs528174408 Chr.16: 54284633-54284633 on Build GRCh38
rs952394474 Chr.16: 54284587-54284587 on Build GRCh38
rs778720191 Chr.16: 54284627-54284627 on Build GRCh38
rs970888743 Chr.16: 54284709-54284709 on Build GRCh38
rs201330122 Chr.16: 54284475-54284475 on Build GRCh38
rs763599592 Chr.16: 54284610-54284610 on Build GRCh38
rs996605918 Chr.16: 54284570-54284570 on Build GRCh38
rs1019395292 Chr.16: 54284386-54284386 on Build GRCh38
rs775802323 Chr.16: 54284774-54284774 on Build GRCh38
rs779557071 Chr.16: 54284623-54284623 on Build GRCh38
rs199707363 Chr.16: 54284359-54284359 on Build GRCh38
rs918033330 Chr.16: 54284464-54284464 on Build GRCh38
rs755582519 Chr.16: 54284621-54284621 on Build GRCh38
rs773827849 Chr.16: 54284700-54284702 on Build GRCh38
rs754623723 Chr.16: 54284626-54284626 on Build GRCh38
rs377057907 Chr.16: 54284546-54284546 on Build GRCh38
rs550874514 Chr.16: 54284768-54284768 on Build GRCh38
rs867243005 Chr.16: 54284648-54284648 on Build GRCh38
rs760695298 Chr.16: 54284488-54284488 on Build GRCh38
rs965481800 Chr.16: 54284401-54284401 on Build GRCh38
rs561881884 Chr.16: 54284735-54284735 on Build GRCh38
rs1028005485 Chr.16: 54284413-54284413 on Build GRCh38
rs1450355 Chr.16: 54284616-54284616 on Build GRCh38
rs764720102 Chr.16: 54284611-54284611 on Build GRCh38
rs768347037 Chr.16: 54284362-54284362 on Build GRCh38
rs774985356 Chr.16: 54284379-54284379 on Build GRCh38
rs148863611 Chr.16: 54284446-54284446 on Build GRCh38
rs529176397 Chr.16: 54284766-54284766 on Build GRCh38
rs573961039 Chr.16: 54284451-54284451 on Build GRCh38
rs574874693 Chr.16: 54284516-54284516 on Build GRCh38
rs1039688103 Chr.16: 54284620-54284620 on Build GRCh38
rs770932556 Chr.16: 54284745-54284745 on Build GRCh38
rs1009425441 Chr.16: 54284543-54284543 on Build GRCh38
rs1028033570 Chr.16: 54284571-54284571 on Build GRCh38
rs1053702691 Chr.16: 54284750-54284750 on Build GRCh38
rs759120276 Chr.16: 54284767-54284768 on Build GRCh38
rs375668113 Chr.16: 54284436-54284436 on Build GRCh38
rs773383338 Chr.16: 54284692-54284692 on Build GRCh38
rs913879357 Chr.16: 54284754-54284754 on Build GRCh38
rs774732094 Chr.16: 54284796-54284796 on Build GRCh38
rs138585447 Chr.16: 54284405-54284405 on Build GRCh38
rs375648427 Chr.16: 54284375-54284375 on Build GRCh38
rs759787704 Chr.16: 54284563-54284563 on Build GRCh38

View Full Product Details