Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 54284680-54285217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRX3
Entrez Gene ID
79191
Gene Name
iroquois homeobox 3
Gene Aliases
IRX-1, IRXB1
UniGene
Hs.499205
Gene Chromosome Location
Chr.16: 54283300-54286772 on Build GRCh38
Transcripts
NM_024336.2 XM_005256139.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3988524 c.725G>A p.G242D Chr.16: 54285156-54285156 on Build GRCh38
6216043 c.1101C>T p.P367P Chr.16: 54284780-54284780 on Build GRCh38
3932366 c.909G>A p.R303R Chr.16: 54284972-54284972 on Build GRCh38
3794912 c.765T>C p.D255D Chr.16: 54285116-54285116 on Build GRCh38
6144832 c.811G>T p.A271S Chr.16: 54285070-54285070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766657567 Chr.16: 54285176-54285177 on Build GRCh38
rs762640142 Chr.16: 54285080-54285080 on Build GRCh38
rs368665720 Chr.16: 54285100-54285100 on Build GRCh38
rs765367373 Chr.16: 54285140-54285140 on Build GRCh38
rs764893242 Chr.16: 54284865-54284865 on Build GRCh38
rs901003833 Chr.16: 54284793-54284793 on Build GRCh38
rs780322160 Chr.16: 54285101-54285101 on Build GRCh38
rs140590187 Chr.16: 54285150-54285150 on Build GRCh38
rs566912327 Chr.16: 54285059-54285059 on Build GRCh38
rs569209098 Chr.16: 54284834-54284834 on Build GRCh38
rs778100589 Chr.16: 54285127-54285127 on Build GRCh38
rs754964542 Chr.16: 54285111-54285111 on Build GRCh38
rs764486984 Chr.16: 54284787-54284787 on Build GRCh38
rs778154024 Chr.16: 54285193-54285193 on Build GRCh38
rs199931909 Chr.16: 54285134-54285134 on Build GRCh38
rs980825274 Chr.16: 54284716-54284716 on Build GRCh38
rs759825805 Chr.16: 54284974-54284974 on Build GRCh38
rs762552381 Chr.16: 54285139-54285139 on Build GRCh38
rs775859472 Chr.16: 54284910-54284910 on Build GRCh38
rs202136358 Chr.16: 54285168-54285168 on Build GRCh38
rs945298586 Chr.16: 54284756-54284756 on Build GRCh38
rs751717317 Chr.16: 54285185-54285187 on Build GRCh38
rs866655438 Chr.16: 54284860-54284860 on Build GRCh38
rs750317837 Chr.16: 54285002-54285002 on Build GRCh38
rs757184714 Chr.16: 54284898-54284898 on Build GRCh38
rs760744850 Chr.16: 54285162-54285162 on Build GRCh38
rs768893938 Chr.16: 54285035-54285035 on Build GRCh38
rs753025533 Chr.16: 54285181-54285181 on Build GRCh38
rs970888743 Chr.16: 54284709-54284709 on Build GRCh38
rs12926111 Chr.16: 54284882-54284882 on Build GRCh38
rs747585644 Chr.16: 54285130-54285130 on Build GRCh38
rs775802323 Chr.16: 54284774-54284774 on Build GRCh38
rs945355385 Chr.16: 54285175-54285175 on Build GRCh38
rs750442189 Chr.16: 54285099-54285099 on Build GRCh38
rs60493962 Chr.16: 54285084-54285084 on Build GRCh38
rs749462155 Chr.16: 54284921-54284921 on Build GRCh38
rs762269451 Chr.16: 54284926-54284926 on Build GRCh38
rs958132177 Chr.16: 54285156-54285156 on Build GRCh38
rs773777305 Chr.16: 54285050-54285050 on Build GRCh38
rs145202228 Chr.16: 54285024-54285024 on Build GRCh38
rs775127089 Chr.16: 54285122-54285124 on Build GRCh38
rs773543799 Chr.16: 54284935-54284935 on Build GRCh38
rs748254948 Chr.16: 54285116-54285116 on Build GRCh38
rs139675671 Chr.16: 54284983-54284983 on Build GRCh38
rs753712248 Chr.16: 54285162-54285164 on Build GRCh38
rs200900756 Chr.16: 54285102-54285102 on Build GRCh38
rs551531660 Chr.16: 54284925-54284925 on Build GRCh38
rs769953483 Chr.16: 54284908-54284908 on Build GRCh38
rs760761000 Chr.16: 54285132-54285137 on Build GRCh38
rs761117558 Chr.16: 54285058-54285058 on Build GRCh38
rs770932556 Chr.16: 54284745-54284745 on Build GRCh38
rs771648858 Chr.16: 54284900-54284901 on Build GRCh38
rs756275106 Chr.16: 54285005-54285005 on Build GRCh38
rs745963616 Chr.16: 54284902-54284902 on Build GRCh38
rs913879357 Chr.16: 54284754-54284754 on Build GRCh38
rs752310447 Chr.16: 54284885-54284885 on Build GRCh38
rs771402262 Chr.16: 54285133-54285133 on Build GRCh38
rs755090625 Chr.16: 54285179-54285179 on Build GRCh38
rs752363516 Chr.16: 54284951-54284951 on Build GRCh38
rs374208191 Chr.16: 54285088-54285088 on Build GRCh38
rs770113994 Chr.16: 54284771-54284771 on Build GRCh38
rs762366659 Chr.16: 54284813-54284813 on Build GRCh38
rs772285575 Chr.16: 54285045-54285045 on Build GRCh38
rs763440889 Chr.16: 54284782-54284782 on Build GRCh38
rs777569439 Chr.16: 54284897-54284897 on Build GRCh38
rs750211299 Chr.16: 54285160-54285160 on Build GRCh38
rs371146591 Chr.16: 54284945-54284945 on Build GRCh38
rs758548201 Chr.16: 54285182-54285182 on Build GRCh38
rs781216923 Chr.16: 54284991-54284991 on Build GRCh38
rs761357993 Chr.16: 54285152-54285152 on Build GRCh38
rs540383621 Chr.16: 54284728-54284728 on Build GRCh38
rs748568502 Chr.16: 54285043-54285043 on Build GRCh38
rs763903294 Chr.16: 54285132-54285134 on Build GRCh38
rs533356563 Chr.16: 54284851-54284851 on Build GRCh38
rs761463912 Chr.16: 54285174-54285175 on Build GRCh38
rs767981006 Chr.16: 54285144-54285144 on Build GRCh38
rs147225096 Chr.16: 54285119-54285119 on Build GRCh38
rs779227141 Chr.16: 54285113-54285113 on Build GRCh38
rs766444262 Chr.16: 54285165-54285165 on Build GRCh38
rs756949771 Chr.16: 54284984-54284984 on Build GRCh38
rs774946084 Chr.16: 54285072-54285072 on Build GRCh38
rs913897230 Chr.16: 54285166-54285166 on Build GRCh38
rs567541390 Chr.16: 54284929-54284929 on Build GRCh38
rs773827849 Chr.16: 54284700-54284702 on Build GRCh38
rs749587099 Chr.16: 54285032-54285032 on Build GRCh38
rs550874514 Chr.16: 54284768-54284768 on Build GRCh38
rs779197191 Chr.16: 54285040-54285040 on Build GRCh38
rs561881884 Chr.16: 54284735-54284735 on Build GRCh38
rs763831741 Chr.16: 54284973-54284973 on Build GRCh38
rs758035815 Chr.16: 54284962-54284962 on Build GRCh38
rs529176397 Chr.16: 54284766-54284766 on Build GRCh38
rs763826248 Chr.16: 54285081-54285081 on Build GRCh38
rs753358833 Chr.16: 54284845-54284845 on Build GRCh38
rs767158835 Chr.16: 54285098-54285098 on Build GRCh38
rs1053702691 Chr.16: 54284750-54284750 on Build GRCh38
rs759120276 Chr.16: 54284767-54284768 on Build GRCh38
rs759081887 Chr.16: 54284938-54284938 on Build GRCh38
rs774732094 Chr.16: 54284796-54284796 on Build GRCh38
rs758197453 Chr.16: 54284891-54284891 on Build GRCh38
rs769353676 Chr.16: 54285066-54285066 on Build GRCh38

View Full Product Details