Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57736839-57737339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNB1
Entrez Gene ID
10300
Gene Name
katanin regulatory subunit B1
Gene Aliases
KAT, LIS6
UniGene
Hs.275675
Gene Chromosome Location
Chr.16: 57735717-57757250 on Build GRCh38
Transcripts
NM_005886.2 XM_005255772.4 XM_006721121.3 XM_006721122.2 XM_006721123.3 XM_011522810.2 XM_017022860.1 XM_017022861.1 XM_017022862.1 XM_017022863.1 XM_017022864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1324387 c.29C>A p.A10D Chr.16: 57737272-57737272 on Build GRCh38
1211360 c.40C>T p.Q14* Chr.16: 57737283-57737283 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs539592764 Chr.16: 57737166-57737166 on Build GRCh38
rs777045525 Chr.16: 57737228-57737228 on Build GRCh38
rs923508087 Chr.16: 57737049-57737049 on Build GRCh38
rs190894237 Chr.16: 57737131-57737131 on Build GRCh38
rs771408060 Chr.16: 57737216-57737216 on Build GRCh38
rs575416159 Chr.16: 57736932-57736932 on Build GRCh38
rs773054342 Chr.16: 57737251-57737251 on Build GRCh38
rs755354626 Chr.16: 57737105-57737105 on Build GRCh38
rs755321066 Chr.16: 57737103-57737103 on Build GRCh38
rs907097050 Chr.16: 57737177-57737177 on Build GRCh38
rs371173167 Chr.16: 57737248-57737248 on Build GRCh38
rs558188216 Chr.16: 57737173-57737173 on Build GRCh38
rs771997237 Chr.16: 57737206-57737206 on Build GRCh38
rs138866853 Chr.16: 57737011-57737011 on Build GRCh38
rs898419934 Chr.16: 57736918-57736918 on Build GRCh38
rs1050730256 Chr.16: 57737095-57737095 on Build GRCh38
rs140697582 Chr.16: 57737258-57737258 on Build GRCh38
rs112425278 Chr.16: 57736921-57736921 on Build GRCh38
rs756084762 Chr.16: 57737002-57737002 on Build GRCh38
rs556118912 Chr.16: 57737305-57737305 on Build GRCh38
rs749084125 Chr.16: 57737036-57737036 on Build GRCh38
rs754763695 Chr.16: 57737291-57737291 on Build GRCh38
rs759826118 Chr.16: 57737229-57737229 on Build GRCh38
rs1023390697 Chr.16: 57736929-57736929 on Build GRCh38
rs550593704 Chr.16: 57737164-57737164 on Build GRCh38
rs766974907 Chr.16: 57737253-57737253 on Build GRCh38
rs146116403 Chr.16: 57736935-57736935 on Build GRCh38
rs1061546 Chr.16: 57737214-57737214 on Build GRCh38
rs762605766 Chr.16: 57737300-57737303 on Build GRCh38
rs761416699 Chr.16: 57737252-57737252 on Build GRCh38
rs974991653 Chr.16: 57737044-57737044 on Build GRCh38
rs368815039 Chr.16: 57737118-57737118 on Build GRCh38
rs1044359808 Chr.16: 57737170-57737170 on Build GRCh38
rs182606259 Chr.16: 57737165-57737165 on Build GRCh38
rs748278052 Chr.16: 57737199-57737199 on Build GRCh38
rs1038195538 Chr.16: 57736908-57736908 on Build GRCh38
rs990292804 Chr.16: 57737004-57737004 on Build GRCh38
rs573369519 Chr.16: 57737174-57737174 on Build GRCh38
rs113501175 Chr.16: 57737153-57737153 on Build GRCh38
rs114475134 Chr.16: 57737104-57737104 on Build GRCh38
rs749929354 Chr.16: 57737254-57737254 on Build GRCh38
rs770277243 Chr.16: 57737197-57737198 on Build GRCh38
rs562745763 Chr.16: 57736990-57736990 on Build GRCh38
rs939721503 Chr.16: 57737126-57737126 on Build GRCh38
rs1002827424 Chr.16: 57736938-57736938 on Build GRCh38
rs774378449 Chr.16: 57737053-57737053 on Build GRCh38
rs1004070650 Chr.16: 57736905-57736905 on Build GRCh38
rs914725111 Chr.16: 57737005-57737005 on Build GRCh38
rs375694212 Chr.16: 57737290-57737290 on Build GRCh38
rs879255518 Chr.16: 57737244-57737244 on Build GRCh38
rs1061547 Chr.16: 57737232-57737232 on Build GRCh38

View Full Product Details