Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58707440-58707945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOT2
Entrez Gene ID
2806
Gene Name
glutamic-oxaloacetic transaminase 2
Gene Aliases
KAT4, KATIV, KYAT4, mitAAT
UniGene
Hs.599470
Gene Chromosome Location
Chr.16: 58707131-58734357 on Build GRCh38
Transcripts
NM_001286220.1 NM_002080.3

SNP Detail

SNP ID Chromosome Location
rs373561929 Chr.16: 58707562-58707562 on Build GRCh38
rs772028571 Chr.16: 58707777-58707777 on Build GRCh38
rs1004921138 Chr.16: 58707584-58707584 on Build GRCh38
rs887813255 Chr.16: 58707542-58707542 on Build GRCh38
rs887762220 Chr.16: 58707773-58707773 on Build GRCh38
rs576135968 Chr.16: 58707862-58707862 on Build GRCh38
rs944484786 Chr.16: 58707719-58707719 on Build GRCh38
rs185678680 Chr.16: 58707764-58707764 on Build GRCh38
rs573870808 Chr.16: 58707765-58707765 on Build GRCh38
rs900721309 Chr.16: 58707505-58707505 on Build GRCh38
rs532786826 Chr.16: 58707834-58707834 on Build GRCh38
rs191852499 Chr.16: 58707622-58707622 on Build GRCh38
rs562723100 Chr.16: 58707793-58707793 on Build GRCh38
rs551161101 Chr.16: 58707607-58707607 on Build GRCh38
rs554628692 Chr.16: 58707807-58707807 on Build GRCh38
rs572913528 Chr.16: 58707717-58707717 on Build GRCh38
rs3208672 Chr.16: 58707665-58707665 on Build GRCh38
rs373228359 Chr.16: 58707608-58707608 on Build GRCh38
rs553780510 Chr.16: 58707593-58707593 on Build GRCh38
rs528510331 Chr.16: 58707858-58707858 on Build GRCh38
rs996402340 Chr.16: 58707506-58707506 on Build GRCh38
rs112684571 Chr.16: 58707893-58707893 on Build GRCh38
rs190556031 Chr.16: 58707883-58707883 on Build GRCh38
rs564219906 Chr.16: 58707808-58707808 on Build GRCh38
rs913000871 Chr.16: 58707718-58707718 on Build GRCh38
rs186732899 Chr.16: 58707672-58707672 on Build GRCh38
rs568730706 Chr.16: 58707518-58707518 on Build GRCh38
rs183665595 Chr.16: 58707645-58707645 on Build GRCh38
rs536223726 Chr.16: 58707801-58707801 on Build GRCh38
rs964205451 Chr.16: 58707686-58707686 on Build GRCh38
rs535708598 Chr.16: 58707578-58707578 on Build GRCh38
rs6993 Chr.16: 58707463-58707463 on Build GRCh38
rs771485679 Chr.16: 58707762-58707762 on Build GRCh38
rs1014477514 Chr.16: 58707603-58707603 on Build GRCh38
rs774486638 Chr.16: 58707652-58707652 on Build GRCh38
rs1049012149 Chr.16: 58707535-58707535 on Build GRCh38
rs555421404 Chr.16: 58707758-58707758 on Build GRCh38
rs544138368 Chr.16: 58707772-58707772 on Build GRCh38
rs1032668571 Chr.16: 58707657-58707657 on Build GRCh38
rs932229097 Chr.16: 58707759-58707759 on Build GRCh38
rs1024162214 Chr.16: 58707614-58707614 on Build GRCh38
rs896975294 Chr.16: 58707594-58707594 on Build GRCh38
rs191539697 Chr.16: 58707732-58707732 on Build GRCh38

View Full Product Details