Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58734024-58734532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOT2
Entrez Gene ID
2806
Gene Name
glutamic-oxaloacetic transaminase 2
Gene Aliases
KAT4, KATIV, KYAT4, mitAAT
UniGene
Hs.599470
Gene Chromosome Location
Chr.16: 58707131-58734357 on Build GRCh38
Transcripts
NM_001286220.1 NM_002080.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4627440 c.51C>A p.F17L Chr.16: 58734178-58734178 on Build GRCh38
6409148 c.80C>T p.A27V Chr.16: 58734149-58734149 on Build GRCh38
3510679 c.35G>A p.G12E Chr.16: 58734194-58734194 on Build GRCh38
460510 c.63C>T p.L21L Chr.16: 58734166-58734166 on Build GRCh38
6672876 c.18C>T p.S6S Chr.16: 58734211-58734211 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934843418 Chr.16: 58734421-58734421 on Build GRCh38
rs886961258 Chr.16: 58734302-58734302 on Build GRCh38
rs746422412 Chr.16: 58734250-58734250 on Build GRCh38
rs954726870 Chr.16: 58734303-58734303 on Build GRCh38
rs568318176 Chr.16: 58734135-58734135 on Build GRCh38
rs919827301 Chr.16: 58734327-58734327 on Build GRCh38
rs990751040 Chr.16: 58734424-58734424 on Build GRCh38
rs551759491 Chr.16: 58734438-58734438 on Build GRCh38
rs893139286 Chr.16: 58734089-58734089 on Build GRCh38
rs1003938020 Chr.16: 58734313-58734313 on Build GRCh38
rs894254310 Chr.16: 58734392-58734392 on Build GRCh38
rs28612351 Chr.16: 58734252-58734252 on Build GRCh38
rs1031012816 Chr.16: 58734467-58734467 on Build GRCh38
rs760486927 Chr.16: 58734265-58734265 on Build GRCh38
rs372273643 Chr.16: 58734294-58734294 on Build GRCh38
rs778088685 Chr.16: 58734130-58734130 on Build GRCh38
rs998112140 Chr.16: 58734300-58734300 on Build GRCh38
rs375471848 Chr.16: 58734247-58734247 on Build GRCh38
rs1018202206 Chr.16: 58734144-58734144 on Build GRCh38
rs1029463415 Chr.16: 58734301-58734301 on Build GRCh38
rs753825716 Chr.16: 58734171-58734171 on Build GRCh38
rs753256036 Chr.16: 58734157-58734158 on Build GRCh38
rs990375908 Chr.16: 58734123-58734123 on Build GRCh38
rs555091099 Chr.16: 58734440-58734440 on Build GRCh38
rs758641913 Chr.16: 58734118-58734118 on Build GRCh38
rs924758531 Chr.16: 58734416-58734416 on Build GRCh38
rs1055540083 Chr.16: 58734375-58734375 on Build GRCh38
rs1023088164 Chr.16: 58734357-58734357 on Build GRCh38
rs11558171 Chr.16: 58734225-58734225 on Build GRCh38
rs202160454 Chr.16: 58734269-58734269 on Build GRCh38
rs6499971 Chr.16: 58734058-58734058 on Build GRCh38
rs370301948 Chr.16: 58734362-58734362 on Build GRCh38
rs1054350373 Chr.16: 58734087-58734087 on Build GRCh38
rs962800775 Chr.16: 58734339-58734339 on Build GRCh38
rs879772577 Chr.16: 58734155-58734155 on Build GRCh38
rs760202020 Chr.16: 58734157-58734180 on Build GRCh38
rs771424682 Chr.16: 58734131-58734131 on Build GRCh38
rs775538232 Chr.16: 58734143-58734143 on Build GRCh38
rs963461857 Chr.16: 58734486-58734486 on Build GRCh38
rs903352631 Chr.16: 58734441-58734442 on Build GRCh38
rs958862072 Chr.16: 58734097-58734097 on Build GRCh38
rs747840361 Chr.16: 58734108-58734108 on Build GRCh38
rs759495822 Chr.16: 58734274-58734274 on Build GRCh38
rs776392130 Chr.16: 58734272-58734272 on Build GRCh38
rs1029540005 Chr.16: 58734162-58734162 on Build GRCh38
rs919732818 Chr.16: 58734490-58734490 on Build GRCh38
rs11558167 Chr.16: 58734246-58734246 on Build GRCh38
rs764456344 Chr.16: 58734149-58734149 on Build GRCh38
rs991169098 Chr.16: 58734343-58734343 on Build GRCh38
rs373726975 Chr.16: 58734488-58734488 on Build GRCh38
rs933873951 Chr.16: 58734505-58734505 on Build GRCh38
rs1046757625 Chr.16: 58734270-58734270 on Build GRCh38
rs916148442 Chr.16: 58734203-58734203 on Build GRCh38
rs754379149 Chr.16: 58734172-58734172 on Build GRCh38
rs529425902 Chr.16: 58734287-58734287 on Build GRCh38
rs933373283 Chr.16: 58734238-58734238 on Build GRCh38
rs1043196027 Chr.16: 58734436-58734436 on Build GRCh38
rs747362549 Chr.16: 58734223-58734223 on Build GRCh38
rs929878365 Chr.16: 58734329-58734329 on Build GRCh38
rs573700244 Chr.16: 58734136-58734136 on Build GRCh38
rs906845672 Chr.16: 58734360-58734360 on Build GRCh38
rs375771750 Chr.16: 58734182-58734182 on Build GRCh38
rs370665986 Chr.16: 58734257-58734257 on Build GRCh38
rs774769517 Chr.16: 58734260-58734260 on Build GRCh38
rs567170160 Chr.16: 58734463-58734463 on Build GRCh38
rs191250519 Chr.16: 58734475-58734475 on Build GRCh38
rs773107472 Chr.16: 58734156-58734156 on Build GRCh38
rs1021889947 Chr.16: 58734141-58734141 on Build GRCh38
rs868600549 Chr.16: 58734241-58734241 on Build GRCh38
rs560899229 Chr.16: 58734282-58734282 on Build GRCh38
rs914641800 Chr.16: 58734435-58734435 on Build GRCh38
rs781615099 Chr.16: 58734245-58734245 on Build GRCh38
rs763622983 Chr.16: 58734158-58734181 on Build GRCh38
rs942516787 Chr.16: 58734321-58734321 on Build GRCh38
rs1011749819 Chr.16: 58734403-58734403 on Build GRCh38
rs752199156 Chr.16: 58734199-58734199 on Build GRCh38
rs981660776 Chr.16: 58734415-58734415 on Build GRCh38
rs943487605 Chr.16: 58734456-58734456 on Build GRCh38
rs760609983 Chr.16: 58734165-58734165 on Build GRCh38
rs1016990322 Chr.16: 58734185-58734185 on Build GRCh38
rs906869510 Chr.16: 58734271-58734271 on Build GRCh38
rs769925491 Chr.16: 58734139-58734139 on Build GRCh38
rs993878759 Chr.16: 58734059-58734059 on Build GRCh38
rs906994196 Chr.16: 58734081-58734081 on Build GRCh38
rs935374842 Chr.16: 58734361-58734361 on Build GRCh38
rs999092666 Chr.16: 58734447-58734447 on Build GRCh38
rs562178599 Chr.16: 58734190-58734190 on Build GRCh38
rs768993241 Chr.16: 58734259-58734259 on Build GRCh38
rs116612242 Chr.16: 58734468-58734468 on Build GRCh38
rs1047427334 Chr.16: 58734351-58734351 on Build GRCh38
rs766405304 Chr.16: 58734170-58734170 on Build GRCh38
rs973970203 Chr.16: 58734322-58734322 on Build GRCh38
rs1007537512 Chr.16: 58734472-58734472 on Build GRCh38
rs962419439 Chr.16: 58734186-58734186 on Build GRCh38
rs775615027 Chr.16: 58734256-58734256 on Build GRCh38
rs374946950 Chr.16: 58734068-58734068 on Build GRCh38
rs758037631 Chr.16: 58734206-58734206 on Build GRCh38
rs763255972 Chr.16: 58734147-58734147 on Build GRCh38
rs577208712 Chr.16: 58734215-58734215 on Build GRCh38
rs1016671499 Chr.16: 58734314-58734314 on Build GRCh38
rs1003032680 Chr.16: 58734082-58734082 on Build GRCh38
rs569323509 Chr.16: 58734332-58734332 on Build GRCh38

View Full Product Details