Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75627648-75628162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KARS
Entrez Gene ID
3735
Gene Name
lysyl-tRNA synthetase
Gene Aliases
CMTRIB, DFNB89, KARS1, KARS2, KRS
UniGene
Hs.3100
Gene Chromosome Location
Chr.16: 75627724-75647687 on Build GRCh38
Transcripts
NM_001130089.1 NM_005548.2 XM_017023217.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2691479 c.1809C>T p.P603P Chr.16: 75627964-75627964 on Build GRCh38
2691477 c.1819_1821delAAG p.K607delK Chr.16: 75627952-75627954 on Build GRCh38
5367889 c.1749_1750ins10 p.T584fs*6 Chr.16: 75627939-75627940 on Build GRCh38
5367888 c.1833_1834ins10 p.T612fs*6 Chr.16: 75627939-75627940 on Build GRCh38
6690112 c.1823A>C p.E608A Chr.16: 75627950-75627950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748316661 Chr.16: 75627871-75627871 on Build GRCh38
rs767554762 Chr.16: 75628040-75628041 on Build GRCh38
rs905855783 Chr.16: 75628034-75628034 on Build GRCh38
rs1036317938 Chr.16: 75628116-75628116 on Build GRCh38
rs974402794 Chr.16: 75627882-75627882 on Build GRCh38
rs762532843 Chr.16: 75627927-75627927 on Build GRCh38
rs188921559 Chr.16: 75628014-75628014 on Build GRCh38
rs941665617 Chr.16: 75627714-75627714 on Build GRCh38
rs534800871 Chr.16: 75628127-75628128 on Build GRCh38
rs1023371022 Chr.16: 75627837-75627837 on Build GRCh38
rs549959474 Chr.16: 75627985-75627985 on Build GRCh38
rs549619745 Chr.16: 75627935-75627935 on Build GRCh38
rs751510371 Chr.16: 75627856-75627856 on Build GRCh38
rs1004981759 Chr.16: 75628138-75628138 on Build GRCh38
rs759971006 Chr.16: 75627856-75627864 on Build GRCh38
rs755761756 Chr.16: 75627900-75627900 on Build GRCh38
rs892710655 Chr.16: 75628115-75628115 on Build GRCh38
rs998945690 Chr.16: 75627795-75627795 on Build GRCh38
rs750234986 Chr.16: 75627863-75627863 on Build GRCh38
rs763797026 Chr.16: 75627929-75627929 on Build GRCh38
rs111679674 Chr.16: 75627862-75627862 on Build GRCh38
rs900391973 Chr.16: 75627738-75627738 on Build GRCh38
rs11505 Chr.16: 75627836-75627836 on Build GRCh38
rs899572544 Chr.16: 75627701-75627701 on Build GRCh38
rs933212240 Chr.16: 75627760-75627760 on Build GRCh38
rs768349236 Chr.16: 75627987-75627987 on Build GRCh38
rs144390077 Chr.16: 75627937-75627937 on Build GRCh38
rs767901895 Chr.16: 75627865-75627866 on Build GRCh38
rs768294238 Chr.16: 75627929-75627929 on Build GRCh38
rs184022101 Chr.16: 75627964-75627964 on Build GRCh38
rs983161571 Chr.16: 75627675-75627675 on Build GRCh38
rs527236481 Chr.16: 75627804-75627804 on Build GRCh38
rs892639439 Chr.16: 75627820-75627820 on Build GRCh38
rs773042139 Chr.16: 75628007-75628007 on Build GRCh38
rs756837046 Chr.16: 75627954-75627958 on Build GRCh38
rs533064131 Chr.16: 75627751-75627751 on Build GRCh38
rs771377883 Chr.16: 75627894-75627894 on Build GRCh38
rs933284596 Chr.16: 75627952-75627952 on Build GRCh38
rs1040805237 Chr.16: 75627687-75627689 on Build GRCh38
rs753346124 Chr.16: 75627952-75627954 on Build GRCh38
rs781580580 Chr.16: 75627973-75627973 on Build GRCh38
rs1035901990 Chr.16: 75627867-75627867 on Build GRCh38
rs761263838 Chr.16: 75628002-75628002 on Build GRCh38
rs766411635 Chr.16: 75628012-75628012 on Build GRCh38
rs370325473 Chr.16: 75628005-75628005 on Build GRCh38
rs1014165252 Chr.16: 75627825-75627825 on Build GRCh38
rs750338644 Chr.16: 75627944-75627944 on Build GRCh38
rs375700761 Chr.16: 75628000-75628000 on Build GRCh38
rs376960548 Chr.16: 75627873-75627873 on Build GRCh38
rs1051631761 Chr.16: 75627790-75627790 on Build GRCh38
rs902343721 Chr.16: 75627793-75627793 on Build GRCh38
rs117561624 Chr.16: 75627947-75627947 on Build GRCh38
rs148798578 Chr.16: 75627965-75627965 on Build GRCh38
rs778053495 Chr.16: 75627874-75627874 on Build GRCh38
rs747105466 Chr.16: 75627878-75627878 on Build GRCh38
rs781705880 Chr.16: 75627895-75627895 on Build GRCh38
rs778496609 Chr.16: 75628032-75628033 on Build GRCh38
rs761998891 Chr.16: 75627708-75627708 on Build GRCh38
rs1802510 Chr.16: 75627977-75627977 on Build GRCh38
rs564062535 Chr.16: 75627713-75627713 on Build GRCh38
rs555726202 Chr.16: 75628129-75628129 on Build GRCh38
rs372437633 Chr.16: 75627955-75627955 on Build GRCh38
rs551766380 Chr.16: 75627912-75627912 on Build GRCh38
rs757309903 Chr.16: 75627967-75627967 on Build GRCh38
rs761365283 Chr.16: 75627940-75627940 on Build GRCh38
rs78654839 Chr.16: 75627869-75627869 on Build GRCh38
rs983171346 Chr.16: 75627870-75627870 on Build GRCh38
rs746320017 Chr.16: 75627974-75627974 on Build GRCh38
rs535745141 Chr.16: 75628093-75628093 on Build GRCh38
rs541098749 Chr.16: 75627707-75627707 on Build GRCh38
rs1802511 Chr.16: 75627857-75627857 on Build GRCh38
rs1038939664 Chr.16: 75627731-75627731 on Build GRCh38
rs770155332 Chr.16: 75627979-75627979 on Build GRCh38
rs780454206 Chr.16: 75627982-75627982 on Build GRCh38
rs932374328 Chr.16: 75627712-75627712 on Build GRCh38
rs760726913 Chr.16: 75628010-75628010 on Build GRCh38
rs1011654982 Chr.16: 75628019-75628019 on Build GRCh38
rs1040629549 Chr.16: 75627839-75627839 on Build GRCh38
rs572117142 Chr.16: 75627688-75627688 on Build GRCh38
rs757209685 Chr.16: 75627859-75627859 on Build GRCh38
rs191584337 Chr.16: 75627907-75627907 on Build GRCh38
rs976988415 Chr.16: 75627963-75627963 on Build GRCh38
rs6834 Chr.16: 75627905-75627905 on Build GRCh38
rs753675208 Chr.16: 75627949-75627949 on Build GRCh38
rs371823244 Chr.16: 75628020-75628020 on Build GRCh38
rs753082472 Chr.16: 75627870-75627871 on Build GRCh38
rs767379297 Chr.16: 75627943-75627943 on Build GRCh38
rs533248832 Chr.16: 75627716-75627716 on Build GRCh38
rs753428124 Chr.16: 75628078-75628078 on Build GRCh38
rs893157086 Chr.16: 75627833-75627833 on Build GRCh38

View Full Product Details