Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84145172-84145660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAAF1
Entrez Gene ID
123872
Gene Name
dynein axonemal assembly factor 1
Gene Aliases
CILD13, LRRC50, ODA7
UniGene
Hs.310164
Gene Chromosome Location
Chr.16: 84145260-84178761 on Build GRCh38
Transcripts
NM_178452.5 XM_006721129.2 XM_011522853.2 XM_011522854.2 XM_011522855.2 XM_011522857.2 XM_011522858.2 XM_017022918.1 XM_017022919.1 NM_001146051.1 NM_031463.4

Gene Symbol
HSDL1
Entrez Gene ID
83693
Gene Name
hydroxysteroid dehydrogenase like 1
Gene Aliases
SDR12C3
UniGene
Hs.555992
Gene Chromosome Location
Chr.16: 84122139-84145195 on Build GRCh38
Transcripts
NM_178452.5 XM_006721129.2 XM_011522853.2 XM_011522854.2 XM_011522855.2 XM_011522857.2 XM_011522858.2 XM_017022918.1 XM_017022919.1 NM_001146051.1 NM_031463.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7304285 c.57G>A p.A19A Chr.16: 84145497-84145497 on Build GRCh38
5799966 c.61G>A p.E21K Chr.16: 84145501-84145501 on Build GRCh38
6575012 c.43G>C p.E15Q Chr.16: 84145483-84145483 on Build GRCh38
5454490 c.56C>T p.A19V Chr.16: 84145496-84145496 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753941664 Chr.16: 84145469-84145469 on Build GRCh38
rs745396262 Chr.16: 84145614-84145614 on Build GRCh38
rs1001491611 Chr.16: 84145328-84145328 on Build GRCh38
rs937216820 Chr.16: 84145546-84145546 on Build GRCh38
rs994873997 Chr.16: 84145580-84145580 on Build GRCh38
rs894588165 Chr.16: 84145618-84145618 on Build GRCh38
rs184588618 Chr.16: 84145610-84145610 on Build GRCh38
rs1024875120 Chr.16: 84145453-84145453 on Build GRCh38
rs1039603911 Chr.16: 84145280-84145280 on Build GRCh38
rs75153645 Chr.16: 84145249-84145249 on Build GRCh38
rs970588606 Chr.16: 84145632-84145632 on Build GRCh38
rs762251379 Chr.16: 84145437-84145437 on Build GRCh38
rs900572188 Chr.16: 84145198-84145198 on Build GRCh38
rs543506666 Chr.16: 84145419-84145419 on Build GRCh38
rs565199726 Chr.16: 84145513-84145513 on Build GRCh38
rs971082297 Chr.16: 84145467-84145467 on Build GRCh38
rs922071695 Chr.16: 84145416-84145416 on Build GRCh38
rs200992452 Chr.16: 84145553-84145553 on Build GRCh38
rs978338480 Chr.16: 84145264-84145264 on Build GRCh38
rs772497904 Chr.16: 84145554-84145554 on Build GRCh38
rs978186795 Chr.16: 84145316-84145316 on Build GRCh38
rs1012069717 Chr.16: 84145620-84145620 on Build GRCh38
rs1034594812 Chr.16: 84145333-84145333 on Build GRCh38
rs1020058258 Chr.16: 84145349-84145349 on Build GRCh38
rs967042626 Chr.16: 84145259-84145259 on Build GRCh38
rs777599188 Chr.16: 84145601-84145601 on Build GRCh38
rs954979646 Chr.16: 84145633-84145633 on Build GRCh38
rs973499688 Chr.16: 84145301-84145301 on Build GRCh38
rs963486644 Chr.16: 84145300-84145300 on Build GRCh38
rs959767181 Chr.16: 84145248-84145248 on Build GRCh38
rs754817465 Chr.16: 84145401-84145401 on Build GRCh38
rs775095917 Chr.16: 84145523-84145523 on Build GRCh38
rs185629224 Chr.16: 84145299-84145299 on Build GRCh38
rs370612797 Chr.16: 84145236-84145236 on Build GRCh38
rs1051715715 Chr.16: 84145211-84145211 on Build GRCh38
rs770276339 Chr.16: 84145414-84145414 on Build GRCh38
rs545356113 Chr.16: 84145486-84145486 on Build GRCh38
rs923568355 Chr.16: 84145505-84145505 on Build GRCh38
rs375586546 Chr.16: 84145576-84145576 on Build GRCh38
rs540178625 Chr.16: 84145231-84145231 on Build GRCh38
rs889730494 Chr.16: 84145189-84145189 on Build GRCh38
rs750920760 Chr.16: 84145440-84145440 on Build GRCh38
rs868661486 Chr.16: 84145447-84145447 on Build GRCh38
rs567383742 Chr.16: 84145190-84145190 on Build GRCh38
rs949080484 Chr.16: 84145242-84145242 on Build GRCh38
rs1051807366 Chr.16: 84145304-84145304 on Build GRCh38
rs1005330220 Chr.16: 84145276-84145276 on Build GRCh38
rs780631154 Chr.16: 84145533-84145533 on Build GRCh38
rs919829936 Chr.16: 84145233-84145233 on Build GRCh38
rs200309272 Chr.16: 84145600-84145600 on Build GRCh38
rs917554094 Chr.16: 84145241-84145241 on Build GRCh38
rs886052362 Chr.16: 84145295-84145295 on Build GRCh38
rs140245131 Chr.16: 84145313-84145313 on Build GRCh38
rs770885929 Chr.16: 84145532-84145532 on Build GRCh38
rs753912123 Chr.16: 84145575-84145575 on Build GRCh38
rs932149217 Chr.16: 84145547-84145547 on Build GRCh38
rs1029220059 Chr.16: 84145626-84145626 on Build GRCh38
rs371930564 Chr.16: 84145428-84145428 on Build GRCh38
rs936968411 Chr.16: 84145275-84145275 on Build GRCh38
rs570145397 Chr.16: 84145241-84145241 on Build GRCh38
rs1016609001 Chr.16: 84145230-84145230 on Build GRCh38
rs997591400 Chr.16: 84145200-84145200 on Build GRCh38
rs149813685 Chr.16: 84145357-84145358 on Build GRCh38
rs899062522 Chr.16: 84145325-84145325 on Build GRCh38
rs763903079 Chr.16: 84145571-84145571 on Build GRCh38
rs968513736 Chr.16: 84145351-84145351 on Build GRCh38
rs768258010 Chr.16: 84145538-84145555 on Build GRCh38
rs907983788 Chr.16: 84145225-84145225 on Build GRCh38
rs769486720 Chr.16: 84145425-84145425 on Build GRCh38
rs921722133 Chr.16: 84145536-84145536 on Build GRCh38
rs895654293 Chr.16: 84145340-84145340 on Build GRCh38
rs752511041 Chr.16: 84145495-84145495 on Build GRCh38
rs967144027 Chr.16: 84145367-84145367 on Build GRCh38
rs779171824 Chr.16: 84145487-84145487 on Build GRCh38
rs562677186 Chr.16: 84145334-84145334 on Build GRCh38
rs767233089 Chr.16: 84145590-84145590 on Build GRCh38
rs886052363 Chr.16: 84145300-84145300 on Build GRCh38
rs529713737 Chr.16: 84145229-84145229 on Build GRCh38
rs565768219 Chr.16: 84145258-84145258 on Build GRCh38
rs151171498 Chr.16: 84145244-84145244 on Build GRCh38
rs369739157 Chr.16: 84145451-84145451 on Build GRCh38
rs558403928 Chr.16: 84145269-84145269 on Build GRCh38
rs1053852291 Chr.16: 84145253-84145253 on Build GRCh38
rs776439760 Chr.16: 84145243-84145243 on Build GRCh38
rs369581422 Chr.16: 84145422-84145422 on Build GRCh38
rs772046071 Chr.16: 84145407-84145407 on Build GRCh38
rs745355772 Chr.16: 84145545-84145545 on Build GRCh38
rs766876325 Chr.16: 84145560-84145560 on Build GRCh38
rs923621112 Chr.16: 84145321-84145321 on Build GRCh38
rs1013979175 Chr.16: 84145341-84145341 on Build GRCh38
rs1038097546 Chr.16: 84145489-84145489 on Build GRCh38
rs193069859 Chr.16: 84145550-84145550 on Build GRCh38
rs890724632 Chr.16: 84145397-84145397 on Build GRCh38
rs886052364 Chr.16: 84145341-84145341 on Build GRCh38
rs35259968 Chr.16: 84145582-84145582 on Build GRCh38
rs373512514 Chr.16: 84145555-84145555 on Build GRCh38
rs900644721 Chr.16: 84145358-84145359 on Build GRCh38
rs886414395 Chr.16: 84145307-84145307 on Build GRCh38
rs758148388 Chr.16: 84145512-84145512 on Build GRCh38
rs957707667 Chr.16: 84145518-84145518 on Build GRCh38
rs902928373 Chr.16: 84145558-84145558 on Build GRCh38
rs991164934 Chr.16: 84145239-84145239 on Build GRCh38
rs886052365 Chr.16: 84145358-84145359 on Build GRCh38
rs201750320 Chr.16: 84145450-84145450 on Build GRCh38
rs760814451 Chr.16: 84145443-84145443 on Build GRCh38
rs368976578 Chr.16: 84145593-84145593 on Build GRCh38
rs529841659 Chr.16: 84145556-84145556 on Build GRCh38
rs778638867 Chr.16: 84145402-84145402 on Build GRCh38
rs780355675 Chr.16: 84145399-84145399 on Build GRCh38
rs983519072 Chr.16: 84145192-84145192 on Build GRCh38
rs768164076 Chr.16: 84145439-84145439 on Build GRCh38
rs1026797863 Chr.16: 84145303-84145303 on Build GRCh38
rs886446995 Chr.16: 84145227-84145227 on Build GRCh38
rs987494938 Chr.16: 84145438-84145438 on Build GRCh38
rs749406461 Chr.16: 84145400-84145400 on Build GRCh38
rs369062424 Chr.16: 84145292-84145292 on Build GRCh38
rs936399947 Chr.16: 84145354-84145354 on Build GRCh38
rs1039228443 Chr.16: 84145193-84145193 on Build GRCh38
rs375290541 Chr.16: 84145405-84145405 on Build GRCh38
rs746549741 Chr.16: 84145411-84145411 on Build GRCh38
rs890452594 Chr.16: 84145272-84145272 on Build GRCh38
rs551990619 Chr.16: 84145552-84145552 on Build GRCh38
rs1037729476 Chr.16: 84145320-84145320 on Build GRCh38
rs190497604 Chr.16: 84145375-84145375 on Build GRCh38
rs978051937 Chr.16: 84145479-84145479 on Build GRCh38
rs781169744 Chr.16: 84145612-84145612 on Build GRCh38
rs899133639 Chr.16: 84145577-84145577 on Build GRCh38
rs751348711 Chr.16: 84145603-84145603 on Build GRCh38

View Full Product Details