Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31620840-31621347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCRTR1
Entrez Gene ID
3061
Gene Name
hypocretin receptor 1
Gene Aliases
OX1R
UniGene
Hs.388226
Gene Chromosome Location
Chr.1: 31617700-31632518 on Build GRCh38
Transcripts
NM_001525.2 XM_017001105.1 XM_017001106.1 XM_017001107.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6678076 c.565C>T p.L189L Chr.1: 31621029-31621029 on Build GRCh38
4031364 c.390G>A p.V130V Chr.1: 31620854-31620854 on Build GRCh38
6842695 c.427G>A p.D143N Chr.1: 31620891-31620891 on Build GRCh38
4031366 c.515C>T p.S172L Chr.1: 31620979-31620979 on Build GRCh38
4491867 c.387C>T p.S129S Chr.1: 31620851-31620851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369797738 Chr.1: 31620920-31620920 on Build GRCh38
rs757214467 Chr.1: 31620943-31620943 on Build GRCh38
rs757082425 Chr.1: 31620885-31620885 on Build GRCh38
rs751234904 Chr.1: 31621062-31621062 on Build GRCh38
rs201146809 Chr.1: 31620888-31620888 on Build GRCh38
rs530187593 Chr.1: 31621047-31621047 on Build GRCh38
rs778794988 Chr.1: 31620894-31620894 on Build GRCh38
rs751652807 Chr.1: 31620939-31620939 on Build GRCh38
rs777757366 Chr.1: 31620878-31620878 on Build GRCh38
rs968526948 Chr.1: 31620962-31620962 on Build GRCh38
rs1004014305 Chr.1: 31620915-31620915 on Build GRCh38
rs921646358 Chr.1: 31621045-31621045 on Build GRCh38
rs895227589 Chr.1: 31621099-31621099 on Build GRCh38
rs374351024 Chr.1: 31620938-31620938 on Build GRCh38
rs896734609 Chr.1: 31621302-31621302 on Build GRCh38
rs199783403 Chr.1: 31620980-31620980 on Build GRCh38
rs2280386 Chr.1: 31621311-31621311 on Build GRCh38
rs763088630 Chr.1: 31621000-31621001 on Build GRCh38
rs771301832 Chr.1: 31621115-31621115 on Build GRCh38
rs113537439 Chr.1: 31620995-31620995 on Build GRCh38
rs746134327 Chr.1: 31621126-31621126 on Build GRCh38
rs752681555 Chr.1: 31621048-31621048 on Build GRCh38
rs746566291 Chr.1: 31620979-31620979 on Build GRCh38
rs780506804 Chr.1: 31620984-31620984 on Build GRCh38
rs1056564939 Chr.1: 31620919-31620919 on Build GRCh38
rs774531930 Chr.1: 31621121-31621121 on Build GRCh38
rs373498313 Chr.1: 31621139-31621139 on Build GRCh38
rs754534724 Chr.1: 31621071-31621071 on Build GRCh38
rs199572096 Chr.1: 31621011-31621011 on Build GRCh38
rs201018126 Chr.1: 31620865-31620865 on Build GRCh38
rs955871989 Chr.1: 31620983-31620983 on Build GRCh38
rs746839329 Chr.1: 31621147-31621147 on Build GRCh38
rs199680510 Chr.1: 31621054-31621054 on Build GRCh38
rs758173191 Chr.1: 31620952-31620952 on Build GRCh38
rs773512118 Chr.1: 31620931-31620931 on Build GRCh38
rs532441867 Chr.1: 31621141-31621141 on Build GRCh38
rs767432563 Chr.1: 31620940-31620940 on Build GRCh38
rs999706496 Chr.1: 31621150-31621150 on Build GRCh38
rs919621513 Chr.1: 31621002-31621002 on Build GRCh38
rs774639908 Chr.1: 31621012-31621012 on Build GRCh38
rs552187651 Chr.1: 31621153-31621153 on Build GRCh38
rs369830366 Chr.1: 31620905-31620905 on Build GRCh38
rs775688884 Chr.1: 31621130-31621130 on Build GRCh38
rs200128876 Chr.1: 31621129-31621129 on Build GRCh38
rs147546381 Chr.1: 31621078-31621078 on Build GRCh38
rs745537980 Chr.1: 31620899-31620899 on Build GRCh38
rs200568446 Chr.1: 31621049-31621049 on Build GRCh38
rs1023671004 Chr.1: 31620959-31620959 on Build GRCh38
rs146426534 Chr.1: 31620918-31620918 on Build GRCh38
rs777190080 Chr.1: 31620921-31620921 on Build GRCh38
rs770241325 Chr.1: 31620991-31620991 on Build GRCh38
rs779645918 Chr.1: 31621055-31621055 on Build GRCh38
rs766465690 Chr.1: 31620864-31620864 on Build GRCh38
rs202061731 Chr.1: 31620942-31620942 on Build GRCh38
rs61749523 Chr.1: 31621053-31621053 on Build GRCh38
rs982714218 Chr.1: 31621015-31621015 on Build GRCh38
rs762157022 Chr.1: 31620923-31620923 on Build GRCh38
rs749730670 Chr.1: 31621114-31621114 on Build GRCh38
rs775180214 Chr.1: 31620908-31620908 on Build GRCh38
rs373065780 Chr.1: 31621089-31621089 on Build GRCh38
rs34733098 Chr.1: 31621033-31621033 on Build GRCh38
rs963816879 Chr.1: 31621250-31621250 on Build GRCh38
rs756675185 Chr.1: 31621105-31621105 on Build GRCh38
rs200717549 Chr.1: 31621025-31621025 on Build GRCh38
rs547909963 Chr.1: 31621193-31621193 on Build GRCh38
rs1032824028 Chr.1: 31620978-31620978 on Build GRCh38
rs3134685 Chr.1: 31621233-31621233 on Build GRCh38
rs929860902 Chr.1: 31621003-31621003 on Build GRCh38
rs201925923 Chr.1: 31620904-31620904 on Build GRCh38
rs974804393 Chr.1: 31621042-31621042 on Build GRCh38
rs973843059 Chr.1: 31621288-31621288 on Build GRCh38
rs377538832 Chr.1: 31621170-31621170 on Build GRCh38
rs770155110 Chr.1: 31620929-31620929 on Build GRCh38
rs951040823 Chr.1: 31621318-31621318 on Build GRCh38
rs201877636 Chr.1: 31620871-31620871 on Build GRCh38
rs569076705 Chr.1: 31621174-31621174 on Build GRCh38
rs759622920 Chr.1: 31621018-31621018 on Build GRCh38
rs201449126 Chr.1: 31620934-31620934 on Build GRCh38
rs377035481 Chr.1: 31620875-31620875 on Build GRCh38
rs200634924 Chr.1: 31620916-31620916 on Build GRCh38
rs144603792 Chr.1: 31620963-31620963 on Build GRCh38
rs369972510 Chr.1: 31621180-31621180 on Build GRCh38
rs772154600 Chr.1: 31621127-31621127 on Build GRCh38
rs376314618 Chr.1: 31621098-31621098 on Build GRCh38
rs201629944 Chr.1: 31621107-31621107 on Build GRCh38
rs200440142 Chr.1: 31620990-31620990 on Build GRCh38
rs201910139 Chr.1: 31620948-31620948 on Build GRCh38
rs140406432 Chr.1: 31620884-31620884 on Build GRCh38
rs200191728 Chr.1: 31620949-31620949 on Build GRCh38
rs183789456 Chr.1: 31621182-31621182 on Build GRCh38

View Full Product Details