Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32316513-32317016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC1
Entrez Gene ID
3065
Gene Name
histone deacetylase 1
Gene Aliases
GON-10, HD1, RPD3, RPD3L1
UniGene
Hs.88556
Gene Chromosome Location
Chr.1: 32292103-32333628 on Build GRCh38
Transcripts
NM_004964.2 XM_011541309.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1194937 c.229C>T p.R77C Chr.1: 32316731-32316731 on Build GRCh38
2077915 c.232T>C p.S78P Chr.1: 32316734-32316734 on Build GRCh38
464440 c.258G>C p.E86D Chr.1: 32316760-32316760 on Build GRCh38
5846399 c.186delG p.E63fs*2 Chr.1: 32316688-32316688 on Build GRCh38
6387433 c.184G>A p.E62K Chr.1: 32316686-32316686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201470442 Chr.1: 32316827-32316827 on Build GRCh38
rs764004549 Chr.1: 32316801-32316801 on Build GRCh38
rs546996277 Chr.1: 32316572-32316572 on Build GRCh38
rs530792959 Chr.1: 32316918-32316918 on Build GRCh38
rs778741268 Chr.1: 32316810-32316810 on Build GRCh38
rs1036471723 Chr.1: 32316799-32316799 on Build GRCh38
rs781428753 Chr.1: 32316749-32316749 on Build GRCh38
rs757270592 Chr.1: 32316809-32316809 on Build GRCh38
rs926162497 Chr.1: 32316543-32316543 on Build GRCh38
rs904831272 Chr.1: 32316849-32316849 on Build GRCh38
rs770313371 Chr.1: 32316769-32316769 on Build GRCh38
rs748444579 Chr.1: 32316751-32316751 on Build GRCh38
rs868716923 Chr.1: 32316716-32316716 on Build GRCh38
rs200357047 Chr.1: 32316710-32316710 on Build GRCh38
rs760485212 Chr.1: 32316794-32316794 on Build GRCh38
rs932049356 Chr.1: 32316948-32316948 on Build GRCh38
rs558598651 Chr.1: 32316755-32316755 on Build GRCh38
rs765973776 Chr.1: 32316667-32316667 on Build GRCh38
rs989061794 Chr.1: 32316859-32316859 on Build GRCh38
rs773512987 Chr.1: 32316628-32316629 on Build GRCh38
rs1023347100 Chr.1: 32316818-32316818 on Build GRCh38
rs931787627 Chr.1: 32316651-32316651 on Build GRCh38
rs536258244 Chr.1: 32316633-32316633 on Build GRCh38
rs147494060 Chr.1: 32316757-32316757 on Build GRCh38
rs922016622 Chr.1: 32316941-32316941 on Build GRCh38
rs988842561 Chr.1: 32316580-32316580 on Build GRCh38
rs751422088 Chr.1: 32316993-32316993 on Build GRCh38
rs780225085 Chr.1: 32316720-32316720 on Build GRCh38
rs762129081 Chr.1: 32316969-32316969 on Build GRCh38
rs548671904 Chr.1: 32316875-32316875 on Build GRCh38
rs753690665 Chr.1: 32316805-32316805 on Build GRCh38
rs141080688 Chr.1: 32316670-32316670 on Build GRCh38
rs767396990 Chr.1: 32316678-32316678 on Build GRCh38
rs921241906 Chr.1: 32316576-32316576 on Build GRCh38
rs977158416 Chr.1: 32316550-32316550 on Build GRCh38
rs777033321 Chr.1: 32316619-32316619 on Build GRCh38
rs139067731 Chr.1: 32316681-32316681 on Build GRCh38
rs755055993 Chr.1: 32316815-32316815 on Build GRCh38
rs868265236 Chr.1: 32316771-32316771 on Build GRCh38
rs377536615 Chr.1: 32316741-32316741 on Build GRCh38
rs763586186 Chr.1: 32316766-32316766 on Build GRCh38
rs546524865 Chr.1: 32316933-32316933 on Build GRCh38
rs143436449 Chr.1: 32316644-32316644 on Build GRCh38
rs762314247 Chr.1: 32316659-32316659 on Build GRCh38
rs957592678 Chr.1: 32316546-32316546 on Build GRCh38
rs777501351 Chr.1: 32316705-32316705 on Build GRCh38
rs909041065 Chr.1: 32316981-32316981 on Build GRCh38
rs1036148135 Chr.1: 32316800-32316800 on Build GRCh38
rs752491074 Chr.1: 32316679-32316679 on Build GRCh38
rs957644810 Chr.1: 32316835-32316835 on Build GRCh38
rs576997727 Chr.1: 32316817-32316817 on Build GRCh38
rs751602920 Chr.1: 32316814-32316814 on Build GRCh38
rs913227814 Chr.1: 32316864-32316864 on Build GRCh38
rs138636514 Chr.1: 32316832-32316832 on Build GRCh38
rs1040421594 Chr.1: 32316620-32316620 on Build GRCh38
rs370709371 Chr.1: 32316821-32316821 on Build GRCh38
rs747213333 Chr.1: 32316732-32316732 on Build GRCh38
rs763449263 Chr.1: 32316783-32316784 on Build GRCh38
rs142941874 Chr.1: 32316874-32316874 on Build GRCh38
rs373563886 Chr.1: 32316822-32316822 on Build GRCh38
rs944826912 Chr.1: 32316601-32316601 on Build GRCh38
rs759301078 Chr.1: 32316671-32316671 on Build GRCh38
rs913225229 Chr.1: 32316597-32316597 on Build GRCh38
rs765694144 Chr.1: 32316641-32316641 on Build GRCh38
rs199533704 Chr.1: 32316648-32316648 on Build GRCh38
rs563389821 Chr.1: 32316903-32316903 on Build GRCh38
rs767237617 Chr.1: 32316781-32316781 on Build GRCh38
rs201518508 Chr.1: 32316830-32316830 on Build GRCh38
rs768979571 Chr.1: 32316617-32316617 on Build GRCh38
rs752398774 Chr.1: 32316791-32316791 on Build GRCh38
rs773959185 Chr.1: 32316773-32316773 on Build GRCh38
rs781690133 Chr.1: 32316984-32316984 on Build GRCh38
rs1032621615 Chr.1: 32316930-32316930 on Build GRCh38
rs759138561 Chr.1: 32316779-32316779 on Build GRCh38
rs189304755 Chr.1: 32316577-32316577 on Build GRCh38
rs145804489 Chr.1: 32316709-32316709 on Build GRCh38

View Full Product Details