Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32328779-32329300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC1
Entrez Gene ID
3065
Gene Name
histone deacetylase 1
Gene Aliases
GON-10, HD1, RPD3, RPD3L1
UniGene
Hs.88556
Gene Chromosome Location
Chr.1: 32292103-32333628 on Build GRCh38
Transcripts
NM_004964.2 XM_011541309.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5667333 c.691G>T p.G231W Chr.1: 32329122-32329122 on Build GRCh38
6678096 c.729G>A p.P243P Chr.1: 32329160-32329160 on Build GRCh38
6678095 c.690C>T p.D230D Chr.1: 32329121-32329121 on Build GRCh38
4403594 c.637-1G>A p.? Chr.1: 32329067-32329067 on Build GRCh38
4031486 c.685C>T p.R229* Chr.1: 32329116-32329116 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751524107 Chr.1: 32329204-32329204 on Build GRCh38
rs766779679 Chr.1: 32329224-32329224 on Build GRCh38
rs888702242 Chr.1: 32329275-32329275 on Build GRCh38
rs753212184 Chr.1: 32329024-32329024 on Build GRCh38
rs200968664 Chr.1: 32329181-32329181 on Build GRCh38
rs996390245 Chr.1: 32328884-32328884 on Build GRCh38
rs927134859 Chr.1: 32329040-32329040 on Build GRCh38
rs754917273 Chr.1: 32329205-32329205 on Build GRCh38
rs984887215 Chr.1: 32328937-32328937 on Build GRCh38
rs114035990 Chr.1: 32328963-32328963 on Build GRCh38
rs1015561922 Chr.1: 32328955-32328955 on Build GRCh38
rs563978052 Chr.1: 32329209-32329209 on Build GRCh38
rs755603614 Chr.1: 32329038-32329038 on Build GRCh38
rs147525948 Chr.1: 32329121-32329121 on Build GRCh38
rs183431499 Chr.1: 32328867-32328867 on Build GRCh38
rs771022207 Chr.1: 32329141-32329141 on Build GRCh38
rs935806595 Chr.1: 32329043-32329043 on Build GRCh38
rs952223346 Chr.1: 32328929-32328929 on Build GRCh38
rs372889510 Chr.1: 32329199-32329199 on Build GRCh38
rs531614495 Chr.1: 32328851-32328851 on Build GRCh38
rs993291146 Chr.1: 32328995-32328995 on Build GRCh38
rs1014984113 Chr.1: 32328960-32328960 on Build GRCh38
rs761209910 Chr.1: 32329169-32329169 on Build GRCh38
rs140106117 Chr.1: 32329163-32329163 on Build GRCh38
rs527991359 Chr.1: 32329195-32329196 on Build GRCh38
rs553868954 Chr.1: 32329269-32329269 on Build GRCh38
rs561669796 Chr.1: 32328878-32328879 on Build GRCh38
rs188558721 Chr.1: 32328889-32328889 on Build GRCh38
rs773477997 Chr.1: 32329122-32329122 on Build GRCh38
rs980437372 Chr.1: 32329227-32329227 on Build GRCh38
rs538773734 Chr.1: 32329201-32329201 on Build GRCh38
rs767842059 Chr.1: 32329018-32329018 on Build GRCh38
rs758227477 Chr.1: 32329111-32329111 on Build GRCh38
rs764741110 Chr.1: 32329197-32329197 on Build GRCh38
rs369035600 Chr.1: 32329190-32329190 on Build GRCh38
rs758124560 Chr.1: 32329202-32329202 on Build GRCh38
rs774635583 Chr.1: 32329149-32329149 on Build GRCh38
rs145372036 Chr.1: 32329116-32329116 on Build GRCh38
rs753457637 Chr.1: 32329060-32329060 on Build GRCh38
rs775015795 Chr.1: 32329094-32329094 on Build GRCh38
rs140876951 Chr.1: 32329088-32329088 on Build GRCh38
rs756907221 Chr.1: 32329073-32329073 on Build GRCh38
rs917110440 Chr.1: 32329006-32329006 on Build GRCh38
rs75753706 Chr.1: 32328846-32328846 on Build GRCh38
rs202153991 Chr.1: 32329112-32329112 on Build GRCh38
rs971706842 Chr.1: 32328983-32328983 on Build GRCh38
rs746543953 Chr.1: 32329130-32329130 on Build GRCh38
rs780763869 Chr.1: 32329255-32329255 on Build GRCh38
rs878924079 Chr.1: 32328818-32328818 on Build GRCh38
rs969392350 Chr.1: 32329210-32329210 on Build GRCh38
rs568008393 Chr.1: 32328906-32328906 on Build GRCh38
rs116148674 Chr.1: 32329133-32329133 on Build GRCh38
rs1029560500 Chr.1: 32328913-32328913 on Build GRCh38
rs917206747 Chr.1: 32328977-32328978 on Build GRCh38
rs997006628 Chr.1: 32328858-32328858 on Build GRCh38
rs373084792 Chr.1: 32329045-32329045 on Build GRCh38
rs765129121 Chr.1: 32328832-32328832 on Build GRCh38
rs376641356 Chr.1: 32329091-32329091 on Build GRCh38
rs754656322 Chr.1: 32329160-32329160 on Build GRCh38
rs139947381 Chr.1: 32329249-32329252 on Build GRCh38
rs763571199 Chr.1: 32329174-32329174 on Build GRCh38
rs763660467 Chr.1: 32329041-32329041 on Build GRCh38
rs1009024630 Chr.1: 32328828-32328828 on Build GRCh38
rs912942584 Chr.1: 32329097-32329097 on Build GRCh38
rs781311606 Chr.1: 32329206-32329206 on Build GRCh38
rs570282927 Chr.1: 32329154-32329154 on Build GRCh38
rs891583258 Chr.1: 32328826-32328826 on Build GRCh38
rs992156562 Chr.1: 32328972-32328972 on Build GRCh38
rs997050523 Chr.1: 32329233-32329233 on Build GRCh38
rs780039381 Chr.1: 32329203-32329203 on Build GRCh38

View Full Product Details