Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32331598-32332105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC1
Entrez Gene ID
3065
Gene Name
histone deacetylase 1
Gene Aliases
GON-10, HD1, RPD3, RPD3L1
UniGene
Hs.88556
Gene Chromosome Location
Chr.1: 32292103-32333628 on Build GRCh38
Transcripts
NM_004964.2 XM_011541309.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3488161 c.1103A>G p.E368G Chr.1: 32331690-32331690 on Build GRCh38
94440 c.1094G>A p.R365Q Chr.1: 32331681-32331681 on Build GRCh38
6006516 c.1168C>T p.P390S Chr.1: 32331755-32331755 on Build GRCh38
6678098 c.1211G>A p.R404H Chr.1: 32331798-32331798 on Build GRCh38
136112 c.1108C>T p.L370F Chr.1: 32331695-32331695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1140673 Chr.1: 32331795-32331795 on Build GRCh38
rs963469181 Chr.1: 32331992-32331992 on Build GRCh38
rs1043246467 Chr.1: 32331851-32331851 on Build GRCh38
rs777590609 Chr.1: 32332053-32332053 on Build GRCh38
rs937711467 Chr.1: 32331799-32331799 on Build GRCh38
rs536858701 Chr.1: 32331715-32331715 on Build GRCh38
rs199881317 Chr.1: 32331914-32331914 on Build GRCh38
rs768915375 Chr.1: 32332082-32332082 on Build GRCh38
rs768477313 Chr.1: 32331645-32331645 on Build GRCh38
rs757896216 Chr.1: 32331748-32331748 on Build GRCh38
rs751023967 Chr.1: 32331736-32331736 on Build GRCh38
rs765762274 Chr.1: 32331817-32331817 on Build GRCh38
rs148321776 Chr.1: 32331784-32331784 on Build GRCh38
rs780814077 Chr.1: 32331845-32331845 on Build GRCh38
rs373199595 Chr.1: 32331651-32331651 on Build GRCh38
rs749127792 Chr.1: 32332054-32332054 on Build GRCh38
rs1031030801 Chr.1: 32331923-32331923 on Build GRCh38
rs369693073 Chr.1: 32331833-32331833 on Build GRCh38
rs750969533 Chr.1: 32331838-32331838 on Build GRCh38
rs143313078 Chr.1: 32331770-32331770 on Build GRCh38
rs376264417 Chr.1: 32331805-32331805 on Build GRCh38
rs375198135 Chr.1: 32331749-32331749 on Build GRCh38
rs762901640 Chr.1: 32331660-32331660 on Build GRCh38
rs552958105 Chr.1: 32331778-32331778 on Build GRCh38
rs568463703 Chr.1: 32331779-32331779 on Build GRCh38
rs920541367 Chr.1: 32332040-32332040 on Build GRCh38
rs534630224 Chr.1: 32331776-32331776 on Build GRCh38
rs746551269 Chr.1: 32331751-32331751 on Build GRCh38
rs16834958 Chr.1: 32331640-32331640 on Build GRCh38
rs529626011 Chr.1: 32331638-32331638 on Build GRCh38
rs529698419 Chr.1: 32332020-32332020 on Build GRCh38
rs147675987 Chr.1: 32331712-32331712 on Build GRCh38
rs546315011 Chr.1: 32331902-32331902 on Build GRCh38
rs562218703 Chr.1: 32332013-32332013 on Build GRCh38
rs766941800 Chr.1: 32331834-32331834 on Build GRCh38
rs747001745 Chr.1: 32332071-32332071 on Build GRCh38
rs16834963 Chr.1: 32331982-32331982 on Build GRCh38
rs575846547 Chr.1: 32331883-32331883 on Build GRCh38
rs764344214 Chr.1: 32331804-32331804 on Build GRCh38
rs754533382 Chr.1: 32331844-32331844 on Build GRCh38
rs1140660 Chr.1: 32331724-32331724 on Build GRCh38
rs540939629 Chr.1: 32332009-32332009 on Build GRCh38
rs1140658 Chr.1: 32331720-32331720 on Build GRCh38
rs866639688 Chr.1: 32331959-32331959 on Build GRCh38
rs967665875 Chr.1: 32331655-32331655 on Build GRCh38
rs999727069 Chr.1: 32331864-32331864 on Build GRCh38
rs759545034 Chr.1: 32331966-32331966 on Build GRCh38
rs973872651 Chr.1: 32332001-32332001 on Build GRCh38
rs149021580 Chr.1: 32331904-32331904 on Build GRCh38
rs773149590 Chr.1: 32331667-32331667 on Build GRCh38
rs954934870 Chr.1: 32331925-32331925 on Build GRCh38
rs1010486225 Chr.1: 32331837-32331837 on Build GRCh38
rs754214092 Chr.1: 32331734-32331734 on Build GRCh38
rs535434877 Chr.1: 32331815-32331815 on Build GRCh38
rs760852226 Chr.1: 32331713-32331713 on Build GRCh38
rs919056789 Chr.1: 32331775-32331775 on Build GRCh38
rs747076698 Chr.1: 32331626-32331627 on Build GRCh38
rs893546978 Chr.1: 32331622-32331622 on Build GRCh38
rs777005876 Chr.1: 32331814-32331814 on Build GRCh38
rs551994577 Chr.1: 32331735-32331735 on Build GRCh38
rs939118121 Chr.1: 32331781-32331781 on Build GRCh38
rs767557241 Chr.1: 32331709-32331709 on Build GRCh38
rs188631841 Chr.1: 32331830-32331830 on Build GRCh38
rs112844293 Chr.1: 32331841-32331852 on Build GRCh38
rs1030225092 Chr.1: 32331903-32331903 on Build GRCh38
rs373556356 Chr.1: 32332067-32332067 on Build GRCh38
rs773437097 Chr.1: 32331800-32331800 on Build GRCh38
rs747865641 Chr.1: 32331853-32331853 on Build GRCh38
rs768154680 Chr.1: 32331763-32331763 on Build GRCh38
rs373048036 Chr.1: 32331785-32331785 on Build GRCh38
rs761628627 Chr.1: 32331653-32331653 on Build GRCh38
rs759651138 Chr.1: 32331694-32331694 on Build GRCh38
rs370426800 Chr.1: 32332052-32332052 on Build GRCh38
rs747885612 Chr.1: 32331769-32331769 on Build GRCh38
rs774348200 Chr.1: 32332068-32332068 on Build GRCh38
rs369560330 Chr.1: 32331782-32331782 on Build GRCh38
rs769656946 Chr.1: 32331662-32331662 on Build GRCh38
rs999015581 Chr.1: 32331908-32331908 on Build GRCh38

View Full Product Details