Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35442799-35443300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0319L
Entrez Gene ID
79932
Gene Name
KIAA0319 like
Gene Aliases
AAVR
UniGene
Hs.456507
Gene Chromosome Location
Chr.1: 35433490-35557670 on Build GRCh38
Transcripts
NM_024874.4 XM_006710907.1 XM_006710909.1 XM_006710910.1 XM_011542179.2 XM_011542180.1 XM_017002366.1 XM_017002367.1 XM_017002368.1 XM_017002369.1 XM_017002370.1 XM_017002371.1 XM_017002372.1 XM_017002374.1 XM_017002375.1 XM_017002376.1 XM_017002377.1 XM_017002378.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
426162 c.2706A>G p.K902K Chr.1: 35442979-35442979 on Build GRCh38
4539284 c.2667G>A p.L889L Chr.1: 35443018-35443018 on Build GRCh38
1667391 c.2683G>A p.G895S Chr.1: 35443002-35443002 on Build GRCh38
1342142 c.2728delT p.W910fs*10 Chr.1: 35442957-35442957 on Build GRCh38
3377149 c.2707C>T p.R903C Chr.1: 35442978-35442978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945677750 Chr.1: 35443116-35443116 on Build GRCh38
rs763491469 Chr.1: 35443073-35443073 on Build GRCh38
rs1041825525 Chr.1: 35443134-35443134 on Build GRCh38
rs773777661 Chr.1: 35442887-35442887 on Build GRCh38
rs775829003 Chr.1: 35442866-35442866 on Build GRCh38
rs369842981 Chr.1: 35442894-35442894 on Build GRCh38
rs142784644 Chr.1: 35442976-35442976 on Build GRCh38
rs936027666 Chr.1: 35443166-35443166 on Build GRCh38
rs889719497 Chr.1: 35443210-35443210 on Build GRCh38
rs201963527 Chr.1: 35442870-35442870 on Build GRCh38
rs766878879 Chr.1: 35442934-35442934 on Build GRCh38
rs1016929083 Chr.1: 35442858-35442858 on Build GRCh38
rs372512432 Chr.1: 35443061-35443061 on Build GRCh38
rs370622628 Chr.1: 35442977-35442977 on Build GRCh38
rs1005795638 Chr.1: 35443229-35443229 on Build GRCh38
rs891723245 Chr.1: 35443089-35443089 on Build GRCh38
rs935901769 Chr.1: 35443048-35443048 on Build GRCh38
rs770904848 Chr.1: 35442862-35442862 on Build GRCh38
rs749088262 Chr.1: 35442859-35442859 on Build GRCh38
rs763292081 Chr.1: 35442888-35442888 on Build GRCh38
rs762171587 Chr.1: 35442877-35442877 on Build GRCh38
rs768939254 Chr.1: 35443023-35443023 on Build GRCh38
rs754682422 Chr.1: 35442925-35442925 on Build GRCh38
rs778877216 Chr.1: 35443006-35443006 on Build GRCh38
rs567988636 Chr.1: 35443113-35443113 on Build GRCh38
rs748257786 Chr.1: 35443060-35443060 on Build GRCh38
rs1049921708 Chr.1: 35443214-35443214 on Build GRCh38
rs758385655 Chr.1: 35442879-35442879 on Build GRCh38
rs2275247 Chr.1: 35442850-35442850 on Build GRCh38
rs983793428 Chr.1: 35443055-35443055 on Build GRCh38
rs755645264 Chr.1: 35442972-35442972 on Build GRCh38
rs1040041149 Chr.1: 35443131-35443131 on Build GRCh38
rs768582106 Chr.1: 35442868-35442868 on Build GRCh38
rs576352244 Chr.1: 35443227-35443227 on Build GRCh38
rs751183160 Chr.1: 35442923-35442923 on Build GRCh38
rs139725127 Chr.1: 35443009-35443009 on Build GRCh38
rs201235642 Chr.1: 35443074-35443074 on Build GRCh38
rs747270260 Chr.1: 35442867-35442867 on Build GRCh38
rs556874446 Chr.1: 35443187-35443187 on Build GRCh38
rs1050419862 Chr.1: 35443171-35443171 on Build GRCh38
rs535344057 Chr.1: 35443149-35443149 on Build GRCh38
rs757083721 Chr.1: 35442989-35442989 on Build GRCh38
rs946211588 Chr.1: 35443129-35443129 on Build GRCh38
rs954266750 Chr.1: 35442847-35442847 on Build GRCh38
rs752255004 Chr.1: 35442958-35442958 on Build GRCh38
rs146388394 Chr.1: 35442926-35442926 on Build GRCh38
rs889695820 Chr.1: 35442873-35442873 on Build GRCh38
rs774710952 Chr.1: 35443076-35443076 on Build GRCh38
rs747154996 Chr.1: 35443008-35443008 on Build GRCh38
rs1017171142 Chr.1: 35443262-35443262 on Build GRCh38
rs979870897 Chr.1: 35442982-35442982 on Build GRCh38
rs748212087 Chr.1: 35443045-35443045 on Build GRCh38
rs773687899 Chr.1: 35443072-35443072 on Build GRCh38
rs904556928 Chr.1: 35442824-35442824 on Build GRCh38

View Full Product Details