Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60043112-60043619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEATR6
Entrez Gene ID
63897
Gene Name
HEAT repeat containing 6
Gene Aliases
ABC1
UniGene
Hs.221899
Gene Chromosome Location
Chr.17: 60041363-60078931 on Build GRCh38
Transcripts
NM_022070.4 XM_005257570.3 XM_011525110.2 XM_011525111.2 XM_017024936.1 XM_017024937.1 XM_017024938.1 XM_017024939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6901176 c.3526C>T p.P1176S Chr.17: 60043583-60043583 on Build GRCh38
7242118 c.3528A>T p.P1176P Chr.17: 60043581-60043581 on Build GRCh38
982132 c.3507T>G p.S1169S Chr.17: 60043602-60043602 on Build GRCh38
5853130 c.3523C>T p.L1175F Chr.17: 60043586-60043586 on Build GRCh38
5578697 c.3512C>T p.S1171L Chr.17: 60043597-60043597 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762569216 Chr.17: 60043575-60043575 on Build GRCh38
rs189032278 Chr.17: 60043207-60043207 on Build GRCh38
rs757931443 Chr.17: 60043519-60043519 on Build GRCh38
rs935455181 Chr.17: 60043321-60043321 on Build GRCh38
rs960045732 Chr.17: 60043411-60043411 on Build GRCh38
rs142900671 Chr.17: 60043226-60043226 on Build GRCh38
rs1011406912 Chr.17: 60043389-60043389 on Build GRCh38
rs977142063 Chr.17: 60043505-60043505 on Build GRCh38
rs772444281 Chr.17: 60043558-60043558 on Build GRCh38
rs556440089 Chr.17: 60043273-60043273 on Build GRCh38
rs922677587 Chr.17: 60043307-60043307 on Build GRCh38
rs952549369 Chr.17: 60043507-60043507 on Build GRCh38
rs180835915 Chr.17: 60043250-60043250 on Build GRCh38
rs964368325 Chr.17: 60043483-60043483 on Build GRCh38
rs571183486 Chr.17: 60043528-60043528 on Build GRCh38
rs375255882 Chr.17: 60043268-60043268 on Build GRCh38
rs923230890 Chr.17: 60043306-60043307 on Build GRCh38
rs891646732 Chr.17: 60043342-60043342 on Build GRCh38
rs554314301 Chr.17: 60043412-60043412 on Build GRCh38
rs1005339196 Chr.17: 60043514-60043514 on Build GRCh38
rs755892944 Chr.17: 60043594-60043594 on Build GRCh38
rs781111870 Chr.17: 60043531-60043531 on Build GRCh38
rs1021488711 Chr.17: 60043170-60043170 on Build GRCh38
rs778248572 Chr.17: 60043592-60043593 on Build GRCh38
rs1035839583 Chr.17: 60043159-60043159 on Build GRCh38
rs1035385502 Chr.17: 60043408-60043408 on Build GRCh38
rs147940060 Chr.17: 60043570-60043570 on Build GRCh38
rs562782641 Chr.17: 60043572-60043572 on Build GRCh38
rs116863705 Chr.17: 60043298-60043298 on Build GRCh38
rs141770217 Chr.17: 60043588-60043588 on Build GRCh38
rs558403592 Chr.17: 60043593-60043593 on Build GRCh38
rs975001910 Chr.17: 60043301-60043301 on Build GRCh38
rs959855692 Chr.17: 60043175-60043175 on Build GRCh38
rs1001605364 Chr.17: 60043407-60043407 on Build GRCh38
rs750094327 Chr.17: 60043574-60043574 on Build GRCh38
rs952861224 Chr.17: 60043454-60043454 on Build GRCh38
rs970129742 Chr.17: 60043134-60043134 on Build GRCh38
rs984391242 Chr.17: 60043260-60043260 on Build GRCh38
rs773654641 Chr.17: 60043504-60043504 on Build GRCh38
rs201811006 Chr.17: 60043529-60043529 on Build GRCh38
rs979842447 Chr.17: 60043151-60043151 on Build GRCh38
rs905813822 Chr.17: 60043392-60043392 on Build GRCh38
rs150797746 Chr.17: 60043254-60043254 on Build GRCh38
rs913294823 Chr.17: 60043339-60043339 on Build GRCh38
rs200810311 Chr.17: 60043536-60043536 on Build GRCh38
rs375867425 Chr.17: 60043545-60043545 on Build GRCh38
rs933327966 Chr.17: 60043317-60043317 on Build GRCh38
rs5821289 Chr.17: 60043592-60043592 on Build GRCh38
rs751067048 Chr.17: 60043582-60043582 on Build GRCh38
rs1042960443 Chr.17: 60043391-60043391 on Build GRCh38
rs1043280387 Chr.17: 60043366-60043366 on Build GRCh38
rs139162824 Chr.17: 60043463-60043463 on Build GRCh38
rs370995532 Chr.17: 60043517-60043517 on Build GRCh38
rs774552842 Chr.17: 60043549-60043549 on Build GRCh38
rs542310568 Chr.17: 60043323-60043323 on Build GRCh38
rs745390730 Chr.17: 60043593-60043594 on Build GRCh38
rs1035310209 Chr.17: 60043205-60043205 on Build GRCh38
rs77648163 Chr.17: 60043464-60043464 on Build GRCh38
rs201186000 Chr.17: 60043264-60043264 on Build GRCh38
rs1052853449 Chr.17: 60043331-60043331 on Build GRCh38
rs760952437 Chr.17: 60043571-60043571 on Build GRCh38
rs950882154 Chr.17: 60043263-60043264 on Build GRCh38
rs749482636 Chr.17: 60043537-60043537 on Build GRCh38
rs755002788 Chr.17: 60043587-60043587 on Build GRCh38
rs959706778 Chr.17: 60043210-60043210 on Build GRCh38
rs942704714 Chr.17: 60043290-60043290 on Build GRCh38
rs765911274 Chr.17: 60043579-60043579 on Build GRCh38
rs577133638 Chr.17: 60043530-60043530 on Build GRCh38
rs1025795279 Chr.17: 60043436-60043436 on Build GRCh38
rs756958988 Chr.17: 60043589-60043591 on Build GRCh38
rs538273826 Chr.17: 60043580-60043580 on Build GRCh38
rs1056979182 Chr.17: 60043437-60043437 on Build GRCh38
rs905758992 Chr.17: 60043202-60043202 on Build GRCh38
rs747112353 Chr.17: 60043240-60043240 on Build GRCh38
rs145634926 Chr.17: 60043591-60043591 on Build GRCh38
rs574852115 Chr.17: 60043291-60043291 on Build GRCh38
rs973293357 Chr.17: 60043204-60043204 on Build GRCh38

View Full Product Details