Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44926268-44926775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF18B
Entrez Gene ID
146909
Gene Name
kinesin family member 18B
Gene Aliases
-
UniGene
Hs.135094
Gene Chromosome Location
Chr.17: 44924711-44947746 on Build GRCh38
Transcripts
NM_001264573.1 NM_001265577.1 XM_011524385.2 XM_011524386.2 XM_011524387.2 XM_011524388.2 XM_011524389.2 XM_011524390.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4267753 c.2461G>A p.A821T Chr.17: 44926468-44926468 on Build GRCh38
4267751 c.2494G>T p.A832S Chr.17: 44926435-44926435 on Build GRCh38
6289126 c.2474C>T p.P825L Chr.17: 44926428-44926428 on Build GRCh38
6855583 c.2430-1G>A p.? Chr.17: 44926500-44926500 on Build GRCh38
6855582 c.2403-1G>A p.? Chr.17: 44926500-44926500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777523695 Chr.17: 44926465-44926465 on Build GRCh38
rs752531561 Chr.17: 44926344-44926344 on Build GRCh38
rs906465104 Chr.17: 44926336-44926336 on Build GRCh38
rs748052207 Chr.17: 44926480-44926480 on Build GRCh38
rs1017240593 Chr.17: 44926706-44926706 on Build GRCh38
rs147949432 Chr.17: 44926596-44926596 on Build GRCh38
rs530427949 Chr.17: 44926406-44926406 on Build GRCh38
rs1011761528 Chr.17: 44926618-44926618 on Build GRCh38
rs1022058683 Chr.17: 44926355-44926355 on Build GRCh38
rs1028583029 Chr.17: 44926713-44926713 on Build GRCh38
rs1053320022 Chr.17: 44926338-44926338 on Build GRCh38
rs974433078 Chr.17: 44926710-44926710 on Build GRCh38
rs535728982 Chr.17: 44926730-44926730 on Build GRCh38
rs1022055795 Chr.17: 44926624-44926624 on Build GRCh38
rs932846230 Chr.17: 44926474-44926474 on Build GRCh38
rs756350303 Chr.17: 44926469-44926469 on Build GRCh38
rs1019001962 Chr.17: 44926420-44926420 on Build GRCh38
rs763430749 Chr.17: 44926536-44926536 on Build GRCh38
rs1018684473 Chr.17: 44926676-44926676 on Build GRCh38
rs528078424 Chr.17: 44926314-44926314 on Build GRCh38
rs745509172 Chr.17: 44926553-44926553 on Build GRCh38
rs999705145 Chr.17: 44926669-44926669 on Build GRCh38
rs530608581 Chr.17: 44926473-44926473 on Build GRCh38
rs778679211 Chr.17: 44926310-44926310 on Build GRCh38
rs370048495 Chr.17: 44926371-44926371 on Build GRCh38
rs997401408 Chr.17: 44926724-44926724 on Build GRCh38
rs375780925 Chr.17: 44926396-44926396 on Build GRCh38
rs902288849 Chr.17: 44926635-44926635 on Build GRCh38
rs764725929 Chr.17: 44926409-44926409 on Build GRCh38
rs142466047 Chr.17: 44926333-44926333 on Build GRCh38
rs1043490089 Chr.17: 44926659-44926659 on Build GRCh38
rs370785118 Chr.17: 44926404-44926404 on Build GRCh38
rs767962830 Chr.17: 44926541-44926541 on Build GRCh38
rs763175234 Chr.17: 44926717-44926717 on Build GRCh38
rs373656052 Chr.17: 44926534-44926534 on Build GRCh38
rs373883057 Chr.17: 44926388-44926388 on Build GRCh38
rs966136622 Chr.17: 44926680-44926680 on Build GRCh38
rs9911406 Chr.17: 44926538-44926538 on Build GRCh38
rs370083146 Chr.17: 44926533-44926533 on Build GRCh38
rs752926049 Chr.17: 44926369-44926369 on Build GRCh38
rs758335577 Chr.17: 44926345-44926345 on Build GRCh38
rs1022002069 Chr.17: 44926398-44926398 on Build GRCh38
rs771472222 Chr.17: 44926370-44926370 on Build GRCh38
rs553333308 Chr.17: 44926696-44926696 on Build GRCh38
rs888530686 Chr.17: 44926298-44926298 on Build GRCh38
rs553257243 Chr.17: 44926576-44926576 on Build GRCh38
rs750551514 Chr.17: 44926542-44926542 on Build GRCh38
rs760714984 Chr.17: 44926544-44926544 on Build GRCh38
rs771981588 Chr.17: 44926484-44926484 on Build GRCh38
rs9890538 Chr.17: 44926539-44926539 on Build GRCh38
rs386797325 Chr.17: 44926538-44926539 on Build GRCh38
rs1017939510 Chr.17: 44926372-44926372 on Build GRCh38
rs909978157 Chr.17: 44926509-44926509 on Build GRCh38
rs778968636 Chr.17: 44926479-44926479 on Build GRCh38
rs910029789 Chr.17: 44926736-44926736 on Build GRCh38
rs940008066 Chr.17: 44926309-44926309 on Build GRCh38
rs750462237 Chr.17: 44926468-44926468 on Build GRCh38
rs751994698 Chr.17: 44926450-44926450 on Build GRCh38
rs780233602 Chr.17: 44926471-44926471 on Build GRCh38
rs866321662 Chr.17: 44926378-44926378 on Build GRCh38
rs551615251 Chr.17: 44926726-44926726 on Build GRCh38
rs569908162 Chr.17: 44926522-44926522 on Build GRCh38
rs961521428 Chr.17: 44926744-44926744 on Build GRCh38
rs1007216087 Chr.17: 44926687-44926687 on Build GRCh38
rs762321612 Chr.17: 44926464-44926464 on Build GRCh38
rs891972314 Chr.17: 44926342-44926342 on Build GRCh38
rs775974061 Chr.17: 44926610-44926610 on Build GRCh38
rs35989381 Chr.17: 44926572-44926573 on Build GRCh38
rs895015365 Chr.17: 44926386-44926386 on Build GRCh38
rs570625419 Chr.17: 44926574-44926574 on Build GRCh38
rs1037409129 Chr.17: 44926318-44926318 on Build GRCh38
rs548456063 Chr.17: 44926449-44926449 on Build GRCh38
rs375991166 Chr.17: 44926537-44926537 on Build GRCh38
rs766654600 Chr.17: 44926546-44926546 on Build GRCh38
rs777459641 Chr.17: 44926512-44926512 on Build GRCh38
rs746472947 Chr.17: 44926529-44926529 on Build GRCh38

View Full Product Details