Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44927943-44928438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF18B
Entrez Gene ID
146909
Gene Name
kinesin family member 18B
Gene Aliases
-
UniGene
Hs.135094
Gene Chromosome Location
Chr.17: 44924711-44947746 on Build GRCh38
Transcripts
NM_001264573.1 NM_001265577.1 XM_011524385.2 XM_011524386.2 XM_011524387.2 XM_011524388.2 XM_011524389.2 XM_011524390.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7259911 c.2006G>A p.C669Y Chr.17: 44928332-44928332 on Build GRCh38
3518181 c.2093G>C p.G698A Chr.17: 44928272-44928272 on Build GRCh38
4267768 c.2141C>T p.T714I Chr.17: 44928224-44928224 on Build GRCh38
6996607 c.2111C>T p.P704L Chr.17: 44928254-44928254 on Build GRCh38
4949098 c.2215C>T p.R739* Chr.17: 44928150-44928150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918183393 Chr.17: 44928381-44928381 on Build GRCh38
rs895601788 Chr.17: 44928102-44928102 on Build GRCh38
rs60374269 Chr.17: 44928354-44928354 on Build GRCh38
rs573673245 Chr.17: 44928269-44928269 on Build GRCh38
rs751358473 Chr.17: 44928037-44928037 on Build GRCh38
rs749349815 Chr.17: 44928255-44928255 on Build GRCh38
rs746403821 Chr.17: 44928240-44928240 on Build GRCh38
rs528725188 Chr.17: 44927991-44927991 on Build GRCh38
rs748528579 Chr.17: 44927986-44927986 on Build GRCh38
rs535808286 Chr.17: 44928094-44928094 on Build GRCh38
rs757281877 Chr.17: 44928050-44928050 on Build GRCh38
rs971873138 Chr.17: 44928294-44928294 on Build GRCh38
rs756421332 Chr.17: 44928414-44928414 on Build GRCh38
rs767466764 Chr.17: 44928052-44928052 on Build GRCh38
rs936046222 Chr.17: 44928351-44928351 on Build GRCh38
rs760473198 Chr.17: 44928205-44928205 on Build GRCh38
rs747520725 Chr.17: 44928000-44928000 on Build GRCh38
rs750361278 Chr.17: 44928204-44928204 on Build GRCh38
rs772611175 Chr.17: 44928076-44928076 on Build GRCh38
rs922738524 Chr.17: 44928001-44928001 on Build GRCh38
rs897519377 Chr.17: 44928073-44928073 on Build GRCh38
rs200544188 Chr.17: 44928237-44928237 on Build GRCh38
rs375235843 Chr.17: 44928288-44928288 on Build GRCh38
rs376484406 Chr.17: 44928150-44928150 on Build GRCh38
rs931802860 Chr.17: 44928023-44928023 on Build GRCh38
rs766478464 Chr.17: 44928332-44928332 on Build GRCh38
rs200526914 Chr.17: 44928287-44928287 on Build GRCh38
rs776599934 Chr.17: 44928008-44928008 on Build GRCh38
rs759406754 Chr.17: 44928380-44928380 on Build GRCh38
rs373300269 Chr.17: 44928226-44928226 on Build GRCh38
rs779631321 Chr.17: 44928067-44928067 on Build GRCh38
rs775764457 Chr.17: 44928021-44928021 on Build GRCh38
rs771389990 Chr.17: 44928005-44928005 on Build GRCh38
rs759020657 Chr.17: 44928283-44928283 on Build GRCh38
rs755104339 Chr.17: 44928356-44928356 on Build GRCh38
rs985925274 Chr.17: 44928026-44928026 on Build GRCh38
rs750785050 Chr.17: 44928039-44928039 on Build GRCh38
rs770901247 Chr.17: 44928371-44928371 on Build GRCh38
rs964674709 Chr.17: 44927975-44927976 on Build GRCh38
rs546084222 Chr.17: 44928357-44928357 on Build GRCh38
rs762699986 Chr.17: 44928402-44928402 on Build GRCh38
rs773509785 Chr.17: 44927997-44927997 on Build GRCh38
rs776532109 Chr.17: 44928374-44928374 on Build GRCh38
rs529539002 Chr.17: 44928070-44928070 on Build GRCh38
rs561983834 Chr.17: 44928051-44928051 on Build GRCh38
rs375515468 Chr.17: 44928262-44928262 on Build GRCh38
rs781349464 Chr.17: 44928239-44928239 on Build GRCh38
rs371805837 Chr.17: 44928368-44928368 on Build GRCh38
rs200703182 Chr.17: 44928166-44928166 on Build GRCh38
rs200684437 Chr.17: 44928189-44928189 on Build GRCh38
rs557711997 Chr.17: 44928210-44928210 on Build GRCh38
rs774935576 Chr.17: 44928398-44928398 on Build GRCh38
rs369709926 Chr.17: 44928186-44928186 on Build GRCh38
rs763624751 Chr.17: 44928403-44928403 on Build GRCh38
rs150206890 Chr.17: 44928216-44928216 on Build GRCh38
rs774702151 Chr.17: 44928258-44928258 on Build GRCh38
rs201358373 Chr.17: 44928238-44928238 on Build GRCh38
rs759643023 Chr.17: 44928011-44928011 on Build GRCh38
rs568495575 Chr.17: 44928088-44928088 on Build GRCh38
rs202002436 Chr.17: 44928278-44928278 on Build GRCh38
rs375683493 Chr.17: 44928009-44928009 on Build GRCh38
rs35432916 Chr.17: 44928397-44928398 on Build GRCh38
rs768948449 Chr.17: 44928178-44928178 on Build GRCh38
rs778143841 Chr.17: 44928132-44928132 on Build GRCh38
rs745989827 Chr.17: 44928155-44928155 on Build GRCh38
rs746748162 Chr.17: 44928369-44928369 on Build GRCh38
rs866587541 Chr.17: 44928272-44928272 on Build GRCh38
rs993143641 Chr.17: 44928276-44928277 on Build GRCh38
rs781639772 Chr.17: 44928290-44928290 on Build GRCh38
rs775362348 Chr.17: 44928167-44928167 on Build GRCh38
rs760682716 Chr.17: 44928277-44928277 on Build GRCh38
rs764743878 Chr.17: 44928388-44928388 on Build GRCh38
rs371129096 Chr.17: 44928350-44928350 on Build GRCh38
rs951624385 Chr.17: 44928222-44928222 on Build GRCh38
rs749196600 Chr.17: 44928220-44928221 on Build GRCh38
rs763982561 Chr.17: 44928033-44928033 on Build GRCh38
rs763790347 Chr.17: 44928404-44928404 on Build GRCh38
rs868670770 Chr.17: 44928198-44928198 on Build GRCh38
rs763296734 Chr.17: 44928172-44928172 on Build GRCh38
rs749862792 Chr.17: 44928263-44928263 on Build GRCh38
rs371854103 Chr.17: 44928260-44928260 on Build GRCh38
rs763207521 Chr.17: 44928031-44928031 on Build GRCh38
rs772824105 Chr.17: 44928273-44928273 on Build GRCh38
rs369728072 Chr.17: 44928014-44928014 on Build GRCh38
rs757115320 Chr.17: 44928223-44928223 on Build GRCh38
rs752355477 Chr.17: 44928289-44928289 on Build GRCh38
rs770153757 Chr.17: 44928254-44928254 on Build GRCh38
rs759820172 Chr.17: 44928364-44928364 on Build GRCh38
rs747328006 Chr.17: 44928140-44928140 on Build GRCh38
rs199693894 Chr.17: 44928190-44928190 on Build GRCh38
rs772134493 Chr.17: 44928367-44928367 on Build GRCh38
rs373529123 Chr.17: 44928057-44928057 on Build GRCh38
rs753384060 Chr.17: 44928280-44928280 on Build GRCh38
rs908974221 Chr.17: 44928043-44928043 on Build GRCh38
rs1056756420 Chr.17: 44928092-44928092 on Build GRCh38
rs755645107 Chr.17: 44928058-44928058 on Build GRCh38

View Full Product Details