Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45149154-45149663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXIM1
Entrez Gene ID
10614
Gene Name
hexamethylene bisacetamide inducible 1
Gene Aliases
CLP1, EDG1, HIS1, MAQ1
UniGene
Hs.586945
Gene Chromosome Location
Chr.17: 45147317-45152101 on Build GRCh38
Transcripts
NM_006460.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3755587 c.330A>G p.E110E Chr.17: 45149520-45149520 on Build GRCh38
3958439 c.130G>C p.D44H Chr.17: 45149320-45149320 on Build GRCh38
3518227 c.136A>C p.R46R Chr.17: 45149326-45149326 on Build GRCh38
3518228 c.231G>A p.Q77Q Chr.17: 45149421-45149421 on Build GRCh38
6226647 c.91G>C p.E31Q Chr.17: 45149281-45149281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746508181 Chr.17: 45149450-45149450 on Build GRCh38
rs755618582 Chr.17: 45149635-45149635 on Build GRCh38
rs774042737 Chr.17: 45149284-45149284 on Build GRCh38
rs371225086 Chr.17: 45149181-45149181 on Build GRCh38
rs765719338 Chr.17: 45149216-45149216 on Build GRCh38
rs770594246 Chr.17: 45149451-45149451 on Build GRCh38
rs751673575 Chr.17: 45149377-45149377 on Build GRCh38
rs753806975 Chr.17: 45149419-45149419 on Build GRCh38
rs536393664 Chr.17: 45149439-45149439 on Build GRCh38
rs149955210 Chr.17: 45149308-45149308 on Build GRCh38
rs1024700122 Chr.17: 45149402-45149402 on Build GRCh38
rs147398972 Chr.17: 45149298-45149298 on Build GRCh38
rs767694506 Chr.17: 45149387-45149387 on Build GRCh38
rs756767765 Chr.17: 45149524-45149524 on Build GRCh38
rs779938632 Chr.17: 45149418-45149418 on Build GRCh38
rs957519217 Chr.17: 45149445-45149445 on Build GRCh38
rs201788609 Chr.17: 45149455-45149455 on Build GRCh38
rs761200584 Chr.17: 45149286-45149286 on Build GRCh38
rs767170073 Chr.17: 45149285-45149286 on Build GRCh38
rs754722145 Chr.17: 45149424-45149424 on Build GRCh38
rs766471661 Chr.17: 45149496-45149496 on Build GRCh38
rs760708795 Chr.17: 45149491-45149491 on Build GRCh38
rs903988968 Chr.17: 45149314-45149314 on Build GRCh38
rs370597507 Chr.17: 45149279-45149279 on Build GRCh38
rs994661450 Chr.17: 45149311-45149311 on Build GRCh38
rs1039487768 Chr.17: 45149346-45149346 on Build GRCh38
rs754918424 Chr.17: 45149508-45149508 on Build GRCh38
rs749146234 Chr.17: 45149271-45149271 on Build GRCh38
rs776691576 Chr.17: 45149566-45149566 on Build GRCh38
rs11558627 Chr.17: 45149352-45149352 on Build GRCh38
rs369049036 Chr.17: 45149592-45149592 on Build GRCh38
rs775152336 Chr.17: 45149538-45149538 on Build GRCh38
rs975607932 Chr.17: 45149452-45149452 on Build GRCh38
rs756233346 Chr.17: 45149415-45149415 on Build GRCh38
rs573692319 Chr.17: 45149602-45149602 on Build GRCh38
rs898947076 Chr.17: 45149552-45149552 on Build GRCh38
rs777149441 Chr.17: 45149286-45149292 on Build GRCh38
rs753721065 Chr.17: 45149499-45149499 on Build GRCh38
rs764378739 Chr.17: 45149306-45149306 on Build GRCh38
rs762094836 Chr.17: 45149380-45149380 on Build GRCh38
rs779434293 Chr.17: 45149637-45149637 on Build GRCh38
rs367729540 Chr.17: 45149249-45149249 on Build GRCh38
rs766775697 Chr.17: 45149199-45149199 on Build GRCh38
rs751186893 Chr.17: 45149615-45149615 on Build GRCh38
rs780022419 Chr.17: 45149319-45149319 on Build GRCh38
rs762138738 Chr.17: 45149479-45149479 on Build GRCh38
rs769413846 Chr.17: 45149259-45149259 on Build GRCh38
rs769192457 Chr.17: 45149542-45149542 on Build GRCh38
rs370060246 Chr.17: 45149574-45149574 on Build GRCh38
rs758700108 Chr.17: 45149228-45149228 on Build GRCh38
rs747061601 Chr.17: 45149236-45149236 on Build GRCh38
rs140416514 Chr.17: 45149255-45149255 on Build GRCh38
rs372722263 Chr.17: 45149476-45149476 on Build GRCh38
rs868552409 Chr.17: 45149342-45149342 on Build GRCh38
rs769075068 Chr.17: 45149457-45149457 on Build GRCh38
rs200079932 Chr.17: 45149337-45149337 on Build GRCh38
rs1024230967 Chr.17: 45149636-45149636 on Build GRCh38
rs754411480 Chr.17: 45149201-45149201 on Build GRCh38
rs752524718 Chr.17: 45149517-45149517 on Build GRCh38
rs377233768 Chr.17: 45149562-45149562 on Build GRCh38
rs756961690 Chr.17: 45149627-45149627 on Build GRCh38
rs777735067 Chr.17: 45149260-45149260 on Build GRCh38
rs373817429 Chr.17: 45149237-45149237 on Build GRCh38
rs765202046 Chr.17: 45149577-45149577 on Build GRCh38
rs761441239 Chr.17: 45149195-45149195 on Build GRCh38
rs778783867 Chr.17: 45149516-45149516 on Build GRCh38
rs372695634 Chr.17: 45149327-45149327 on Build GRCh38
rs776178507 Chr.17: 45149370-45149370 on Build GRCh38
rs967415341 Chr.17: 45149405-45149405 on Build GRCh38
rs12051846 Chr.17: 45149520-45149520 on Build GRCh38
rs750499219 Chr.17: 45149409-45149409 on Build GRCh38
rs898998482 Chr.17: 45149305-45149305 on Build GRCh38
rs977466725 Chr.17: 45149417-45149417 on Build GRCh38
rs771429350 Chr.17: 45149343-45149343 on Build GRCh38
rs774768717 Chr.17: 45149462-45149462 on Build GRCh38
rs780853522 Chr.17: 45149533-45149533 on Build GRCh38
rs763523915 Chr.17: 45149184-45149184 on Build GRCh38
rs772784303 Chr.17: 45149345-45149345 on Build GRCh38
rs745438153 Chr.17: 45149536-45149536 on Build GRCh38
rs781344787 Chr.17: 45149313-45149313 on Build GRCh38
rs746679626 Chr.17: 45149523-45149523 on Build GRCh38
rs572482083 Chr.17: 45149431-45149431 on Build GRCh38
rs768281978 Chr.17: 45149281-45149281 on Build GRCh38
rs748513171 Chr.17: 45149558-45149558 on Build GRCh38
rs202151146 Chr.17: 45149564-45149564 on Build GRCh38
rs760170895 Chr.17: 45149356-45149356 on Build GRCh38
rs745348258 Chr.17: 45149632-45149632 on Build GRCh38
rs776090403 Chr.17: 45149453-45149453 on Build GRCh38
rs374259515 Chr.17: 45149610-45149610 on Build GRCh38
rs763269543 Chr.17: 45149590-45149590 on Build GRCh38
rs867729042 Chr.17: 45149548-45149548 on Build GRCh38
rs980549845 Chr.17: 45149484-45149484 on Build GRCh38
rs764574971 Chr.17: 45149376-45149376 on Build GRCh38
rs759674454 Chr.17: 45149567-45149567 on Build GRCh38
rs749057704 Chr.17: 45149325-45149325 on Build GRCh38
rs745839893 Chr.17: 45149258-45149258 on Build GRCh38
rs539194359 Chr.17: 45149373-45149373 on Build GRCh38
rs142769439 Chr.17: 45149295-45149295 on Build GRCh38
rs767088972 Chr.17: 45149290-45149290 on Build GRCh38
rs758892714 Chr.17: 45149304-45149304 on Build GRCh38
rs762208355 Chr.17: 45149314-45149316 on Build GRCh38
rs750750456 Chr.17: 45149594-45149595 on Build GRCh38
rs778210451 Chr.17: 45149231-45149231 on Build GRCh38
rs773843252 Chr.17: 45149190-45149190 on Build GRCh38
rs147427660 Chr.17: 45149429-45149429 on Build GRCh38
rs564812671 Chr.17: 45149561-45149561 on Build GRCh38
rs752910152 Chr.17: 45149222-45149222 on Build GRCh38
rs745722539 Chr.17: 45149315-45149315 on Build GRCh38
rs755261268 Chr.17: 45149203-45149203 on Build GRCh38
rs770412496 Chr.17: 45149363-45149363 on Build GRCh38
rs17850101 Chr.17: 45149518-45149518 on Build GRCh38
rs765817679 Chr.17: 45149470-45149470 on Build GRCh38
rs772356897 Chr.17: 45149559-45149559 on Build GRCh38
rs756070648 Chr.17: 45149317-45149317 on Build GRCh38
rs371316696 Chr.17: 45149521-45149521 on Build GRCh38
rs752620066 Chr.17: 45149578-45149578 on Build GRCh38
rs777350008 Chr.17: 45149447-45149447 on Build GRCh38
rs868242690 Chr.17: 45149539-45149539 on Build GRCh38
rs139710804 Chr.17: 45149302-45149302 on Build GRCh38
rs200239219 Chr.17: 45149565-45149565 on Build GRCh38
rs774768886 Chr.17: 45149545-45149545 on Build GRCh38
rs201778839 Chr.17: 45149485-45149485 on Build GRCh38
rs767754935 Chr.17: 45149480-45149480 on Build GRCh38
rs763806684 Chr.17: 45149597-45149597 on Build GRCh38
rs894323965 Chr.17: 45149585-45149585 on Build GRCh38

View Full Product Details