Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48622282-48622775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXB8
Entrez Gene ID
3218
Gene Name
homeobox B8
Gene Aliases
HOX2, HOX2D, Hox-2.4
UniGene
Hs.514292
Gene Chromosome Location
Chr.17: 48612346-48622853 on Build GRCh38
Transcripts
XM_005257286.2 XM_017024564.1 NM_024017.4

Gene Symbol
HOXB9
Entrez Gene ID
3219
Gene Name
homeobox B9
Gene Aliases
HOX-2.5, HOX2, HOX2E
UniGene
Hs.463350
Gene Chromosome Location
Chr.17: 48621156-48626473 on Build GRCh38
Transcripts
XM_005257286.2 XM_017024564.1 NM_024017.4

SNP Detail

SNP ID Chromosome Location
rs565093808 Chr.17: 48622644-48622644 on Build GRCh38
rs748571883 Chr.17: 48622419-48622419 on Build GRCh38
rs542950345 Chr.17: 48622510-48622510 on Build GRCh38
rs1026427470 Chr.17: 48622702-48622702 on Build GRCh38
rs915516832 Chr.17: 48622556-48622556 on Build GRCh38
rs770485306 Chr.17: 48622603-48622603 on Build GRCh38
rs41300616 Chr.17: 48622508-48622508 on Build GRCh38
rs772675520 Chr.17: 48622432-48622432 on Build GRCh38
rs910075085 Chr.17: 48622421-48622421 on Build GRCh38
rs903660536 Chr.17: 48622595-48622595 on Build GRCh38
rs186208263 Chr.17: 48622532-48622532 on Build GRCh38
rs976156012 Chr.17: 48622369-48622369 on Build GRCh38
rs547572900 Chr.17: 48622707-48622707 on Build GRCh38
rs776003623 Chr.17: 48622457-48622457 on Build GRCh38
rs1023479460 Chr.17: 48622440-48622440 on Build GRCh38
rs896822606 Chr.17: 48622346-48622346 on Build GRCh38
rs113702226 Chr.17: 48622423-48622423 on Build GRCh38
rs776077798 Chr.17: 48622709-48622709 on Build GRCh38
rs892540326 Chr.17: 48622561-48622561 on Build GRCh38
rs1017546877 Chr.17: 48622318-48622318 on Build GRCh38
rs4239158 Chr.17: 48622489-48622489 on Build GRCh38
rs2303486 Chr.17: 48622307-48622307 on Build GRCh38
rs879199164 Chr.17: 48622325-48622325 on Build GRCh38
rs41315066 Chr.17: 48622326-48622326 on Build GRCh38
rs896808480 Chr.17: 48622637-48622637 on Build GRCh38
rs923933603 Chr.17: 48622459-48622459 on Build GRCh38
rs1043852824 Chr.17: 48622590-48622590 on Build GRCh38
rs932253960 Chr.17: 48622559-48622559 on Build GRCh38
rs1015978131 Chr.17: 48622733-48622733 on Build GRCh38
rs993865125 Chr.17: 48622664-48622664 on Build GRCh38
rs183369267 Chr.17: 48622370-48622370 on Build GRCh38
rs527790273 Chr.17: 48622675-48622675 on Build GRCh38
rs545401023 Chr.17: 48622630-48622630 on Build GRCh38
rs866084455 Chr.17: 48622392-48622392 on Build GRCh38
rs997183249 Chr.17: 48622652-48622652 on Build GRCh38
rs367858610 Chr.17: 48622531-48622531 on Build GRCh38
rs901795553 Chr.17: 48622635-48622635 on Build GRCh38
rs1040841548 Chr.17: 48622636-48622636 on Build GRCh38
rs191784064 Chr.17: 48622567-48622567 on Build GRCh38
rs1038930282 Chr.17: 48622608-48622608 on Build GRCh38
rs926872766 Chr.17: 48622445-48622445 on Build GRCh38
rs982897985 Chr.17: 48622416-48622416 on Build GRCh38
rs965705999 Chr.17: 48622359-48622359 on Build GRCh38
rs79931349 Chr.17: 48622371-48622371 on Build GRCh38
rs147466612 Chr.17: 48622434-48622434 on Build GRCh38

View Full Product Details