Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82052198-82052699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPS1
Entrez Gene ID
2873
Gene Name
G protein pathway suppressor 1
Gene Aliases
COPS1, CSN1, SGN1
UniGene
Hs.268530
Gene Chromosome Location
Chr.17: 82051477-82057470 on Build GRCh38
Transcripts
NM_001321089.1 NM_001321090.1 NM_001321091.1 NM_001321092.1 NM_001321093.1 NM_004127.5 NM_212492.2 XM_005256350.1 XM_005256353.1 XM_005256359.1 XM_005256360.3 XM_011523569.1 XM_011523570.1 XM_017024524.1 XM_017024525.1 XM_017024526.1 XM_017024527.1 XM_017024528.1 XM_017024529.1 XM_017024530.1 XM_017024531.1 XM_017024532.1 XM_017024533.1 XM_017024534.1 XM_017024535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7291760 c.16G>A p.A6T Chr.17: 82052322-82052322 on Build GRCh38
3773343 c.33C>A p.S11S Chr.17: 82052339-82052339 on Build GRCh38
437815 c.138T>C p.C46C Chr.17: 82052444-82052444 on Build GRCh38
2806020 c.144G>A p.L48L Chr.17: 82052450-82052450 on Build GRCh38
2806013 c.12C>T p.S4S Chr.17: 82052318-82052318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758578519 Chr.17: 82052448-82052448 on Build GRCh38
rs780324128 Chr.17: 82052281-82052281 on Build GRCh38
rs373286823 Chr.17: 82052433-82052433 on Build GRCh38
rs773224502 Chr.17: 82052388-82052388 on Build GRCh38
rs894736807 Chr.17: 82052664-82052664 on Build GRCh38
rs1041704897 Chr.17: 82052618-82052618 on Build GRCh38
rs761518596 Chr.17: 82052338-82052338 on Build GRCh38
rs971421113 Chr.17: 82052546-82052546 on Build GRCh38
rs984475178 Chr.17: 82052592-82052592 on Build GRCh38
rs750390121 Chr.17: 82052267-82052267 on Build GRCh38
rs750215459 Chr.17: 82052341-82052341 on Build GRCh38
rs956062132 Chr.17: 82052514-82052514 on Build GRCh38
rs367679480 Chr.17: 82052463-82052463 on Build GRCh38
rs756101174 Chr.17: 82052280-82052280 on Build GRCh38
rs376089228 Chr.17: 82052226-82052226 on Build GRCh38
rs956030974 Chr.17: 82052346-82052346 on Build GRCh38
rs371663355 Chr.17: 82052467-82052467 on Build GRCh38
rs767568919 Chr.17: 82052282-82052282 on Build GRCh38
rs542090557 Chr.17: 82052399-82052399 on Build GRCh38
rs942510289 Chr.17: 82052594-82052594 on Build GRCh38
rs1000364181 Chr.17: 82052246-82052246 on Build GRCh38
rs368049856 Chr.17: 82052494-82052494 on Build GRCh38
rs764019253 Chr.17: 82052352-82052353 on Build GRCh38
rs537526868 Chr.17: 82052675-82052675 on Build GRCh38
rs939686955 Chr.17: 82052607-82052607 on Build GRCh38
rs747561442 Chr.17: 82052327-82052327 on Build GRCh38
rs551520949 Chr.17: 82052640-82052640 on Build GRCh38
rs773893764 Chr.17: 82052404-82052404 on Build GRCh38
rs1014391786 Chr.17: 82052667-82052667 on Build GRCh38
rs984470247 Chr.17: 82052544-82052544 on Build GRCh38
rs201302867 Chr.17: 82052480-82052480 on Build GRCh38
rs764326046 Chr.17: 82052453-82052453 on Build GRCh38
rs748412379 Chr.17: 82052232-82052232 on Build GRCh38
rs372303943 Chr.17: 82052495-82052495 on Build GRCh38
rs775452065 Chr.17: 82052507-82052507 on Build GRCh38
rs771718012 Chr.17: 82052408-82052408 on Build GRCh38
rs572264154 Chr.17: 82052440-82052440 on Build GRCh38
rs146708684 Chr.17: 82052331-82052331 on Build GRCh38
rs775358337 Chr.17: 82052316-82052327 on Build GRCh38
rs371023131 Chr.17: 82052442-82052442 on Build GRCh38
rs557827043 Chr.17: 82052242-82052242 on Build GRCh38
rs761166603 Chr.17: 82052239-82052239 on Build GRCh38
rs57313438 Chr.17: 82052308-82052308 on Build GRCh38
rs529594445 Chr.17: 82052536-82052536 on Build GRCh38
rs760726493 Chr.17: 82052344-82052344 on Build GRCh38
rs753980537 Chr.17: 82052355-82052355 on Build GRCh38
rs376384816 Chr.17: 82052372-82052372 on Build GRCh38
rs140262257 Chr.17: 82052393-82052393 on Build GRCh38
rs545970806 Chr.17: 82052455-82052455 on Build GRCh38
rs756616541 Chr.17: 82052483-82052483 on Build GRCh38
rs768113538 Chr.17: 82052403-82052403 on Build GRCh38
rs375617874 Chr.17: 82052443-82052443 on Build GRCh38
rs755240495 Chr.17: 82052615-82052615 on Build GRCh38
rs766372671 Chr.17: 82052348-82052348 on Build GRCh38
rs772647543 Chr.17: 82052234-82052234 on Build GRCh38
rs548018929 Chr.17: 82052264-82052264 on Build GRCh38
rs771448835 Chr.17: 82052395-82052395 on Build GRCh38
rs761415005 Chr.17: 82052406-82052406 on Build GRCh38
rs373403707 Chr.17: 82052332-82052332 on Build GRCh38
rs760491080 Chr.17: 82052428-82052428 on Build GRCh38
rs565979687 Chr.17: 82052603-82052603 on Build GRCh38
rs547675123 Chr.17: 82052589-82052589 on Build GRCh38
rs892017027 Chr.17: 82052663-82052663 on Build GRCh38
rs752920597 Chr.17: 82052383-82052383 on Build GRCh38
rs928783804 Chr.17: 82052317-82052317 on Build GRCh38
rs780270950 Chr.17: 82052220-82052220 on Build GRCh38
rs764782776 Chr.17: 82052241-82052241 on Build GRCh38
rs1028877747 Chr.17: 82052497-82052497 on Build GRCh38
rs926488036 Chr.17: 82052570-82052570 on Build GRCh38
rs981969017 Chr.17: 82052560-82052560 on Build GRCh38
rs569832593 Chr.17: 82052665-82052665 on Build GRCh38
rs771395733 Chr.17: 82052329-82052329 on Build GRCh38
rs773525057 Chr.17: 82052236-82052236 on Build GRCh38
rs755275782 Chr.17: 82052296-82052296 on Build GRCh38
rs1049491352 Chr.17: 82052642-82052642 on Build GRCh38
rs767344099 Chr.17: 82052266-82052266 on Build GRCh38
rs369798418 Chr.17: 82052387-82052387 on Build GRCh38
rs561704070 Chr.17: 82052519-82052519 on Build GRCh38
rs763693350 Chr.17: 82052259-82052259 on Build GRCh38
rs889978420 Chr.17: 82052647-82052647 on Build GRCh38
rs1000349452 Chr.17: 82052652-82052652 on Build GRCh38
rs1024356213 Chr.17: 82052532-82052532 on Build GRCh38
rs370086561 Chr.17: 82052627-82052627 on Build GRCh38
rs756941562 Chr.17: 82052262-82052262 on Build GRCh38
rs1025766846 Chr.17: 82052527-82052527 on Build GRCh38
rs751388258 Chr.17: 82052260-82052260 on Build GRCh38
rs752128405 Chr.17: 82052499-82052499 on Build GRCh38
rs918471852 Chr.17: 82052390-82052393 on Build GRCh38
rs1033240380 Chr.17: 82052662-82052662 on Build GRCh38
rs144100968 Chr.17: 82052427-82052427 on Build GRCh38
rs778945347 Chr.17: 82052305-82052305 on Build GRCh38
rs748551957 Chr.17: 82052311-82052311 on Build GRCh38
rs988882764 Chr.17: 82052367-82052367 on Build GRCh38
rs748797995 Chr.17: 82052400-82052400 on Build GRCh38
rs781529094 Chr.17: 82052479-82052479 on Build GRCh38
rs767267218 Chr.17: 82052339-82052339 on Build GRCh38
rs376662207 Chr.17: 82052290-82052290 on Build GRCh38
rs763484145 Chr.17: 82052452-82052452 on Build GRCh38
rs865861875 Chr.17: 82052522-82052522 on Build GRCh38
rs937852941 Chr.17: 82052573-82052573 on Build GRCh38
rs1056420524 Chr.17: 82052633-82052633 on Build GRCh38
rs952648492 Chr.17: 82052541-82052541 on Build GRCh38
rs201126611 Chr.17: 82052318-82052318 on Build GRCh38
rs202075762 Chr.17: 82052336-82052336 on Build GRCh38
rs753930921 Chr.17: 82052288-82052288 on Build GRCh38
rs752788575 Chr.17: 82052444-82052444 on Build GRCh38
rs760665070 Chr.17: 82052322-82052323 on Build GRCh38
rs778320214 Chr.17: 82052490-82052490 on Build GRCh38
rs765179699 Chr.17: 82052506-82052506 on Build GRCh38
rs200199758 Chr.17: 82052335-82052335 on Build GRCh38
rs778185702 Chr.17: 82052323-82052323 on Build GRCh38
rs762631627 Chr.17: 82052251-82052251 on Build GRCh38
rs911161307 Chr.17: 82052588-82052588 on Build GRCh38
rs539651313 Chr.17: 82052623-82052623 on Build GRCh38

View Full Product Details