Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82054420-82054929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPS1
Entrez Gene ID
2873
Gene Name
G protein pathway suppressor 1
Gene Aliases
COPS1, CSN1, SGN1
UniGene
Hs.268530
Gene Chromosome Location
Chr.17: 82051477-82057470 on Build GRCh38
Transcripts
NM_001321089.1 NM_001321090.1 NM_001321091.1 NM_001321092.1 NM_001321093.1 NM_004127.5 NM_212492.2 XM_005256350.1 XM_005256353.1 XM_005256359.1 XM_005256360.3 XM_011523569.1 XM_011523570.1 XM_017024524.1 XM_017024525.1 XM_017024526.1 XM_017024527.1 XM_017024528.1 XM_017024529.1 XM_017024530.1 XM_017024531.1 XM_017024532.1 XM_017024533.1 XM_017024534.1 XM_017024535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3738240 c.659G>T p.C220F Chr.17: 82054740-82054740 on Build GRCh38
6351651 c.492G>A p.T164T Chr.17: 82054573-82054573 on Build GRCh38
6353954 c.730-5G>A p.? Chr.17: 82054893-82054893 on Build GRCh38
6351652 c.384G>A p.T128T Chr.17: 82054573-82054573 on Build GRCh38
4139965 c.621C>G p.H207Q Chr.17: 82054702-82054702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778184632 Chr.17: 82054481-82054481 on Build GRCh38
rs373486488 Chr.17: 82054830-82054830 on Build GRCh38
rs775565772 Chr.17: 82054834-82054834 on Build GRCh38
rs754578240 Chr.17: 82054673-82054673 on Build GRCh38
rs779600084 Chr.17: 82054885-82054886 on Build GRCh38
rs890627172 Chr.17: 82054443-82054443 on Build GRCh38
rs532618201 Chr.17: 82054455-82054455 on Build GRCh38
rs556950675 Chr.17: 82054651-82054651 on Build GRCh38
rs778096914 Chr.17: 82054550-82054550 on Build GRCh38
rs369450843 Chr.17: 82054827-82054827 on Build GRCh38
rs763681337 Chr.17: 82054878-82054878 on Build GRCh38
rs370407144 Chr.17: 82054840-82054840 on Build GRCh38
rs758597336 Chr.17: 82054648-82054648 on Build GRCh38
rs760726274 Chr.17: 82054621-82054621 on Build GRCh38
rs745600248 Chr.17: 82054887-82054887 on Build GRCh38
rs778597131 Chr.17: 82054813-82054813 on Build GRCh38
rs966724245 Chr.17: 82054494-82054494 on Build GRCh38
rs763190588 Chr.17: 82054766-82054766 on Build GRCh38
rs767965990 Chr.17: 82054857-82054857 on Build GRCh38
rs752974709 Chr.17: 82054645-82054645 on Build GRCh38
rs758827950 Chr.17: 82054549-82054549 on Build GRCh38
rs143568996 Chr.17: 82054624-82054624 on Build GRCh38
rs760671219 Chr.17: 82054528-82054528 on Build GRCh38
rs758138640 Chr.17: 82054817-82054817 on Build GRCh38
rs139212789 Chr.17: 82054657-82054657 on Build GRCh38
rs779898057 Chr.17: 82054888-82054888 on Build GRCh38
rs114775081 Chr.17: 82054867-82054867 on Build GRCh38
rs747792808 Chr.17: 82054603-82054603 on Build GRCh38
rs750992483 Chr.17: 82054801-82054801 on Build GRCh38
rs768311600 Chr.17: 82054502-82054502 on Build GRCh38
rs878893994 Chr.17: 82054748-82054748 on Build GRCh38
rs202153481 Chr.17: 82054892-82054892 on Build GRCh38
rs959951901 Chr.17: 82054846-82054846 on Build GRCh38
rs775512549 Chr.17: 82054765-82054765 on Build GRCh38
rs375895683 Chr.17: 82054454-82054454 on Build GRCh38
rs375853187 Chr.17: 82054771-82054771 on Build GRCh38
rs541936561 Chr.17: 82054547-82054547 on Build GRCh38
rs370601543 Chr.17: 82054862-82054862 on Build GRCh38
rs144937537 Chr.17: 82054729-82054729 on Build GRCh38
rs774035765 Chr.17: 82054505-82054505 on Build GRCh38
rs369415235 Chr.17: 82054789-82054789 on Build GRCh38
rs764478554 Chr.17: 82054649-82054649 on Build GRCh38
rs749530554 Chr.17: 82054526-82054526 on Build GRCh38
rs36080512 Chr.17: 82054706-82054706 on Build GRCh38
rs561281296 Chr.17: 82054471-82054471 on Build GRCh38
rs756551905 Chr.17: 82054803-82054803 on Build GRCh38
rs571991947 Chr.17: 82054531-82054531 on Build GRCh38
rs11077966 Chr.17: 82054573-82054573 on Build GRCh38
rs139982509 Chr.17: 82054590-82054590 on Build GRCh38
rs762571536 Chr.17: 82054732-82054732 on Build GRCh38
rs747847902 Chr.17: 82054688-82054688 on Build GRCh38
rs550977110 Chr.17: 82054480-82054480 on Build GRCh38
rs966671883 Chr.17: 82054690-82054690 on Build GRCh38
rs141937467 Chr.17: 82054562-82054562 on Build GRCh38
rs766291739 Chr.17: 82054622-82054622 on Build GRCh38
rs750538430 Chr.17: 82054444-82054444 on Build GRCh38
rs1051696078 Chr.17: 82054714-82054714 on Build GRCh38
rs140038239 Chr.17: 82054501-82054501 on Build GRCh38
rs752206815 Chr.17: 82054815-82054815 on Build GRCh38
rs773344800 Chr.17: 82054523-82054523 on Build GRCh38
rs748498826 Chr.17: 82054477-82054477 on Build GRCh38
rs116882446 Chr.17: 82054750-82054750 on Build GRCh38
rs768868833 Chr.17: 82054841-82054841 on Build GRCh38
rs376959069 Chr.17: 82054847-82054847 on Build GRCh38
rs368324967 Chr.17: 82054890-82054890 on Build GRCh38
rs765226186 Chr.17: 82054639-82054639 on Build GRCh38
rs768201465 Chr.17: 82054600-82054600 on Build GRCh38
rs201830902 Chr.17: 82054831-82054831 on Build GRCh38
rs973347591 Chr.17: 82054900-82054900 on Build GRCh38
rs11539403 Chr.17: 82054540-82054540 on Build GRCh38
rs1026859984 Chr.17: 82054442-82054442 on Build GRCh38
rs1035379619 Chr.17: 82054593-82054593 on Build GRCh38
rs1009050183 Chr.17: 82054654-82054654 on Build GRCh38
rs762959455 Chr.17: 82054836-82054836 on Build GRCh38
rs777799705 Chr.17: 82054445-82054445 on Build GRCh38
rs375230398 Chr.17: 82054826-82054826 on Build GRCh38
rs750713783 Chr.17: 82054861-82054861 on Build GRCh38
rs868110239 Chr.17: 82054527-82054527 on Build GRCh38
rs78902137 Chr.17: 82054634-82054634 on Build GRCh38
rs751271572 Chr.17: 82054884-82054884 on Build GRCh38
rs530041584 Chr.17: 82054886-82054886 on Build GRCh38
rs149095451 Chr.17: 82054781-82054781 on Build GRCh38
rs772045477 Chr.17: 82054611-82054611 on Build GRCh38
rs371231940 Chr.17: 82054535-82054535 on Build GRCh38
rs935356970 Chr.17: 82054721-82054721 on Build GRCh38
rs544365812 Chr.17: 82054866-82054866 on Build GRCh38
rs372833632 Chr.17: 82054666-82054666 on Build GRCh38
rs746744726 Chr.17: 82054703-82054703 on Build GRCh38
rs546416950 Chr.17: 82054904-82054904 on Build GRCh38
rs750021803 Chr.17: 82054823-82054825 on Build GRCh38
rs748705896 Chr.17: 82054886-82054886 on Build GRCh38
rs751931963 Chr.17: 82054776-82054776 on Build GRCh38
rs775038430 Chr.17: 82054498-82054498 on Build GRCh38
rs890703734 Chr.17: 82054515-82054515 on Build GRCh38
rs769351385 Chr.17: 82054491-82054491 on Build GRCh38
rs757910030 Chr.17: 82054882-82054883 on Build GRCh38
rs779177158 Chr.17: 82054476-82054476 on Build GRCh38
rs777733533 Chr.17: 82054699-82054699 on Build GRCh38
rs11655185 Chr.17: 82054893-82054893 on Build GRCh38
rs761636383 Chr.17: 82054506-82054506 on Build GRCh38
rs893945180 Chr.17: 82054530-82054530 on Build GRCh38
rs868217672 Chr.17: 82054704-82054704 on Build GRCh38
rs999592009 Chr.17: 82054720-82054720 on Build GRCh38
rs898918255 Chr.17: 82054576-82054576 on Build GRCh38
rs890478605 Chr.17: 82054717-82054717 on Build GRCh38
rs750893372 Chr.17: 82054669-82054669 on Build GRCh38
rs755196906 Chr.17: 82054472-82054472 on Build GRCh38
rs770816362 Chr.17: 82054824-82054824 on Build GRCh38
rs777502521 Chr.17: 82054819-82054819 on Build GRCh38
rs367741530 Chr.17: 82054678-82054678 on Build GRCh38
rs748433801 Chr.17: 82054891-82054891 on Build GRCh38
rs771810470 Chr.17: 82054696-82054696 on Build GRCh38
rs780272863 Chr.17: 82054451-82054451 on Build GRCh38
rs369888073 Chr.17: 82054529-82054529 on Build GRCh38
rs549000263 Chr.17: 82054546-82054546 on Build GRCh38
rs747559436 Chr.17: 82054556-82054556 on Build GRCh38
rs747510042 Chr.17: 82054487-82054487 on Build GRCh38
rs774123575 Chr.17: 82054602-82054602 on Build GRCh38
rs371520281 Chr.17: 82054885-82054885 on Build GRCh38
rs574941019 Chr.17: 82054863-82054863 on Build GRCh38
rs200876960 Chr.17: 82054842-82054842 on Build GRCh38
rs373015830 Chr.17: 82054522-82054522 on Build GRCh38
rs769965931 Chr.17: 82054756-82054756 on Build GRCh38
rs373314198 Chr.17: 82054823-82054823 on Build GRCh38
rs773111199 Chr.17: 82054618-82054618 on Build GRCh38
rs770776929 Chr.17: 82054724-82054724 on Build GRCh38

View Full Product Details