Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 547124-547629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GZMM
Entrez Gene ID
3004
Gene Name
granzyme M
Gene Aliases
LMET1, MET1
UniGene
Hs.465511
Gene Chromosome Location
Chr.19: 544034-549920 on Build GRCh38
Transcripts
NM_001258351.1 NM_005317.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7370612 c.121G>A p.A41T Chr.19: 547345-547345 on Build GRCh38
6344640 c.56-5G>T p.? Chr.19: 547275-547275 on Build GRCh38
3538625 c.62C>T p.S21F Chr.19: 547286-547286 on Build GRCh38
1001045 c.114G>A p.P38P Chr.19: 547338-547338 on Build GRCh38
6677332 c.110G>A p.R37H Chr.19: 547334-547334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764766156 Chr.19: 547488-547488 on Build GRCh38
rs201257717 Chr.19: 547287-547287 on Build GRCh38
rs769969742 Chr.19: 547377-547377 on Build GRCh38
rs373840917 Chr.19: 547312-547312 on Build GRCh38
rs980136628 Chr.19: 547217-547217 on Build GRCh38
rs962279423 Chr.19: 547266-547266 on Build GRCh38
rs765915582 Chr.19: 547362-547362 on Build GRCh38
rs780211377 Chr.19: 547448-547448 on Build GRCh38
rs771020787 Chr.19: 547263-547263 on Build GRCh38
rs757450436 Chr.19: 547370-547370 on Build GRCh38
rs185429686 Chr.19: 547544-547544 on Build GRCh38
rs759077712 Chr.19: 547485-547485 on Build GRCh38
rs1021538122 Chr.19: 547188-547188 on Build GRCh38
rs2070796 Chr.19: 547555-547555 on Build GRCh38
rs780087868 Chr.19: 547406-547415 on Build GRCh38
rs755067260 Chr.19: 547415-547416 on Build GRCh38
rs779267733 Chr.19: 547235-547235 on Build GRCh38
rs374939287 Chr.19: 547412-547412 on Build GRCh38
rs774142092 Chr.19: 547390-547390 on Build GRCh38
rs867165968 Chr.19: 547349-547349 on Build GRCh38
rs756467224 Chr.19: 547438-547438 on Build GRCh38
rs768715164 Chr.19: 547380-547380 on Build GRCh38
rs749910314 Chr.19: 547309-547309 on Build GRCh38
rs768668456 Chr.19: 547460-547460 on Build GRCh38
rs770576095 Chr.19: 547482-547482 on Build GRCh38
rs753181415 Chr.19: 547364-547364 on Build GRCh38
rs755458400 Chr.19: 547310-547310 on Build GRCh38
rs146184990 Chr.19: 547223-547223 on Build GRCh38
rs1015053406 Chr.19: 547258-547258 on Build GRCh38
rs763964114 Chr.19: 547343-547343 on Build GRCh38
rs753936908 Chr.19: 547410-547411 on Build GRCh38
rs556484742 Chr.19: 547306-547306 on Build GRCh38
rs776715218 Chr.19: 547332-547332 on Build GRCh38
rs762870720 Chr.19: 547280-547280 on Build GRCh38
rs1034717815 Chr.19: 547560-547560 on Build GRCh38
rs761581140 Chr.19: 547228-547228 on Build GRCh38
rs1048668692 Chr.19: 547268-547268 on Build GRCh38
rs143995851 Chr.19: 547313-547313 on Build GRCh38
rs968685063 Chr.19: 547189-547189 on Build GRCh38
rs908920474 Chr.19: 547545-547545 on Build GRCh38
rs764607104 Chr.19: 547426-547426 on Build GRCh38
rs748522250 Chr.19: 547245-547245 on Build GRCh38
rs759450901 Chr.19: 547267-547267 on Build GRCh38
rs199725777 Chr.19: 547467-547467 on Build GRCh38
rs200360663 Chr.19: 547327-547327 on Build GRCh38
rs569468334 Chr.19: 547147-547147 on Build GRCh38
rs749396827 Chr.19: 547458-547458 on Build GRCh38
rs145060399 Chr.19: 547331-547331 on Build GRCh38
rs755991300 Chr.19: 547405-547406 on Build GRCh38
rs763030017 Chr.19: 547341-547341 on Build GRCh38
rs753412913 Chr.19: 547416-547416 on Build GRCh38
rs538294785 Chr.19: 547554-547554 on Build GRCh38
rs368042562 Chr.19: 547338-547338 on Build GRCh38
rs147907546 Chr.19: 547429-547429 on Build GRCh38
rs889330290 Chr.19: 547586-547586 on Build GRCh38
rs935250038 Chr.19: 547221-547221 on Build GRCh38
rs778106883 Chr.19: 547365-547365 on Build GRCh38
rs755613208 Chr.19: 547356-547356 on Build GRCh38
rs751231756 Chr.19: 547296-547296 on Build GRCh38
rs760316637 Chr.19: 547473-547473 on Build GRCh38
rs748710730 Chr.19: 547587-547587 on Build GRCh38
rs923821587 Chr.19: 547198-547198 on Build GRCh38
rs2070794 Chr.19: 547483-547483 on Build GRCh38
rs4919846 Chr.19: 547224-547224 on Build GRCh38
rs942980035 Chr.19: 547476-547476 on Build GRCh38
rs751910126 Chr.19: 547427-547427 on Build GRCh38
rs151192827 Chr.19: 547347-547347 on Build GRCh38
rs566770452 Chr.19: 547222-547222 on Build GRCh38
rs779153027 Chr.19: 547449-547458 on Build GRCh38
rs772230082 Chr.19: 547251-547251 on Build GRCh38
rs146522321 Chr.19: 547378-547378 on Build GRCh38
rs992020751 Chr.19: 547516-547516 on Build GRCh38
rs543874497 Chr.19: 547404-547404 on Build GRCh38
rs766184418 Chr.19: 547339-547339 on Build GRCh38
rs746140221 Chr.19: 547376-547376 on Build GRCh38
rs527907006 Chr.19: 547500-547500 on Build GRCh38
rs760925690 Chr.19: 547259-547259 on Build GRCh38
rs1051454332 Chr.19: 547492-547492 on Build GRCh38
rs761539542 Chr.19: 547401-547401 on Build GRCh38
rs767160387 Chr.19: 547402-547402 on Build GRCh38
rs1025694096 Chr.19: 547594-547594 on Build GRCh38
rs2070795 Chr.19: 547491-547491 on Build GRCh38
rs948004680 Chr.19: 547540-547540 on Build GRCh38
rs868146010 Chr.19: 547302-547302 on Build GRCh38
rs917570055 Chr.19: 547342-547342 on Build GRCh38
rs386805631 Chr.19: 547223-547224 on Build GRCh38
rs773452629 Chr.19: 547257-547257 on Build GRCh38
rs772396880 Chr.19: 547314-547314 on Build GRCh38
rs746061534 Chr.19: 547436-547436 on Build GRCh38
rs765862575 Chr.19: 547413-547413 on Build GRCh38
rs1051948755 Chr.19: 547583-547583 on Build GRCh38
rs781655159 Chr.19: 547431-547431 on Build GRCh38
rs548378481 Chr.19: 547191-547191 on Build GRCh38
rs140327613 Chr.19: 547372-547372 on Build GRCh38
rs148691419 Chr.19: 547333-547333 on Build GRCh38
rs776632660 Chr.19: 547264-547264 on Build GRCh38
rs193277130 Chr.19: 547532-547532 on Build GRCh38
rs778751059 Chr.19: 547465-547465 on Build GRCh38
rs752489191 Chr.19: 547272-547272 on Build GRCh38
rs867370843 Chr.19: 547329-547329 on Build GRCh38
rs773118602 Chr.19: 547470-547470 on Build GRCh38
rs188296189 Chr.19: 547160-547160 on Build GRCh38
rs61729586 Chr.19: 547346-547346 on Build GRCh38
rs369284240 Chr.19: 547469-547469 on Build GRCh38
rs2070797 Chr.19: 547561-547561 on Build GRCh38
rs139076851 Chr.19: 547305-547305 on Build GRCh38
rs769729181 Chr.19: 547337-547337 on Build GRCh38
rs933455848 Chr.19: 547569-547569 on Build GRCh38
rs930166545 Chr.19: 547239-547239 on Build GRCh38
rs887546083 Chr.19: 547238-547238 on Build GRCh38
rs370173494 Chr.19: 547344-547344 on Build GRCh38

View Full Product Details