Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 549444-549986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GZMM
Entrez Gene ID
3004
Gene Name
granzyme M
Gene Aliases
LMET1, MET1
UniGene
Hs.465511
Gene Chromosome Location
Chr.19: 544034-549920 on Build GRCh38
Transcripts
NM_001258351.1 NM_005317.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3835713 c.694G>T p.D232Y Chr.19: 549711-549711 on Build GRCh38
4942233 c.762C>G p.G254G Chr.19: 549779-549779 on Build GRCh38
4689403 c.692C>T p.T231I Chr.19: 549709-549709 on Build GRCh38
7370610 c.614G>T p.G205V Chr.19: 549631-549631 on Build GRCh38
6371447 c.619T>C p.S207P Chr.19: 549636-549636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760924563 Chr.19: 549753-549753 on Build GRCh38
rs12459388 Chr.19: 549509-549509 on Build GRCh38
rs773665762 Chr.19: 549812-549812 on Build GRCh38
rs778518334 Chr.19: 549638-549638 on Build GRCh38
rs761156084 Chr.19: 549817-549817 on Build GRCh38
rs1002626095 Chr.19: 549868-549868 on Build GRCh38
rs549002952 Chr.19: 549776-549776 on Build GRCh38
rs141658632 Chr.19: 549752-549752 on Build GRCh38
rs75943115 Chr.19: 549496-549496 on Build GRCh38
rs866471498 Chr.19: 549873-549873 on Build GRCh38
rs1010046340 Chr.19: 549830-549830 on Build GRCh38
rs139629210 Chr.19: 549707-549707 on Build GRCh38
rs1599882 Chr.19: 549678-549678 on Build GRCh38
rs151036435 Chr.19: 549781-549781 on Build GRCh38
rs770415036 Chr.19: 549657-549657 on Build GRCh38
rs751967137 Chr.19: 549617-549617 on Build GRCh38
rs962238712 Chr.19: 549532-549532 on Build GRCh38
rs771243383 Chr.19: 549819-549819 on Build GRCh38
rs569015004 Chr.19: 549916-549916 on Build GRCh38
rs1040874046 Chr.19: 549487-549487 on Build GRCh38
rs748318382 Chr.19: 549915-549915 on Build GRCh38
rs780866675 Chr.19: 549787-549787 on Build GRCh38
rs374806995 Chr.19: 549654-549654 on Build GRCh38
rs953598382 Chr.19: 549948-549948 on Build GRCh38
rs200659579 Chr.19: 549729-549729 on Build GRCh38
rs542623279 Chr.19: 549645-549645 on Build GRCh38
rs370155439 Chr.19: 549790-549790 on Build GRCh38
rs778944154 Chr.19: 549702-549702 on Build GRCh38
rs759359247 Chr.19: 549565-549565 on Build GRCh38
rs748596570 Chr.19: 549807-549807 on Build GRCh38
rs778685093 Chr.19: 549547-549565 on Build GRCh38
rs973987351 Chr.19: 549732-549732 on Build GRCh38
rs906622519 Chr.19: 549854-549854 on Build GRCh38
rs111763479 Chr.19: 549829-549829 on Build GRCh38
rs17845771 Chr.19: 549679-549679 on Build GRCh38
rs376548111 Chr.19: 549785-549785 on Build GRCh38
rs766502816 Chr.19: 549685-549685 on Build GRCh38
rs970822731 Chr.19: 549512-549512 on Build GRCh38
rs931443732 Chr.19: 549491-549491 on Build GRCh38
rs745413232 Chr.19: 549733-549733 on Build GRCh38
rs572901883 Chr.19: 549673-549673 on Build GRCh38
rs776208077 Chr.19: 549659-549659 on Build GRCh38
rs758398362 Chr.19: 549780-549780 on Build GRCh38
rs919637009 Chr.19: 549770-549770 on Build GRCh38
rs530541565 Chr.19: 549518-549518 on Build GRCh38
rs539206442 Chr.19: 549620-549620 on Build GRCh38
rs759556462 Chr.19: 549474-549474 on Build GRCh38
rs1028218014 Chr.19: 549947-549947 on Build GRCh38
rs761463798 Chr.19: 549677-549677 on Build GRCh38
rs556663346 Chr.19: 549581-549581 on Build GRCh38
rs745551320 Chr.19: 549789-549789 on Build GRCh38
rs982870432 Chr.19: 549927-549927 on Build GRCh38
rs58562291 Chr.19: 549725-549725 on Build GRCh38
rs958159385 Chr.19: 549564-549564 on Build GRCh38
rs996870769 Chr.19: 549517-549517 on Build GRCh38
rs965130278 Chr.19: 549961-549961 on Build GRCh38
rs538877467 Chr.19: 549870-549870 on Build GRCh38
rs748077045 Chr.19: 549706-549706 on Build GRCh38
rs753183174 Chr.19: 549596-549596 on Build GRCh38
rs547224436 Chr.19: 549502-549502 on Build GRCh38
rs761306555 Chr.19: 549594-549595 on Build GRCh38
rs760557962 Chr.19: 549728-549729 on Build GRCh38
rs772722407 Chr.19: 549808-549808 on Build GRCh38
rs76318413 Chr.19: 549607-549607 on Build GRCh38
rs1032400417 Chr.19: 549561-549561 on Build GRCh38
rs754826849 Chr.19: 549693-549693 on Build GRCh38
rs755055521 Chr.19: 549769-549769 on Build GRCh38
rs898267162 Chr.19: 549485-549485 on Build GRCh38
rs768644937 Chr.19: 549637-549637 on Build GRCh38
rs777605067 Chr.19: 549713-549713 on Build GRCh38
rs139484295 Chr.19: 549631-549631 on Build GRCh38
rs548964053 Chr.19: 549568-549568 on Build GRCh38
rs764684750 Chr.19: 549666-549666 on Build GRCh38
rs142892784 Chr.19: 549777-549777 on Build GRCh38
rs908254099 Chr.19: 549940-549940 on Build GRCh38
rs944192890 Chr.19: 549786-549786 on Build GRCh38
rs759735037 Chr.19: 549827-549827 on Build GRCh38
rs780505573 Chr.19: 549745-549745 on Build GRCh38
rs200816488 Chr.19: 549799-549799 on Build GRCh38
rs887355076 Chr.19: 549493-549493 on Build GRCh38
rs79638775 Chr.19: 549920-549920 on Build GRCh38
rs374422648 Chr.19: 549688-549688 on Build GRCh38
rs915104792 Chr.19: 549814-549814 on Build GRCh38
rs368863219 Chr.19: 549741-549741 on Build GRCh38
rs199678590 Chr.19: 549580-549580 on Build GRCh38
rs1032705355 Chr.19: 549497-549497 on Build GRCh38
rs371239880 Chr.19: 549683-549683 on Build GRCh38
rs937076836 Chr.19: 549473-549473 on Build GRCh38
rs772860823 Chr.19: 549724-549725 on Build GRCh38
rs973678909 Chr.19: 549546-549546 on Build GRCh38
rs377514288 Chr.19: 549611-549611 on Build GRCh38
rs910946294 Chr.19: 549610-549610 on Build GRCh38
rs991127619 Chr.19: 549612-549612 on Build GRCh38
rs767256530 Chr.19: 549613-549614 on Build GRCh38
rs776889894 Chr.19: 549823-549823 on Build GRCh38
rs893803104 Chr.19: 549499-549499 on Build GRCh38
rs116707574 Chr.19: 549875-549875 on Build GRCh38
rs58601458 Chr.19: 549728-549728 on Build GRCh38
rs756238054 Chr.19: 549627-549627 on Build GRCh38
rs1599884 Chr.19: 549492-549492 on Build GRCh38
rs112722986 Chr.19: 549515-549515 on Build GRCh38
rs1042450493 Chr.19: 549849-549849 on Build GRCh38
rs762826023 Chr.19: 549834-549834 on Build GRCh38
rs780336298 Chr.19: 549630-549630 on Build GRCh38
rs753826167 Chr.19: 549761-549761 on Build GRCh38
rs750658853 Chr.19: 549680-549680 on Build GRCh38
rs771986372 Chr.19: 549838-549838 on Build GRCh38
rs769223757 Chr.19: 549738-549738 on Build GRCh38
rs994991540 Chr.19: 549946-549946 on Build GRCh38
rs935152288 Chr.19: 549803-549803 on Build GRCh38
rs1041243012 Chr.19: 549806-549806 on Build GRCh38
rs769996882 Chr.19: 549945-549945 on Build GRCh38
rs1599883 Chr.19: 549595-549595 on Build GRCh38
rs558079813 Chr.19: 549705-549705 on Build GRCh38
rs371890199 Chr.19: 549784-549784 on Build GRCh38
rs530579163 Chr.19: 549746-549746 on Build GRCh38
rs113273340 Chr.19: 549889-549889 on Build GRCh38
rs764048962 Chr.19: 549835-549835 on Build GRCh38
rs748000490 Chr.19: 549640-549640 on Build GRCh38
rs750710140 Chr.19: 549621-549621 on Build GRCh38
rs370750803 Chr.19: 549619-549619 on Build GRCh38
rs59539774 Chr.19: 549667-549667 on Build GRCh38
rs771673830 Chr.19: 549642-549642 on Build GRCh38
rs932930012 Chr.19: 549818-549818 on Build GRCh38
rs763308928 Chr.19: 549665-549665 on Build GRCh38
rs201561127 Chr.19: 549633-549633 on Build GRCh38

View Full Product Details