Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 603878-604396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCN2
Entrez Gene ID
610
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 2
Gene Aliases
BCNG-2, BCNG2, HAC-1
UniGene
Hs.124161
Gene Chromosome Location
Chr.19: 589893-617159 on Build GRCh38
Transcripts
NM_001194.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5435970 c.984C>T p.F328F Chr.19: 603895-603895 on Build GRCh38
6377653 c.1048T>G p.W350G Chr.19: 603959-603959 on Build GRCh38
5435971 c.1002C>A p.L334L Chr.19: 603913-603913 on Build GRCh38
1002821 c.979C>T p.R327C Chr.19: 603890-603890 on Build GRCh38
6226174 c.1034G>A p.R345H Chr.19: 603945-603945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146340939 Chr.19: 603934-603934 on Build GRCh38
rs140389473 Chr.19: 604327-604327 on Build GRCh38
rs57672134 Chr.19: 604190-604190 on Build GRCh38
rs59300335 Chr.19: 604197-604197 on Build GRCh38
rs756082561 Chr.19: 603974-603974 on Build GRCh38
rs1055501283 Chr.19: 604149-604149 on Build GRCh38
rs376159402 Chr.19: 603985-603985 on Build GRCh38
rs12973067 Chr.19: 604089-604089 on Build GRCh38
rs1032653660 Chr.19: 604127-604127 on Build GRCh38
rs182960665 Chr.19: 604304-604304 on Build GRCh38
rs763330197 Chr.19: 603922-603922 on Build GRCh38
rs767814420 Chr.19: 604012-604012 on Build GRCh38
rs564226585 Chr.19: 603985-603986 on Build GRCh38
rs768576209 Chr.19: 603986-603986 on Build GRCh38
rs753019727 Chr.19: 603920-603920 on Build GRCh38
rs1055968761 Chr.19: 603975-603975 on Build GRCh38
rs991237816 Chr.19: 604343-604343 on Build GRCh38
rs775990609 Chr.19: 604006-604006 on Build GRCh38
rs58064834 Chr.19: 604156-604156 on Build GRCh38
rs543617634 Chr.19: 604359-604359 on Build GRCh38
rs887063911 Chr.19: 603996-603996 on Build GRCh38
rs57519629 Chr.19: 604163-604163 on Build GRCh38
rs1038893637 Chr.19: 603992-603992 on Build GRCh38
rs757350744 Chr.19: 603943-603943 on Build GRCh38
rs564460148 Chr.19: 604048-604048 on Build GRCh38
rs59210872 Chr.19: 604164-604164 on Build GRCh38
rs57537998 Chr.19: 604205-604205 on Build GRCh38
rs553831274 Chr.19: 604021-604021 on Build GRCh38
rs770236849 Chr.19: 604005-604005 on Build GRCh38
rs1038338281 Chr.19: 603998-603998 on Build GRCh38
rs545357612 Chr.19: 604027-604027 on Build GRCh38
rs760013170 Chr.19: 603913-603913 on Build GRCh38
rs12981107 Chr.19: 604211-604211 on Build GRCh38
rs59535474 Chr.19: 604171-604171 on Build GRCh38
rs58503897 Chr.19: 604204-604204 on Build GRCh38
rs368986151 Chr.19: 603997-603997 on Build GRCh38
rs59313139 Chr.19: 604099-604099 on Build GRCh38
rs751789521 Chr.19: 604010-604010 on Build GRCh38
rs765553182 Chr.19: 603917-603917 on Build GRCh38
rs60627977 Chr.19: 604129-604129 on Build GRCh38
rs371982176 Chr.19: 604342-604342 on Build GRCh38
rs12980894 Chr.19: 604170-604170 on Build GRCh38
rs60680473 Chr.19: 604130-604130 on Build GRCh38
rs1048015858 Chr.19: 603931-603931 on Build GRCh38
rs781035673 Chr.19: 603975-603980 on Build GRCh38
rs574487685 Chr.19: 603995-603995 on Build GRCh38
rs780166639 Chr.19: 603981-603981 on Build GRCh38
rs58035646 Chr.19: 604151-604151 on Build GRCh38
rs562650221 Chr.19: 604293-604293 on Build GRCh38
rs747814781 Chr.19: 603994-603994 on Build GRCh38
rs1032707360 Chr.19: 604350-604350 on Build GRCh38
rs1009778548 Chr.19: 604059-604059 on Build GRCh38
rs764239250 Chr.19: 603928-603928 on Build GRCh38
rs1029782345 Chr.19: 604037-604037 on Build GRCh38
rs968258551 Chr.19: 604071-604071 on Build GRCh38
rs777183942 Chr.19: 603898-603898 on Build GRCh38
rs56078004 Chr.19: 604154-604154 on Build GRCh38
rs1021211335 Chr.19: 604060-604060 on Build GRCh38
rs763232737 Chr.19: 604007-604007 on Build GRCh38
rs201131143 Chr.19: 603971-603971 on Build GRCh38
rs775163400 Chr.19: 604011-604012 on Build GRCh38
rs12981115 Chr.19: 604222-604222 on Build GRCh38
rs936708984 Chr.19: 604140-604140 on Build GRCh38
rs57385698 Chr.19: 604115-604115 on Build GRCh38
rs781185447 Chr.19: 603958-603958 on Build GRCh38
rs899832922 Chr.19: 604011-604011 on Build GRCh38
rs557400402 Chr.19: 604064-604064 on Build GRCh38
rs1008687495 Chr.19: 603973-603973 on Build GRCh38
rs12980903 Chr.19: 604181-604181 on Build GRCh38
rs572629999 Chr.19: 604069-604069 on Build GRCh38
rs58581944 Chr.19: 604122-604122 on Build GRCh38
rs868748276 Chr.19: 604263-604263 on Build GRCh38
rs770688397 Chr.19: 603974-603974 on Build GRCh38
rs980096937 Chr.19: 604072-604072 on Build GRCh38
rs10409268 Chr.19: 604022-604022 on Build GRCh38
rs58220447 Chr.19: 604123-604123 on Build GRCh38
rs764505312 Chr.19: 604009-604009 on Build GRCh38
rs778705073 Chr.19: 603987-603987 on Build GRCh38
rs745549980 Chr.19: 603980-603980 on Build GRCh38

View Full Product Details