Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 616331-616859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCN2
Entrez Gene ID
610
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 2
Gene Aliases
BCNG-2, BCNG2, HAC-1
UniGene
Hs.124161
Gene Chromosome Location
Chr.19: 589893-617159 on Build GRCh38
Transcripts
NM_001194.3

SNP Detail

SNP ID Chromosome Location
rs983434550 Chr.19: 616361-616361 on Build GRCh38
rs959310355 Chr.19: 616350-616350 on Build GRCh38
rs1014575910 Chr.19: 616837-616837 on Build GRCh38
rs933505249 Chr.19: 616388-616388 on Build GRCh38
rs541952823 Chr.19: 616429-616429 on Build GRCh38
rs879888906 Chr.19: 616706-616706 on Build GRCh38
rs1042047460 Chr.19: 616501-616501 on Build GRCh38
rs950691169 Chr.19: 616357-616357 on Build GRCh38
rs959212277 Chr.19: 616753-616753 on Build GRCh38
rs922134842 Chr.19: 616378-616378 on Build GRCh38
rs865910456 Chr.19: 616508-616508 on Build GRCh38
rs893402945 Chr.19: 616640-616640 on Build GRCh38
rs999691553 Chr.19: 616664-616683 on Build GRCh38
rs917508821 Chr.19: 616759-616759 on Build GRCh38
rs1046215802 Chr.19: 616609-616609 on Build GRCh38
rs916290337 Chr.19: 616672-616672 on Build GRCh38
rs917842519 Chr.19: 616356-616356 on Build GRCh38
rs56291780 Chr.19: 616568-616568 on Build GRCh38
rs546915808 Chr.19: 616636-616636 on Build GRCh38
rs533527260 Chr.19: 616812-616812 on Build GRCh38
rs981727575 Chr.19: 616836-616836 on Build GRCh38
rs1016186302 Chr.19: 616545-616546 on Build GRCh38
rs55810748 Chr.19: 616509-616509 on Build GRCh38
rs899062581 Chr.19: 616699-616701 on Build GRCh38
rs920479331 Chr.19: 616786-616786 on Build GRCh38
rs771330705 Chr.19: 616370-616370 on Build GRCh38
rs1013354033 Chr.19: 616524-616524 on Build GRCh38
rs533345141 Chr.19: 616553-616553 on Build GRCh38
rs529839402 Chr.19: 616683-616683 on Build GRCh38
rs949060760 Chr.19: 616678-616678 on Build GRCh38
rs1051981474 Chr.19: 616395-616395 on Build GRCh38
rs1039196668 Chr.19: 616375-616375 on Build GRCh38
rs761681143 Chr.19: 616420-616420 on Build GRCh38
rs886234524 Chr.19: 616614-616614 on Build GRCh38
rs550827450 Chr.19: 616667-616667 on Build GRCh38
rs940477636 Chr.19: 616775-616775 on Build GRCh38
rs907641443 Chr.19: 616689-616689 on Build GRCh38
rs755732935 Chr.19: 616492-616492 on Build GRCh38
rs759760819 Chr.19: 616383-616383 on Build GRCh38
rs775586517 Chr.19: 616392-616392 on Build GRCh38
rs933445414 Chr.19: 616493-616493 on Build GRCh38
rs970339628 Chr.19: 616765-616765 on Build GRCh38
rs931810092 Chr.19: 616798-616798 on Build GRCh38
rs866146431 Chr.19: 616817-616817 on Build GRCh38
rs1004682418 Chr.19: 616615-616615 on Build GRCh38
rs527643702 Chr.19: 616574-616574 on Build GRCh38
rs963493143 Chr.19: 616476-616476 on Build GRCh38
rs913436518 Chr.19: 616564-616564 on Build GRCh38
rs987623949 Chr.19: 616562-616562 on Build GRCh38
rs894610402 Chr.19: 616827-616827 on Build GRCh38
rs1037991522 Chr.19: 616620-616620 on Build GRCh38
rs1041769788 Chr.19: 616810-616813 on Build GRCh38
rs967480546 Chr.19: 616717-616717 on Build GRCh38
rs913465932 Chr.19: 616496-616496 on Build GRCh38
rs1013517402 Chr.19: 616831-616831 on Build GRCh38
rs894914501 Chr.19: 616519-616519 on Build GRCh38
rs973635121 Chr.19: 616782-616782 on Build GRCh38
rs963672499 Chr.19: 616547-616547 on Build GRCh38
rs894939849 Chr.19: 616444-616444 on Build GRCh38
rs1024486891 Chr.19: 616833-616833 on Build GRCh38
rs56069324 Chr.19: 616595-616595 on Build GRCh38
rs1037562161 Chr.19: 616697-616697 on Build GRCh38
rs916318381 Chr.19: 616597-616597 on Build GRCh38
rs1020358787 Chr.19: 616712-616712 on Build GRCh38
rs1004796982 Chr.19: 616541-616541 on Build GRCh38
rs56357145 Chr.19: 616671-616671 on Build GRCh38
rs1016714692 Chr.19: 616475-616475 on Build GRCh38
rs967504970 Chr.19: 616661-616661 on Build GRCh38
rs996560073 Chr.19: 616635-616635 on Build GRCh38
rs565148977 Chr.19: 616639-616639 on Build GRCh38
rs751606466 Chr.19: 616409-616409 on Build GRCh38
rs890389958 Chr.19: 616801-616801 on Build GRCh38
rs944672520 Chr.19: 616809-616809 on Build GRCh38
rs946310583 Chr.19: 616497-616497 on Build GRCh38
rs1024796043 Chr.19: 616527-616527 on Build GRCh38
rs1035478679 Chr.19: 616824-616824 on Build GRCh38
rs532361528 Chr.19: 616659-616659 on Build GRCh38
rs56114824 Chr.19: 616611-616611 on Build GRCh38
rs1013386762 Chr.19: 616445-616445 on Build GRCh38
rs1033513763 Chr.19: 616435-616435 on Build GRCh38
rs975195009 Chr.19: 616484-616484 on Build GRCh38
rs926180899 Chr.19: 616664-616664 on Build GRCh38
rs886352184 Chr.19: 616531-616531 on Build GRCh38
rs974782383 Chr.19: 616549-616549 on Build GRCh38
rs750216734 Chr.19: 616432-616432 on Build GRCh38
rs747358302 Chr.19: 616367-616367 on Build GRCh38
rs1009024957 Chr.19: 616644-616644 on Build GRCh38
rs55703451 Chr.19: 616701-616701 on Build GRCh38
rs1050299126 Chr.19: 616799-616799 on Build GRCh38
rs936311996 Chr.19: 616586-616586 on Build GRCh38
rs940494626 Chr.19: 616690-616690 on Build GRCh38
rs947604075 Chr.19: 616651-616651 on Build GRCh38
rs1050414411 Chr.19: 616702-616702 on Build GRCh38
rs987654819 Chr.19: 616494-616494 on Build GRCh38
rs992075582 Chr.19: 616355-616355 on Build GRCh38
rs991718762 Chr.19: 616754-616754 on Build GRCh38
rs954798485 Chr.19: 616556-616556 on Build GRCh38
rs942141604 Chr.19: 616369-616369 on Build GRCh38
rs904046835 Chr.19: 616634-616644 on Build GRCh38
rs1055255306 Chr.19: 616514-616514 on Build GRCh38
rs907610436 Chr.19: 616770-616770 on Build GRCh38
rs1024825486 Chr.19: 616456-616456 on Build GRCh38
rs776681778 Chr.19: 616372-616372 on Build GRCh38
rs1020381905 Chr.19: 616657-616657 on Build GRCh38
rs868829990 Chr.19: 616605-616605 on Build GRCh38
rs971643647 Chr.19: 616834-616834 on Build GRCh38
rs1033216599 Chr.19: 616738-616738 on Build GRCh38
rs962097384 Chr.19: 616838-616838 on Build GRCh38
rs1042099295 Chr.19: 616412-616412 on Build GRCh38
rs954767778 Chr.19: 616642-616642 on Build GRCh38
rs979383040 Chr.19: 616572-616572 on Build GRCh38
rs903588933 Chr.19: 616417-616417 on Build GRCh38
rs1033958981 Chr.19: 616349-616349 on Build GRCh38
rs1000759469 Chr.19: 616722-616722 on Build GRCh38
rs983406508 Chr.19: 616469-616469 on Build GRCh38
rs981758840 Chr.19: 616766-616767 on Build GRCh38
rs764125612 Chr.19: 616399-616399 on Build GRCh38
rs996142026 Chr.19: 616699-616699 on Build GRCh38
rs950659914 Chr.19: 616459-616459 on Build GRCh38
rs1033515819 Chr.19: 616825-616825 on Build GRCh38
rs1009211475 Chr.19: 616402-616402 on Build GRCh38
rs1008909178 Chr.19: 616707-616707 on Build GRCh38
rs1000324973 Chr.19: 616816-616816 on Build GRCh38
rs55760240 Chr.19: 616688-616688 on Build GRCh38
rs1000687055 Chr.19: 616348-616348 on Build GRCh38

View Full Product Details