Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1105005-1105513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPX4
Entrez Gene ID
2879
Gene Name
glutathione peroxidase 4
Gene Aliases
GPx-4, GSHPx-4, MCSP, PHGPx, SMDS, snGPx, snPHGPx
UniGene
Hs.433951
Gene Chromosome Location
Chr.19: 1103926-1106789 on Build GRCh38
Transcripts
NM_001039847.2 NM_001039848.2 NM_002085.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5854598 c.265C>T p.R89* Chr.19: 1105451-1105451 on Build GRCh38
4580600 c.157A>G p.M53V Chr.19: 1105258-1105258 on Build GRCh38
6205618 c.90G>T p.A30A Chr.19: 1105191-1105191 on Build GRCh38
5476022 c.148G>A p.D50N Chr.19: 1105249-1105249 on Build GRCh38
3668178 c.138C>A p.A46A Chr.19: 1105239-1105239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760039914 Chr.19: 1105462-1105462 on Build GRCh38
rs756485267 Chr.19: 1105279-1105279 on Build GRCh38
rs764426541 Chr.19: 1105331-1105331 on Build GRCh38
rs763783774 Chr.19: 1105256-1105256 on Build GRCh38
rs765117498 Chr.19: 1105149-1105149 on Build GRCh38
rs770740384 Chr.19: 1105319-1105319 on Build GRCh38
rs752167709 Chr.19: 1105432-1105432 on Build GRCh38
rs1025575075 Chr.19: 1105092-1105092 on Build GRCh38
rs780173011 Chr.19: 1105445-1105445 on Build GRCh38
rs753179152 Chr.19: 1105288-1105288 on Build GRCh38
rs201903291 Chr.19: 1105386-1105386 on Build GRCh38
rs750939686 Chr.19: 1105349-1105349 on Build GRCh38
rs772198561 Chr.19: 1105385-1105385 on Build GRCh38
rs945662789 Chr.19: 1105222-1105222 on Build GRCh38
rs757673527 Chr.19: 1105343-1105343 on Build GRCh38
rs778651341 Chr.19: 1105369-1105369 on Build GRCh38
rs762088801 Chr.19: 1105455-1105455 on Build GRCh38
rs974879773 Chr.19: 1105124-1105124 on Build GRCh38
rs756571473 Chr.19: 1105200-1105200 on Build GRCh38
rs768267697 Chr.19: 1105227-1105227 on Build GRCh38
rs530063614 Chr.19: 1105122-1105122 on Build GRCh38
rs746579938 Chr.19: 1105225-1105225 on Build GRCh38
rs374349562 Chr.19: 1105473-1105473 on Build GRCh38
rs548528852 Chr.19: 1105123-1105123 on Build GRCh38
rs754504650 Chr.19: 1105295-1105295 on Build GRCh38
rs754288488 Chr.19: 1105286-1105286 on Build GRCh38
rs765611709 Chr.19: 1105348-1105348 on Build GRCh38
rs759634320 Chr.19: 1105325-1105325 on Build GRCh38
rs981510076 Chr.19: 1105452-1105452 on Build GRCh38
rs760877738 Chr.19: 1105389-1105389 on Build GRCh38
rs755330241 Chr.19: 1105029-1105029 on Build GRCh38
rs778748373 Chr.19: 1105483-1105483 on Build GRCh38
rs983475978 Chr.19: 1105113-1105113 on Build GRCh38
rs199743575 Chr.19: 1105435-1105435 on Build GRCh38
rs545598881 Chr.19: 1105091-1105091 on Build GRCh38
rs374238675 Chr.19: 1105387-1105387 on Build GRCh38
rs754417099 Chr.19: 1105355-1105355 on Build GRCh38
rs571770278 Chr.19: 1105057-1105057 on Build GRCh38
rs763031415 Chr.19: 1105164-1105164 on Build GRCh38
rs749154135 Chr.19: 1105317-1105317 on Build GRCh38
rs8178971 Chr.19: 1105363-1105363 on Build GRCh38
rs745799475 Chr.19: 1105373-1105373 on Build GRCh38
rs566974741 Chr.19: 1105237-1105237 on Build GRCh38
rs575039682 Chr.19: 1105450-1105450 on Build GRCh38
rs1011734962 Chr.19: 1105059-1105059 on Build GRCh38
rs776936631 Chr.19: 1105406-1105406 on Build GRCh38
rs201951088 Chr.19: 1105460-1105460 on Build GRCh38
rs747887804 Chr.19: 1105249-1105249 on Build GRCh38
rs8178969 Chr.19: 1105100-1105100 on Build GRCh38
rs376432780 Chr.19: 1105384-1105384 on Build GRCh38
rs550603355 Chr.19: 1105312-1105312 on Build GRCh38
rs77421492 Chr.19: 1105364-1105364 on Build GRCh38
rs778370256 Chr.19: 1105203-1105203 on Build GRCh38
rs745707351 Chr.19: 1105443-1105443 on Build GRCh38
rs961334143 Chr.19: 1105130-1105130 on Build GRCh38
rs766374099 Chr.19: 1105178-1105178 on Build GRCh38
rs11555179 Chr.19: 1105449-1105449 on Build GRCh38
rs373138217 Chr.19: 1105314-1105314 on Build GRCh38
rs913439103 Chr.19: 1105424-1105424 on Build GRCh38
rs747013916 Chr.19: 1105447-1105447 on Build GRCh38
rs770775625 Chr.19: 1105367-1105367 on Build GRCh38
rs761699782 Chr.19: 1105437-1105438 on Build GRCh38
rs952819943 Chr.19: 1105211-1105211 on Build GRCh38
rs370419921 Chr.19: 1105451-1105451 on Build GRCh38
rs772104779 Chr.19: 1105444-1105444 on Build GRCh38
rs376929853 Chr.19: 1105270-1105270 on Build GRCh38
rs898252784 Chr.19: 1105486-1105486 on Build GRCh38
rs779416718 Chr.19: 1105214-1105214 on Build GRCh38
rs771535901 Chr.19: 1105210-1105210 on Build GRCh38
rs777518288 Chr.19: 1105361-1105361 on Build GRCh38
rs760967203 Chr.19: 1105330-1105330 on Build GRCh38
rs769967246 Chr.19: 1105456-1105456 on Build GRCh38
rs776390793 Chr.19: 1105137-1105137 on Build GRCh38
rs376032528 Chr.19: 1105316-1105316 on Build GRCh38
rs376007177 Chr.19: 1105439-1105439 on Build GRCh38
rs370119671 Chr.19: 1105328-1105328 on Build GRCh38
rs200071465 Chr.19: 1105411-1105411 on Build GRCh38
rs372370002 Chr.19: 1105236-1105236 on Build GRCh38
rs766916114 Chr.19: 1105415-1105415 on Build GRCh38
rs755235087 Chr.19: 1105195-1105195 on Build GRCh38
rs750853594 Chr.19: 1105414-1105414 on Build GRCh38
rs369712159 Chr.19: 1105196-1105196 on Build GRCh38
rs766828184 Chr.19: 1105459-1105459 on Build GRCh38
rs750347551 Chr.19: 1105157-1105157 on Build GRCh38
rs771121995 Chr.19: 1105062-1105062 on Build GRCh38
rs761637110 Chr.19: 1105144-1105144 on Build GRCh38
rs561995303 Chr.19: 1105106-1105106 on Build GRCh38
rs8178970 Chr.19: 1105311-1105311 on Build GRCh38
rs780763270 Chr.19: 1105296-1105296 on Build GRCh38
rs937006628 Chr.19: 1105458-1105458 on Build GRCh38
rs200859501 Chr.19: 1105259-1105259 on Build GRCh38
rs913501602 Chr.19: 1105166-1105166 on Build GRCh38
rs545537200 Chr.19: 1105489-1105489 on Build GRCh38
rs749766545 Chr.19: 1105207-1105207 on Build GRCh38
rs536607574 Chr.19: 1105457-1105457 on Build GRCh38
rs778371843 Chr.19: 1105388-1105388 on Build GRCh38
rs756916315 Chr.19: 1105478-1105478 on Build GRCh38
rs994139572 Chr.19: 1105133-1105133 on Build GRCh38
rs995860594 Chr.19: 1105074-1105074 on Build GRCh38
rs367564504 Chr.19: 1105488-1105488 on Build GRCh38
rs373533365 Chr.19: 1105359-1105359 on Build GRCh38
rs778842247 Chr.19: 1105442-1105442 on Build GRCh38
rs751317795 Chr.19: 1105322-1105323 on Build GRCh38
rs1045408258 Chr.19: 1105226-1105226 on Build GRCh38
rs563988259 Chr.19: 1105094-1105094 on Build GRCh38
rs769589678 Chr.19: 1105253-1105253 on Build GRCh38
rs756929751 Chr.19: 1105441-1105441 on Build GRCh38
rs768722652 Chr.19: 1105395-1105395 on Build GRCh38
rs773134735 Chr.19: 1105256-1105256 on Build GRCh38
rs753350196 Chr.19: 1105474-1105474 on Build GRCh38
rs922127639 Chr.19: 1105153-1105153 on Build GRCh38
rs752983685 Chr.19: 1105276-1105276 on Build GRCh38
rs779516356 Chr.19: 1105287-1105287 on Build GRCh38
rs549843877 Chr.19: 1105103-1105103 on Build GRCh38
rs762792092 Chr.19: 1105258-1105258 on Build GRCh38
rs556598510 Chr.19: 1105412-1105412 on Build GRCh38
rs751723961 Chr.19: 1105186-1105186 on Build GRCh38
rs754121801 Chr.19: 1105333-1105333 on Build GRCh38
rs1055087141 Chr.19: 1105158-1105158 on Build GRCh38
rs1027012803 Chr.19: 1105171-1105172 on Build GRCh38
rs777236046 Chr.19: 1105436-1105436 on Build GRCh38
rs780675386 Chr.19: 1105358-1105358 on Build GRCh38

View Full Product Details