Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6380137-6380629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2F1
Entrez Gene ID
2962
Gene Name
general transcription factor IIF subunit 1
Gene Aliases
BTF4, RAP74, TF2F1, TFIIF
UniGene
Hs.68257
Gene Chromosome Location
Chr.19: 6379569-6393280 on Build GRCh38
Transcripts
NM_002096.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6487865 c.1525G>T p.D509Y Chr.19: 6380310-6380310 on Build GRCh38
4526151 c.1375G>A p.V459M Chr.19: 6380460-6380460 on Build GRCh38
1002874 c.1379G>A p.R460H Chr.19: 6380456-6380456 on Build GRCh38
2727438 c.1294C>A p.L432M Chr.19: 6380628-6380628 on Build GRCh38
5581272 c.1407C>T p.T469T Chr.19: 6380428-6380428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776430368 Chr.19: 6380599-6380599 on Build GRCh38
rs45483994 Chr.19: 6380524-6380524 on Build GRCh38
rs532144980 Chr.19: 6380466-6380466 on Build GRCh38
rs746224485 Chr.19: 6380362-6380362 on Build GRCh38
rs367874489 Chr.19: 6380239-6380239 on Build GRCh38
rs949605746 Chr.19: 6380548-6380548 on Build GRCh38
rs776196364 Chr.19: 6380511-6380511 on Build GRCh38
rs761413505 Chr.19: 6380609-6380609 on Build GRCh38
rs985777820 Chr.19: 6380203-6380203 on Build GRCh38
rs756438444 Chr.19: 6380574-6380574 on Build GRCh38
rs779379265 Chr.19: 6380252-6380252 on Build GRCh38
rs766958129 Chr.19: 6380541-6380541 on Build GRCh38
rs763149040 Chr.19: 6380181-6380181 on Build GRCh38
rs751866107 Chr.19: 6380551-6380551 on Build GRCh38
rs755816393 Chr.19: 6380350-6380350 on Build GRCh38
rs749748984 Chr.19: 6380254-6380254 on Build GRCh38
rs774978790 Chr.19: 6380558-6380560 on Build GRCh38
rs200104282 Chr.19: 6380502-6380502 on Build GRCh38
rs931827214 Chr.19: 6380182-6380182 on Build GRCh38
rs1041444132 Chr.19: 6380213-6380213 on Build GRCh38
rs752571667 Chr.19: 6380449-6380449 on Build GRCh38
rs749978997 Chr.19: 6380526-6380526 on Build GRCh38
rs778190675 Chr.19: 6380251-6380252 on Build GRCh38
rs199893553 Chr.19: 6380361-6380361 on Build GRCh38
rs780478511 Chr.19: 6380392-6380392 on Build GRCh38
rs962109702 Chr.19: 6380171-6380171 on Build GRCh38
rs568273336 Chr.19: 6380495-6380495 on Build GRCh38
rs759532759 Chr.19: 6380433-6380433 on Build GRCh38
rs768953273 Chr.19: 6380407-6380407 on Build GRCh38
rs200973251 Chr.19: 6380488-6380488 on Build GRCh38
rs202050363 Chr.19: 6380575-6380575 on Build GRCh38
rs1027436046 Chr.19: 6380377-6380377 on Build GRCh38
rs536272120 Chr.19: 6380175-6380175 on Build GRCh38
rs372430323 Chr.19: 6380552-6380552 on Build GRCh38
rs919502860 Chr.19: 6380547-6380547 on Build GRCh38
rs752236063 Chr.19: 6380349-6380349 on Build GRCh38
rs753390449 Chr.19: 6380354-6380354 on Build GRCh38
rs767170843 Chr.19: 6380448-6380448 on Build GRCh38
rs758799388 Chr.19: 6380534-6380534 on Build GRCh38
rs575662882 Chr.19: 6380594-6380594 on Build GRCh38
rs911847281 Chr.19: 6380194-6380194 on Build GRCh38
rs781700950 Chr.19: 6380562-6380562 on Build GRCh38
rs150223769 Chr.19: 6380481-6380481 on Build GRCh38
rs770988666 Chr.19: 6380426-6380426 on Build GRCh38
rs201709498 Chr.19: 6380501-6380501 on Build GRCh38
rs781628968 Chr.19: 6380482-6380482 on Build GRCh38
rs763789216 Chr.19: 6380455-6380455 on Build GRCh38
rs372092692 Chr.19: 6380329-6380329 on Build GRCh38
rs17625356 Chr.19: 6380458-6380458 on Build GRCh38
rs747691288 Chr.19: 6380240-6380240 on Build GRCh38
rs560145579 Chr.19: 6380460-6380460 on Build GRCh38
rs774365042 Chr.19: 6380432-6380432 on Build GRCh38
rs776821239 Chr.19: 6380416-6380416 on Build GRCh38
rs753328893 Chr.19: 6380456-6380456 on Build GRCh38
rs773451010 Chr.19: 6380489-6380489 on Build GRCh38
rs948136797 Chr.19: 6380529-6380529 on Build GRCh38
rs769355414 Chr.19: 6380553-6380553 on Build GRCh38
rs777359292 Chr.19: 6380247-6380247 on Build GRCh38
rs749438643 Chr.19: 6380576-6380576 on Build GRCh38
rs1023748926 Chr.19: 6380287-6380287 on Build GRCh38
rs568959027 Chr.19: 6380485-6380485 on Build GRCh38
rs746277546 Chr.19: 6380274-6380274 on Build GRCh38
rs745399854 Chr.19: 6380296-6380297 on Build GRCh38
rs149599017 Chr.19: 6380339-6380339 on Build GRCh38
rs545144327 Chr.19: 6380200-6380200 on Build GRCh38
rs376701000 Chr.19: 6380457-6380457 on Build GRCh38
rs138224011 Chr.19: 6380193-6380193 on Build GRCh38
rs944638809 Chr.19: 6380212-6380212 on Build GRCh38
rs762042946 Chr.19: 6380419-6380419 on Build GRCh38
rs747440550 Chr.19: 6380472-6380472 on Build GRCh38
rs1045212532 Chr.19: 6380538-6380538 on Build GRCh38
rs754798149 Chr.19: 6380332-6380332 on Build GRCh38
rs760217721 Chr.19: 6380452-6380452 on Build GRCh38
rs45466997 Chr.19: 6380490-6380490 on Build GRCh38
rs573003291 Chr.19: 6380199-6380199 on Build GRCh38
rs146571246 Chr.19: 6380324-6380324 on Build GRCh38
rs973148648 Chr.19: 6380174-6380174 on Build GRCh38
rs982242610 Chr.19: 6380556-6380556 on Build GRCh38
rs772474765 Chr.19: 6380250-6380251 on Build GRCh38
rs771895027 Chr.19: 6380578-6380578 on Build GRCh38
rs774671746 Chr.19: 6380261-6380261 on Build GRCh38
rs772320932 Chr.19: 6380286-6380286 on Build GRCh38
rs192571038 Chr.19: 6380353-6380353 on Build GRCh38
rs370032823 Chr.19: 6380461-6380461 on Build GRCh38
rs558616331 Chr.19: 6380224-6380224 on Build GRCh38
rs893872946 Chr.19: 6380256-6380256 on Build GRCh38
rs775638504 Chr.19: 6380303-6380303 on Build GRCh38
rs772256816 Chr.19: 6380376-6380376 on Build GRCh38
rs747149114 Chr.19: 6380400-6380400 on Build GRCh38
rs369704738 Chr.19: 6380311-6380311 on Build GRCh38
rs775560246 Chr.19: 6380580-6380580 on Build GRCh38
rs111666835 Chr.19: 6380504-6380504 on Build GRCh38
rs748734190 Chr.19: 6380255-6380255 on Build GRCh38
rs760953612 Chr.19: 6380310-6380310 on Build GRCh38
rs893925186 Chr.19: 6380528-6380528 on Build GRCh38
rs766468930 Chr.19: 6380325-6380325 on Build GRCh38
rs1000029043 Chr.19: 6380251-6380251 on Build GRCh38
rs375548583 Chr.19: 6380563-6380563 on Build GRCh38
rs746921999 Chr.19: 6380587-6380587 on Build GRCh38

View Full Product Details