Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6380762-6381279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2F1
Entrez Gene ID
2962
Gene Name
general transcription factor IIF subunit 1
Gene Aliases
BTF4, RAP74, TF2F1, TFIIF
UniGene
Hs.68257
Gene Chromosome Location
Chr.19: 6379569-6393280 on Build GRCh38
Transcripts
NM_002096.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1397269 c.1168C>T p.P390S Chr.19: 6380967-6380967 on Build GRCh38
1397270 c.1103C>T p.T368M Chr.19: 6381032-6381032 on Build GRCh38
1291036 c.1146G>T p.R382S Chr.19: 6380989-6380989 on Build GRCh38
3797659 c.1064G>A p.S355N Chr.19: 6381150-6381150 on Build GRCh38
6412235 c.1090G>T p.A364S Chr.19: 6381124-6381124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770531901 Chr.19: 6380883-6380883 on Build GRCh38
rs745472895 Chr.19: 6381045-6381045 on Build GRCh38
rs200336819 Chr.19: 6381199-6381199 on Build GRCh38
rs373030598 Chr.19: 6381188-6381188 on Build GRCh38
rs773225692 Chr.19: 6381031-6381033 on Build GRCh38
rs746526459 Chr.19: 6381107-6381107 on Build GRCh38
rs910459568 Chr.19: 6381001-6381001 on Build GRCh38
rs767636409 Chr.19: 6381093-6381093 on Build GRCh38
rs575943672 Chr.19: 6381161-6381161 on Build GRCh38
rs112259604 Chr.19: 6381121-6381121 on Build GRCh38
rs760675173 Chr.19: 6381033-6381035 on Build GRCh38
rs752299707 Chr.19: 6381187-6381187 on Build GRCh38
rs765088198 Chr.19: 6381100-6381100 on Build GRCh38
rs771134395 Chr.19: 6381148-6381148 on Build GRCh38
rs778868830 Chr.19: 6381042-6381042 on Build GRCh38
rs1000091228 Chr.19: 6381231-6381231 on Build GRCh38
rs995051190 Chr.19: 6380854-6380854 on Build GRCh38
rs751676609 Chr.19: 6381023-6381023 on Build GRCh38
rs189869674 Chr.19: 6381182-6381182 on Build GRCh38
rs773356588 Chr.19: 6381007-6381007 on Build GRCh38
rs766404728 Chr.19: 6381015-6381015 on Build GRCh38
rs746552099 Chr.19: 6381057-6381057 on Build GRCh38
rs112441441 Chr.19: 6381120-6381120 on Build GRCh38
rs781164089 Chr.19: 6380856-6380856 on Build GRCh38
rs143330643 Chr.19: 6380840-6380840 on Build GRCh38
rs199570172 Chr.19: 6381242-6381242 on Build GRCh38
rs148354538 Chr.19: 6381149-6381149 on Build GRCh38
rs748935468 Chr.19: 6380857-6380858 on Build GRCh38
rs779681975 Chr.19: 6381051-6381051 on Build GRCh38
rs369743152 Chr.19: 6381198-6381198 on Build GRCh38
rs772539013 Chr.19: 6381023-6381024 on Build GRCh38
rs777032976 Chr.19: 6381002-6381002 on Build GRCh38
rs943598007 Chr.19: 6381220-6381220 on Build GRCh38
rs761280869 Chr.19: 6380920-6380920 on Build GRCh38
rs375498391 Chr.19: 6381086-6381086 on Build GRCh38
rs748693442 Chr.19: 6381070-6381070 on Build GRCh38
rs1045104687 Chr.19: 6380822-6380822 on Build GRCh38
rs201144023 Chr.19: 6380927-6380927 on Build GRCh38
rs372773954 Chr.19: 6381065-6381065 on Build GRCh38
rs761629237 Chr.19: 6380988-6380997 on Build GRCh38
rs753656181 Chr.19: 6381094-6381094 on Build GRCh38
rs146231205 Chr.19: 6380872-6380873 on Build GRCh38
rs201244435 Chr.19: 6381115-6381115 on Build GRCh38
rs115426992 Chr.19: 6380824-6380824 on Build GRCh38
rs1037886497 Chr.19: 6380848-6380848 on Build GRCh38
rs143977043 Chr.19: 6380979-6380979 on Build GRCh38
rs758541092 Chr.19: 6380942-6380942 on Build GRCh38
rs557666933 Chr.19: 6380945-6380945 on Build GRCh38
rs866198431 Chr.19: 6381024-6381024 on Build GRCh38
rs754188897 Chr.19: 6380925-6380925 on Build GRCh38
rs780535392 Chr.19: 6381180-6381180 on Build GRCh38
rs939565132 Chr.19: 6380825-6380825 on Build GRCh38
rs773978756 Chr.19: 6380886-6380886 on Build GRCh38
rs769574746 Chr.19: 6380875-6380875 on Build GRCh38
rs143281847 Chr.19: 6381194-6381194 on Build GRCh38
rs73563879 Chr.19: 6381134-6381134 on Build GRCh38
rs186727244 Chr.19: 6380813-6380813 on Build GRCh38
rs764473497 Chr.19: 6380921-6380921 on Build GRCh38
rs368046275 Chr.19: 6380962-6380962 on Build GRCh38
rs561964456 Chr.19: 6380812-6380812 on Build GRCh38
rs765442181 Chr.19: 6381005-6381005 on Build GRCh38
rs762999068 Chr.19: 6381233-6381233 on Build GRCh38
rs772698034 Chr.19: 6380906-6380906 on Build GRCh38
rs761716206 Chr.19: 6380992-6380992 on Build GRCh38
rs1025982402 Chr.19: 6380897-6380897 on Build GRCh38
rs766631327 Chr.19: 6381082-6381083 on Build GRCh38
rs910838971 Chr.19: 6381207-6381207 on Build GRCh38
rs780120435 Chr.19: 6380943-6380943 on Build GRCh38
rs770324925 Chr.19: 6381123-6381123 on Build GRCh38
rs370397042 Chr.19: 6380930-6380930 on Build GRCh38
rs777274532 Chr.19: 6381246-6381246 on Build GRCh38
rs749797850 Chr.19: 6381190-6381190 on Build GRCh38
rs753746645 Chr.19: 6380947-6380947 on Build GRCh38
rs763129543 Chr.19: 6381081-6381081 on Build GRCh38
rs775166072 Chr.19: 6380978-6380978 on Build GRCh38
rs148360187 Chr.19: 6381046-6381046 on Build GRCh38
rs757286429 Chr.19: 6381097-6381097 on Build GRCh38
rs777452377 Chr.19: 6380882-6380882 on Build GRCh38
rs147291523 Chr.19: 6381032-6381032 on Build GRCh38
rs777072677 Chr.19: 6381068-6381068 on Build GRCh38
rs377485256 Chr.19: 6381236-6381236 on Build GRCh38
rs761714995 Chr.19: 6380894-6380894 on Build GRCh38
rs182194001 Chr.19: 6380794-6380794 on Build GRCh38
rs755552195 Chr.19: 6381245-6381245 on Build GRCh38
rs758103933 Chr.19: 6381101-6381101 on Build GRCh38
rs374734306 Chr.19: 6380893-6380893 on Build GRCh38
rs770601677 Chr.19: 6380968-6380968 on Build GRCh38
rs759814022 Chr.19: 6381150-6381150 on Build GRCh38
rs201652392 Chr.19: 6381137-6381137 on Build GRCh38
rs1053304122 Chr.19: 6381253-6381253 on Build GRCh38
rs773597792 Chr.19: 6381075-6381075 on Build GRCh38
rs537844648 Chr.19: 6381243-6381243 on Build GRCh38
rs371917624 Chr.19: 6380971-6380971 on Build GRCh38
rs868843814 Chr.19: 6381058-6381058 on Build GRCh38
rs777591705 Chr.19: 6380961-6380961 on Build GRCh38
rs745515448 Chr.19: 6380969-6380969 on Build GRCh38
rs200785566 Chr.19: 6380987-6380987 on Build GRCh38
rs762066559 Chr.19: 6380926-6380926 on Build GRCh38
rs915202365 Chr.19: 6380811-6380811 on Build GRCh38
rs756054151 Chr.19: 6381031-6381031 on Build GRCh38
rs773217883 Chr.19: 6381225-6381225 on Build GRCh38
rs935908431 Chr.19: 6380793-6380793 on Build GRCh38
rs769460685 Chr.19: 6380931-6380931 on Build GRCh38
rs757961315 Chr.19: 6381214-6381214 on Build GRCh38
rs770095830 Chr.19: 6381071-6381071 on Build GRCh38
rs376779199 Chr.19: 6381155-6381155 on Build GRCh38
rs756931564 Chr.19: 6381035-6381035 on Build GRCh38
rs200428042 Chr.19: 6381235-6381235 on Build GRCh38
rs527508942 Chr.19: 6381193-6381193 on Build GRCh38
rs769086537 Chr.19: 6380980-6380980 on Build GRCh38
rs748563388 Chr.19: 6381122-6381122 on Build GRCh38
rs762909769 Chr.19: 6381164-6381164 on Build GRCh38
rs113322069 Chr.19: 6381102-6381102 on Build GRCh38
rs199758892 Chr.19: 6381087-6381087 on Build GRCh38
rs765317059 Chr.19: 6381160-6381160 on Build GRCh38
rs774038708 Chr.19: 6380938-6380940 on Build GRCh38
rs759088805 Chr.19: 6381240-6381240 on Build GRCh38
rs115294901 Chr.19: 6381200-6381200 on Build GRCh38
rs898117908 Chr.19: 6380845-6380845 on Build GRCh38
rs747933162 Chr.19: 6380871-6380871 on Build GRCh38
rs375189684 Chr.19: 6381014-6381014 on Build GRCh38
rs753921613 Chr.19: 6381138-6381140 on Build GRCh38
rs370175450 Chr.19: 6381203-6381203 on Build GRCh38
rs763184642 Chr.19: 6381008-6381008 on Build GRCh38
rs753659161 Chr.19: 6381033-6381033 on Build GRCh38
rs760443858 Chr.19: 6381080-6381081 on Build GRCh38
rs768334020 Chr.19: 6380858-6380863 on Build GRCh38
rs200961335 Chr.19: 6381172-6381172 on Build GRCh38
rs375204831 Chr.19: 6380960-6380960 on Build GRCh38
rs776078092 Chr.19: 6380911-6380911 on Build GRCh38
rs754380951 Chr.19: 6381108-6381108 on Build GRCh38
rs779431420 Chr.19: 6380876-6380876 on Build GRCh38
rs761814202 Chr.19: 6381157-6381157 on Build GRCh38
rs888753031 Chr.19: 6380901-6380901 on Build GRCh38
rs766201648 Chr.19: 6381169-6381169 on Build GRCh38
rs892874833 Chr.19: 6381259-6381259 on Build GRCh38
rs754690722 Chr.19: 6381179-6381179 on Build GRCh38

View Full Product Details