Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43671548-43672084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM4A
Entrez Gene ID
9682
Gene Name
lysine demethylase 4A
Gene Aliases
JHDM3A, JMJD2, JMJD2A, TDRD14A
UniGene
Hs.155983
Gene Chromosome Location
Chr.1: 43650118-43705518 on Build GRCh38
Transcripts
NM_014663.2 XM_005271354.3 XM_005271355.3 XM_005271356.3 XM_011542467.2 XM_017002912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7131573 c.1553G>T p.R518L Chr.1: 43671694-43671694 on Build GRCh38
6692387 c.1701C>T p.A567A Chr.1: 43671842-43671842 on Build GRCh38
7126879 c.1534C>T p.L512L Chr.1: 43671675-43671675 on Build GRCh38
5922042 c.1555G>A p.D519N Chr.1: 43671696-43671696 on Build GRCh38
1196531 c.1529C>G p.S510C Chr.1: 43671670-43671670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750198893 Chr.1: 43671624-43671624 on Build GRCh38
rs373467522 Chr.1: 43671648-43671648 on Build GRCh38
rs764064619 Chr.1: 43671607-43671607 on Build GRCh38
rs750358339 Chr.1: 43671705-43671705 on Build GRCh38
rs767686593 Chr.1: 43671588-43671588 on Build GRCh38
rs527454755 Chr.1: 43672046-43672046 on Build GRCh38
rs199980633 Chr.1: 43671914-43671914 on Build GRCh38
rs138721228 Chr.1: 43671619-43671619 on Build GRCh38
rs368103707 Chr.1: 43671913-43671929 on Build GRCh38
rs773486048 Chr.1: 43671827-43671827 on Build GRCh38
rs899558059 Chr.1: 43671824-43671824 on Build GRCh38
rs774631250 Chr.1: 43671577-43671577 on Build GRCh38
rs375615103 Chr.1: 43671633-43671633 on Build GRCh38
rs991423814 Chr.1: 43672061-43672061 on Build GRCh38
rs879068780 Chr.1: 43671907-43671907 on Build GRCh38
rs775542197 Chr.1: 43671592-43671592 on Build GRCh38
rs201313043 Chr.1: 43671647-43671647 on Build GRCh38
rs150381773 Chr.1: 43671735-43671735 on Build GRCh38
rs942144181 Chr.1: 43671671-43671671 on Build GRCh38
rs201318621 Chr.1: 43671649-43671649 on Build GRCh38
rs769037274 Chr.1: 43671899-43671899 on Build GRCh38
rs763357725 Chr.1: 43671574-43671574 on Build GRCh38
rs868527797 Chr.1: 43671647-43671650 on Build GRCh38
rs192175863 Chr.1: 43671757-43671757 on Build GRCh38
rs1031181212 Chr.1: 43672021-43672021 on Build GRCh38
rs766075877 Chr.1: 43671725-43671725 on Build GRCh38
rs759399492 Chr.1: 43671842-43671842 on Build GRCh38
rs185752390 Chr.1: 43671813-43671813 on Build GRCh38
rs775648431 Chr.1: 43671782-43671782 on Build GRCh38
rs780911853 Chr.1: 43671745-43671745 on Build GRCh38
rs746359582 Chr.1: 43671769-43671769 on Build GRCh38
rs142569925 Chr.1: 43671749-43671749 on Build GRCh38
rs538082991 Chr.1: 43671693-43671693 on Build GRCh38
rs747080315 Chr.1: 43671646-43671647 on Build GRCh38
rs752324740 Chr.1: 43671847-43671847 on Build GRCh38
rs373296726 Chr.1: 43671616-43671616 on Build GRCh38
rs758770894 Chr.1: 43671880-43671880 on Build GRCh38
rs586339 Chr.1: 43671586-43671586 on Build GRCh38
rs961135515 Chr.1: 43672035-43672035 on Build GRCh38
rs753347514 Chr.1: 43671870-43671870 on Build GRCh38
rs142484784 Chr.1: 43671668-43671668 on Build GRCh38
rs1035939282 Chr.1: 43671931-43671931 on Build GRCh38
rs368052422 Chr.1: 43671843-43671843 on Build GRCh38
rs758462057 Chr.1: 43671708-43671708 on Build GRCh38
rs776946990 Chr.1: 43671817-43671817 on Build GRCh38
rs376335983 Chr.1: 43671742-43671742 on Build GRCh38
rs777260014 Chr.1: 43671861-43671861 on Build GRCh38
rs750147690 Chr.1: 43671829-43671829 on Build GRCh38
rs754106082 Chr.1: 43672002-43672002 on Build GRCh38
rs766346614 Chr.1: 43671839-43671839 on Build GRCh38
rs769715632 Chr.1: 43671910-43671926 on Build GRCh38
rs943818142 Chr.1: 43671862-43671862 on Build GRCh38
rs760799528 Chr.1: 43671694-43671694 on Build GRCh38
rs190275733 Chr.1: 43671840-43671840 on Build GRCh38
rs201742445 Chr.1: 43671912-43671912 on Build GRCh38
rs777311423 Chr.1: 43671760-43671760 on Build GRCh38
rs763381742 Chr.1: 43671993-43671994 on Build GRCh38
rs772631446 Chr.1: 43671776-43671776 on Build GRCh38
rs149544266 Chr.1: 43671938-43671938 on Build GRCh38
rs763138350 Chr.1: 43671922-43671922 on Build GRCh38
rs747410051 Chr.1: 43671791-43671791 on Build GRCh38
rs755820197 Chr.1: 43671853-43671853 on Build GRCh38
rs565093649 Chr.1: 43671959-43671959 on Build GRCh38
rs183975839 Chr.1: 43671626-43671626 on Build GRCh38
rs556233659 Chr.1: 43671882-43671882 on Build GRCh38
rs781462368 Chr.1: 43671901-43671901 on Build GRCh38
rs145410085 Chr.1: 43671644-43671644 on Build GRCh38
rs747292198 Chr.1: 43671895-43671895 on Build GRCh38
rs201339111 Chr.1: 43671656-43671656 on Build GRCh38
rs150730301 Chr.1: 43671603-43671603 on Build GRCh38
rs769974000 Chr.1: 43671823-43671823 on Build GRCh38
rs774869980 Chr.1: 43671652-43671652 on Build GRCh38
rs574481883 Chr.1: 43671658-43671658 on Build GRCh38
rs1039454721 Chr.1: 43671770-43671770 on Build GRCh38
rs773361159 Chr.1: 43671920-43671920 on Build GRCh38
rs369667794 Chr.1: 43671634-43671634 on Build GRCh38
rs764379296 Chr.1: 43671696-43671696 on Build GRCh38
rs557915054 Chr.1: 43671698-43671698 on Build GRCh38
rs35317523 Chr.1: 43671620-43671620 on Build GRCh38
rs148308072 Chr.1: 43671702-43671702 on Build GRCh38
rs999743738 Chr.1: 43671998-43671998 on Build GRCh38
rs762171653 Chr.1: 43671819-43671819 on Build GRCh38

View Full Product Details