Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43957314-43957821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO13
Entrez Gene ID
9670
Gene Name
importin 13
Gene Aliases
IMP13, KAP13, LGL2, RANBP13
UniGene
Hs.158497
Gene Chromosome Location
Chr.1: 43946806-43968022 on Build GRCh38
Transcripts
NM_014652.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4466827 c.1454C>T p.S485F Chr.1: 43957463-43957463 on Build GRCh38
1687595 c.1424C>T p.S475F Chr.1: 43957433-43957433 on Build GRCh38
4692820 c.1442A>G p.D481G Chr.1: 43957451-43957451 on Build GRCh38
3805356 c.1500C>T p.I500I Chr.1: 43957509-43957509 on Build GRCh38
7095912 c.1446C>G p.V482V Chr.1: 43957455-43957455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149447036 Chr.1: 43957371-43957371 on Build GRCh38
rs368982144 Chr.1: 43957443-43957443 on Build GRCh38
rs777589389 Chr.1: 43957448-43957448 on Build GRCh38
rs750750254 Chr.1: 43957412-43957412 on Build GRCh38
rs773886936 Chr.1: 43957396-43957396 on Build GRCh38
rs984676672 Chr.1: 43957602-43957602 on Build GRCh38
rs775021377 Chr.1: 43957487-43957487 on Build GRCh38
rs766557306 Chr.1: 43957419-43957419 on Build GRCh38
rs551250779 Chr.1: 43957422-43957422 on Build GRCh38
rs773102392 Chr.1: 43957360-43957360 on Build GRCh38
rs1024936682 Chr.1: 43957715-43957715 on Build GRCh38
rs557704310 Chr.1: 43957704-43957704 on Build GRCh38
rs766714188 Chr.1: 43957556-43957556 on Build GRCh38
rs763242380 Chr.1: 43957415-43957415 on Build GRCh38
rs749066021 Chr.1: 43957366-43957366 on Build GRCh38
rs755114546 Chr.1: 43957347-43957347 on Build GRCh38
rs765486831 Chr.1: 43957411-43957411 on Build GRCh38
rs762356792 Chr.1: 43957407-43957407 on Build GRCh38
rs760200148 Chr.1: 43957507-43957507 on Build GRCh38
rs373347796 Chr.1: 43957397-43957397 on Build GRCh38
rs377240680 Chr.1: 43957516-43957516 on Build GRCh38
rs777107111 Chr.1: 43957405-43957405 on Build GRCh38
rs746461655 Chr.1: 43957753-43957753 on Build GRCh38
rs375754477 Chr.1: 43957564-43957564 on Build GRCh38
rs909104106 Chr.1: 43957362-43957362 on Build GRCh38
rs926689564 Chr.1: 43957791-43957791 on Build GRCh38
rs756071679 Chr.1: 43957446-43957446 on Build GRCh38
rs143414529 Chr.1: 43957615-43957615 on Build GRCh38
rs34114279 Chr.1: 43957700-43957701 on Build GRCh38
rs777696126 Chr.1: 43957591-43957591 on Build GRCh38
rs777179135 Chr.1: 43957617-43957617 on Build GRCh38
rs200729944 Chr.1: 43957527-43957527 on Build GRCh38
rs375735197 Chr.1: 43957434-43957434 on Build GRCh38
rs747054629 Chr.1: 43957346-43957346 on Build GRCh38
rs751812759 Chr.1: 43957559-43957559 on Build GRCh38
rs756272759 Chr.1: 43957568-43957568 on Build GRCh38
rs538154313 Chr.1: 43957657-43957657 on Build GRCh38
rs767079625 Chr.1: 43957398-43957398 on Build GRCh38
rs1054070596 Chr.1: 43957417-43957417 on Build GRCh38
rs770634062 Chr.1: 43957458-43957458 on Build GRCh38
rs370459141 Chr.1: 43957563-43957563 on Build GRCh38
rs201948211 Chr.1: 43957428-43957428 on Build GRCh38
rs569169406 Chr.1: 43957654-43957654 on Build GRCh38
rs906944913 Chr.1: 43957470-43957470 on Build GRCh38
rs755098089 Chr.1: 43957425-43957425 on Build GRCh38
rs1048992851 Chr.1: 43957337-43957337 on Build GRCh38
rs923412137 Chr.1: 43957676-43957676 on Build GRCh38
rs986316000 Chr.1: 43957711-43957711 on Build GRCh38
rs146216435 Chr.1: 43957515-43957515 on Build GRCh38
rs976077327 Chr.1: 43957650-43957650 on Build GRCh38
rs186187535 Chr.1: 43957452-43957452 on Build GRCh38
rs745577104 Chr.1: 43957482-43957482 on Build GRCh38
rs771547347 Chr.1: 43957483-43957483 on Build GRCh38
rs778621101 Chr.1: 43957481-43957481 on Build GRCh38
rs923330424 Chr.1: 43957357-43957357 on Build GRCh38
rs568461145 Chr.1: 43957755-43957755 on Build GRCh38
rs531675429 Chr.1: 43957348-43957348 on Build GRCh38
rs375136589 Chr.1: 43957355-43957355 on Build GRCh38
rs201470482 Chr.1: 43957353-43957353 on Build GRCh38
rs548619205 Chr.1: 43957679-43957682 on Build GRCh38
rs1014835480 Chr.1: 43957609-43957609 on Build GRCh38
rs113864346 Chr.1: 43957386-43957386 on Build GRCh38

View Full Product Details