Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15119452-15119957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ILVBL
Entrez Gene ID
10994
Gene Name
ilvB acetolactate synthase like
Gene Aliases
209L8, AHAS, HACL1L, ILV2H
UniGene
Hs.78880
Gene Chromosome Location
Chr.19: 15114974-15125799 on Build GRCh38
Transcripts
NM_006844.4 XM_005259717.4 XM_011527651.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7174558 c.820C>A p.Q274K Chr.19: 15119512-15119512 on Build GRCh38
6003929 c.840G>C p.L280L Chr.19: 15119492-15119492 on Build GRCh38
3282877 c.838C>T p.L280L Chr.19: 15119494-15119494 on Build GRCh38
992044 c.823C>T p.R275C Chr.19: 15119509-15119509 on Build GRCh38
3422483 c.877C>A p.Q293K Chr.19: 15119455-15119455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375796907 Chr.19: 15119488-15119488 on Build GRCh38
rs970676008 Chr.19: 15119912-15119912 on Build GRCh38
rs1038700021 Chr.19: 15119756-15119756 on Build GRCh38
rs749283160 Chr.19: 15119487-15119488 on Build GRCh38
rs1033757820 Chr.19: 15119605-15119605 on Build GRCh38
rs375508917 Chr.19: 15119653-15119653 on Build GRCh38
rs369349850 Chr.19: 15119859-15119859 on Build GRCh38
rs745443448 Chr.19: 15119587-15119587 on Build GRCh38
rs540099152 Chr.19: 15119841-15119841 on Build GRCh38
rs901537872 Chr.19: 15119781-15119781 on Build GRCh38
rs908263847 Chr.19: 15119669-15119669 on Build GRCh38
rs952441851 Chr.19: 15119609-15119609 on Build GRCh38
rs976537122 Chr.19: 15119902-15119902 on Build GRCh38
rs149721143 Chr.19: 15119487-15119487 on Build GRCh38
rs943149227 Chr.19: 15119704-15119704 on Build GRCh38
rs765419787 Chr.19: 15119510-15119510 on Build GRCh38
rs569835539 Chr.19: 15119560-15119561 on Build GRCh38
rs145280202 Chr.19: 15119836-15119836 on Build GRCh38
rs758432782 Chr.19: 15119515-15119515 on Build GRCh38
rs756476211 Chr.19: 15119534-15119534 on Build GRCh38
rs774547221 Chr.19: 15119550-15119553 on Build GRCh38
rs573474304 Chr.19: 15119869-15119869 on Build GRCh38
rs959481449 Chr.19: 15119858-15119858 on Build GRCh38
rs144521840 Chr.19: 15119508-15119508 on Build GRCh38
rs1032454641 Chr.19: 15119525-15119525 on Build GRCh38
rs376142251 Chr.19: 15119545-15119545 on Build GRCh38
rs961075357 Chr.19: 15119896-15119896 on Build GRCh38
rs754346294 Chr.19: 15119604-15119604 on Build GRCh38
rs970916804 Chr.19: 15119600-15119600 on Build GRCh38
rs936708142 Chr.19: 15119737-15119737 on Build GRCh38
rs746222205 Chr.19: 15119526-15119526 on Build GRCh38
rs781609278 Chr.19: 15119523-15119523 on Build GRCh38
rs2074264 Chr.19: 15119843-15119843 on Build GRCh38
rs778668560 Chr.19: 15119539-15119539 on Build GRCh38
rs771738981 Chr.19: 15119486-15119486 on Build GRCh38
rs372535768 Chr.19: 15119794-15119794 on Build GRCh38
rs747631733 Chr.19: 15119485-15119485 on Build GRCh38
rs1016310618 Chr.19: 15119603-15119603 on Build GRCh38
rs374239382 Chr.19: 15119482-15119482 on Build GRCh38
rs544384447 Chr.19: 15119675-15119675 on Build GRCh38
rs11673486 Chr.19: 15119695-15119695 on Build GRCh38
rs751951816 Chr.19: 15119522-15119522 on Build GRCh38
rs375406074 Chr.19: 15119887-15119887 on Build GRCh38
rs1054214532 Chr.19: 15119738-15119738 on Build GRCh38
rs768689620 Chr.19: 15119802-15119802 on Build GRCh38
rs922421750 Chr.19: 15119933-15119933 on Build GRCh38
rs574925490 Chr.19: 15119905-15119934 on Build GRCh38
rs1015682512 Chr.19: 15119870-15119870 on Build GRCh38
rs768975700 Chr.19: 15119543-15119546 on Build GRCh38
rs747829531 Chr.19: 15119541-15119541 on Build GRCh38
rs985381220 Chr.19: 15119598-15119598 on Build GRCh38
rs367772538 Chr.19: 15119495-15119495 on Build GRCh38
rs147819940 Chr.19: 15119696-15119696 on Build GRCh38
rs752952432 Chr.19: 15119511-15119511 on Build GRCh38
rs547381507 Chr.19: 15119662-15119662 on Build GRCh38
rs949953584 Chr.19: 15119681-15119681 on Build GRCh38
rs773015464 Chr.19: 15119920-15119920 on Build GRCh38
rs764180162 Chr.19: 15119518-15119518 on Build GRCh38
rs746570411 Chr.19: 15119559-15119559 on Build GRCh38
rs539795993 Chr.19: 15119720-15119720 on Build GRCh38
rs370725036 Chr.19: 15119509-15119509 on Build GRCh38
rs141346014 Chr.19: 15119793-15119793 on Build GRCh38
rs151010404 Chr.19: 15119840-15119840 on Build GRCh38
rs575380971 Chr.19: 15119572-15119573 on Build GRCh38
rs981355703 Chr.19: 15119694-15119694 on Build GRCh38
rs892877555 Chr.19: 15119698-15119698 on Build GRCh38
rs1024920596 Chr.19: 15119873-15119873 on Build GRCh38
rs769499091 Chr.19: 15119635-15119635 on Build GRCh38
rs961431071 Chr.19: 15119674-15119674 on Build GRCh38
rs772857203 Chr.19: 15119552-15119552 on Build GRCh38
rs1016879592 Chr.19: 15119849-15119849 on Build GRCh38
rs368571962 Chr.19: 15119697-15119697 on Build GRCh38
rs770885943 Chr.19: 15119932-15119932 on Build GRCh38

View Full Product Details