Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18436550-18437044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISYNA1
Entrez Gene ID
51477
Gene Name
inositol-3-phosphate synthase 1
Gene Aliases
INO1, INOS, IPS, IPS 1, IPS-1
UniGene
Hs.405873
Gene Chromosome Location
Chr.19: 18434388-18438301 on Build GRCh38
Transcripts
NM_001170938.1 NM_001253389.1 NM_016368.4 NR_045573.1 NR_045574.1 XM_011528059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6687666 c.416-3delC p.? Chr.19: 18436880-18436880 on Build GRCh38
4693196 c.450G>A p.A150A Chr.19: 18436843-18436843 on Build GRCh38
4693197 c.372C>T p.S124S Chr.19: 18437016-18437016 on Build GRCh38
6043252 c.492A>C p.Q164H Chr.19: 18436801-18436801 on Build GRCh38
4693198 c.345G>T p.E115D Chr.19: 18437043-18437043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778890208 Chr.19: 18436717-18436717 on Build GRCh38
rs765365223 Chr.19: 18436900-18436900 on Build GRCh38
rs776303300 Chr.19: 18436625-18436625 on Build GRCh38
rs376309732 Chr.19: 18436966-18436966 on Build GRCh38
rs756524230 Chr.19: 18436773-18436773 on Build GRCh38
rs749075844 Chr.19: 18436925-18436925 on Build GRCh38
rs993231647 Chr.19: 18436812-18436812 on Build GRCh38
rs1031191050 Chr.19: 18436962-18436962 on Build GRCh38
rs780972489 Chr.19: 18436831-18436831 on Build GRCh38
rs374113839 Chr.19: 18436691-18436691 on Build GRCh38
rs951916746 Chr.19: 18436855-18436855 on Build GRCh38
rs752824769 Chr.19: 18436961-18436961 on Build GRCh38
rs752408483 Chr.19: 18436653-18436653 on Build GRCh38
rs773743984 Chr.19: 18436741-18436741 on Build GRCh38
rs751639570 Chr.19: 18436988-18436988 on Build GRCh38
rs770779084 Chr.19: 18436719-18436719 on Build GRCh38
rs1055154489 Chr.19: 18436670-18436670 on Build GRCh38
rs775289903 Chr.19: 18436843-18436843 on Build GRCh38
rs540468469 Chr.19: 18436665-18436665 on Build GRCh38
rs766889926 Chr.19: 18436776-18436776 on Build GRCh38
rs779754659 Chr.19: 18436924-18436924 on Build GRCh38
rs745650840 Chr.19: 18436723-18436723 on Build GRCh38
rs144717477 Chr.19: 18436676-18436676 on Build GRCh38
rs1041887983 Chr.19: 18436581-18436581 on Build GRCh38
rs924800575 Chr.19: 18436588-18436588 on Build GRCh38
rs977535109 Chr.19: 18436983-18436983 on Build GRCh38
rs770712032 Chr.19: 18436623-18436623 on Build GRCh38
rs149381559 Chr.19: 18436576-18436576 on Build GRCh38
rs764100802 Chr.19: 18436880-18436880 on Build GRCh38
rs752248602 Chr.19: 18436716-18436717 on Build GRCh38
rs761212791 Chr.19: 18436870-18436870 on Build GRCh38
rs369738821 Chr.19: 18436982-18436982 on Build GRCh38
rs773261369 Chr.19: 18436607-18436607 on Build GRCh38
rs779951043 Chr.19: 18436995-18436995 on Build GRCh38
rs148517966 Chr.19: 18436804-18436804 on Build GRCh38
rs770150882 Chr.19: 18436574-18436574 on Build GRCh38
rs751563209 Chr.19: 18436893-18436894 on Build GRCh38
rs753819243 Chr.19: 18437004-18437004 on Build GRCh38
rs368377568 Chr.19: 18436657-18436657 on Build GRCh38
rs771441661 Chr.19: 18436942-18436942 on Build GRCh38
rs768913421 Chr.19: 18436606-18436606 on Build GRCh38
rs1029172674 Chr.19: 18436673-18436673 on Build GRCh38
rs924769521 Chr.19: 18436986-18436986 on Build GRCh38
rs369808766 Chr.19: 18436897-18436897 on Build GRCh38
rs748877705 Chr.19: 18436848-18436848 on Build GRCh38
rs761197113 Chr.19: 18436947-18436947 on Build GRCh38
rs777336870 Chr.19: 18436950-18436950 on Build GRCh38
rs570653689 Chr.19: 18436914-18436914 on Build GRCh38
rs762629715 Chr.19: 18436735-18436735 on Build GRCh38
rs754696099 Chr.19: 18437007-18437007 on Build GRCh38
rs749293912 Chr.19: 18436762-18436762 on Build GRCh38
rs1018931216 Chr.19: 18436615-18436615 on Build GRCh38
rs759242237 Chr.19: 18436690-18436716 on Build GRCh38
rs764866864 Chr.19: 18436645-18436645 on Build GRCh38
rs376639419 Chr.19: 18436849-18436849 on Build GRCh38
rs529148937 Chr.19: 18436718-18436718 on Build GRCh38
rs754073061 Chr.19: 18436781-18436781 on Build GRCh38
rs747873233 Chr.19: 18436933-18436933 on Build GRCh38
rs759798488 Chr.19: 18436954-18436954 on Build GRCh38
rs898375630 Chr.19: 18436792-18436792 on Build GRCh38
rs775096976 Chr.19: 18436726-18436726 on Build GRCh38
rs147482450 Chr.19: 18436690-18436690 on Build GRCh38
rs767101089 Chr.19: 18436879-18436879 on Build GRCh38
rs747706315 Chr.19: 18437014-18437014 on Build GRCh38
rs773823049 Chr.19: 18436883-18436883 on Build GRCh38
rs772871610 Chr.19: 18436944-18436944 on Build GRCh38
rs996102315 Chr.19: 18436898-18436898 on Build GRCh38
rs201189874 Chr.19: 18436923-18436923 on Build GRCh38
rs756055475 Chr.19: 18436920-18436920 on Build GRCh38
rs755077720 Chr.19: 18436713-18436713 on Build GRCh38
rs533150372 Chr.19: 18436582-18436582 on Build GRCh38
rs768550882 Chr.19: 18436928-18436928 on Build GRCh38
rs760569282 Chr.19: 18436619-18436619 on Build GRCh38
rs199555469 Chr.19: 18436815-18436815 on Build GRCh38
rs974986104 Chr.19: 18436663-18436663 on Build GRCh38
rs778798808 Chr.19: 18437013-18437013 on Build GRCh38
rs200584853 Chr.19: 18436796-18436796 on Build GRCh38
rs757391449 Chr.19: 18436993-18436993 on Build GRCh38
rs761294933 Chr.19: 18436885-18436885 on Build GRCh38
rs774117364 Chr.19: 18436852-18436852 on Build GRCh38
rs767903366 Chr.19: 18436759-18436759 on Build GRCh38
rs745849041 Chr.19: 18436837-18436837 on Build GRCh38
rs754259197 Chr.19: 18436891-18436891 on Build GRCh38
rs762410485 Chr.19: 18436730-18436733 on Build GRCh38
rs759163947 Chr.19: 18436629-18436629 on Build GRCh38
rs917606458 Chr.19: 18436750-18436750 on Build GRCh38
rs762430471 Chr.19: 18436654-18436654 on Build GRCh38
rs764857591 Chr.19: 18436717-18436718 on Build GRCh38
rs763638072 Chr.19: 18436655-18436655 on Build GRCh38
rs755265547 Chr.19: 18436785-18436785 on Build GRCh38
rs1006136397 Chr.19: 18436864-18436864 on Build GRCh38
rs777942533 Chr.19: 18436908-18436908 on Build GRCh38
rs762311401 Chr.19: 18436754-18436754 on Build GRCh38
rs201814656 Chr.19: 18436902-18436902 on Build GRCh38
rs776699315 Chr.19: 18436720-18436720 on Build GRCh38
rs776083627 Chr.19: 18436618-18436619 on Build GRCh38
rs769628350 Chr.19: 18436724-18436724 on Build GRCh38
rs886929409 Chr.19: 18436860-18436860 on Build GRCh38
rs372324582 Chr.19: 18436901-18436901 on Build GRCh38
rs367647209 Chr.19: 18436697-18436697 on Build GRCh38
rs765676731 Chr.19: 18436957-18436957 on Build GRCh38

View Full Product Details