Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18436853-18437343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISYNA1
Entrez Gene ID
51477
Gene Name
inositol-3-phosphate synthase 1
Gene Aliases
INO1, INOS, IPS, IPS 1, IPS-1
UniGene
Hs.405873
Gene Chromosome Location
Chr.19: 18434388-18438301 on Build GRCh38
Transcripts
NM_001170938.1 NM_001253389.1 NM_016368.4 NR_045573.1 NR_045574.1 XM_011528059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6687666 c.416-3delC p.? Chr.19: 18436880-18436880 on Build GRCh38
6017378 c.303G>C p.S101S Chr.19: 18437085-18437085 on Build GRCh38
4693197 c.372C>T p.S124S Chr.19: 18437016-18437016 on Build GRCh38
4693198 c.345G>T p.E115D Chr.19: 18437043-18437043 on Build GRCh38
5495559 c.359T>C p.F120S Chr.19: 18437029-18437029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753989088 Chr.19: 18437273-18437273 on Build GRCh38
rs765365223 Chr.19: 18436900-18436900 on Build GRCh38
rs770008089 Chr.19: 18437073-18437073 on Build GRCh38
rs903831659 Chr.19: 18437274-18437274 on Build GRCh38
rs931274632 Chr.19: 18437173-18437173 on Build GRCh38
rs1033575598 Chr.19: 18437235-18437235 on Build GRCh38
rs376309732 Chr.19: 18436966-18436966 on Build GRCh38
rs749075844 Chr.19: 18436925-18436925 on Build GRCh38
rs759798488 Chr.19: 18436954-18436954 on Build GRCh38
rs1031191050 Chr.19: 18436962-18436962 on Build GRCh38
rs752824769 Chr.19: 18436961-18436961 on Build GRCh38
rs761855774 Chr.19: 18437112-18437112 on Build GRCh38
rs929050874 Chr.19: 18437193-18437193 on Build GRCh38
rs767101089 Chr.19: 18436879-18436879 on Build GRCh38
rs767621422 Chr.19: 18437114-18437114 on Build GRCh38
rs753676609 Chr.19: 18437124-18437124 on Build GRCh38
rs772871610 Chr.19: 18436944-18436944 on Build GRCh38
rs773823049 Chr.19: 18436883-18436883 on Build GRCh38
rs747706315 Chr.19: 18437014-18437014 on Build GRCh38
rs996102315 Chr.19: 18436898-18436898 on Build GRCh38
rs751639570 Chr.19: 18436988-18436988 on Build GRCh38
rs971425669 Chr.19: 18437300-18437300 on Build GRCh38
rs887573944 Chr.19: 18437206-18437206 on Build GRCh38
rs977463313 Chr.19: 18437303-18437303 on Build GRCh38
rs917637479 Chr.19: 18437048-18437048 on Build GRCh38
rs201189874 Chr.19: 18436923-18436923 on Build GRCh38
rs756055475 Chr.19: 18436920-18436920 on Build GRCh38
rs1006476987 Chr.19: 18437222-18437222 on Build GRCh38
rs779754659 Chr.19: 18436924-18436924 on Build GRCh38
rs763014105 Chr.19: 18437083-18437083 on Build GRCh38
rs1031767599 Chr.19: 18437306-18437306 on Build GRCh38
rs768550882 Chr.19: 18436928-18436928 on Build GRCh38
rs919585290 Chr.19: 18437146-18437146 on Build GRCh38
rs977535109 Chr.19: 18436983-18436983 on Build GRCh38
rs535730541 Chr.19: 18437179-18437179 on Build GRCh38
rs778798808 Chr.19: 18437013-18437013 on Build GRCh38
rs764100802 Chr.19: 18436880-18436880 on Build GRCh38
rs369738821 Chr.19: 18436982-18436982 on Build GRCh38
rs1040445422 Chr.19: 18437240-18437240 on Build GRCh38
rs779951043 Chr.19: 18436995-18436995 on Build GRCh38
rs373317657 Chr.19: 18437034-18437034 on Build GRCh38
rs757391449 Chr.19: 18436993-18436993 on Build GRCh38
rs943026151 Chr.19: 18437109-18437109 on Build GRCh38
rs535629613 Chr.19: 18437241-18437241 on Build GRCh38
rs761294933 Chr.19: 18436885-18436885 on Build GRCh38
rs895034789 Chr.19: 18437244-18437244 on Build GRCh38
rs753819243 Chr.19: 18437004-18437004 on Build GRCh38
rs751563209 Chr.19: 18436893-18436894 on Build GRCh38
rs771441661 Chr.19: 18436942-18436942 on Build GRCh38
rs567066618 Chr.19: 18437135-18437135 on Build GRCh38
rs996216843 Chr.19: 18437252-18437252 on Build GRCh38
rs754259197 Chr.19: 18436891-18436891 on Build GRCh38
rs924769521 Chr.19: 18436986-18436986 on Build GRCh38
rs369808766 Chr.19: 18436897-18436897 on Build GRCh38
rs764882294 Chr.19: 18437134-18437134 on Build GRCh38
rs575307400 Chr.19: 18437199-18437199 on Build GRCh38
rs761197113 Chr.19: 18436947-18436947 on Build GRCh38
rs556027884 Chr.19: 18437181-18437181 on Build GRCh38
rs910807563 Chr.19: 18437066-18437066 on Build GRCh38
rs796373956 Chr.19: 18437101-18437101 on Build GRCh38
rs764292495 Chr.19: 18437085-18437085 on Build GRCh38
rs777336870 Chr.19: 18436950-18436950 on Build GRCh38
rs777248875 Chr.19: 18437063-18437063 on Build GRCh38
rs754963960 Chr.19: 18437126-18437126 on Build GRCh38
rs1001141260 Chr.19: 18437319-18437319 on Build GRCh38
rs570653689 Chr.19: 18436914-18436914 on Build GRCh38
rs777942533 Chr.19: 18436908-18436908 on Build GRCh38
rs746505603 Chr.19: 18437058-18437058 on Build GRCh38
rs775940285 Chr.19: 18437074-18437074 on Build GRCh38
rs376059509 Chr.19: 18437110-18437110 on Build GRCh38
rs754696099 Chr.19: 18437007-18437007 on Build GRCh38
rs11552754 Chr.19: 18437045-18437045 on Build GRCh38
rs1013531326 Chr.19: 18437271-18437271 on Build GRCh38
rs901547128 Chr.19: 18437191-18437191 on Build GRCh38
rs201814656 Chr.19: 18436902-18436902 on Build GRCh38
rs777547297 Chr.19: 18437148-18437148 on Build GRCh38
rs760122823 Chr.19: 18437067-18437067 on Build GRCh38
rs372324582 Chr.19: 18436901-18436901 on Build GRCh38
rs747873233 Chr.19: 18436933-18436933 on Build GRCh38
rs1047510840 Chr.19: 18437205-18437205 on Build GRCh38
rs765676731 Chr.19: 18436957-18436957 on Build GRCh38

View Full Product Details