Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19255963-19256459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAPLN4
Entrez Gene ID
404037
Gene Name
hyaluronan and proteoglycan link protein 4
Gene Aliases
BRAL2
UniGene
Hs.367829
Gene Chromosome Location
Chr.19: 19255641-19262787 on Build GRCh38
Transcripts
NM_023002.2

SNP Detail

SNP ID Chromosome Location
rs183075598 Chr.19: 19256378-19256378 on Build GRCh38
rs111446817 Chr.19: 19256075-19256075 on Build GRCh38
rs538128997 Chr.19: 19256412-19256412 on Build GRCh38
rs534816033 Chr.19: 19256111-19256111 on Build GRCh38
rs1021266752 Chr.19: 19256035-19256035 on Build GRCh38
rs958695114 Chr.19: 19256125-19256125 on Build GRCh38
rs559621337 Chr.19: 19256155-19256155 on Build GRCh38
rs765214761 Chr.19: 19256371-19256371 on Build GRCh38
rs555619789 Chr.19: 19256012-19256012 on Build GRCh38
rs780597414 Chr.19: 19256090-19256090 on Build GRCh38
rs558131441 Chr.19: 19256418-19256418 on Build GRCh38
rs557945016 Chr.19: 19256094-19256094 on Build GRCh38
rs1054194157 Chr.19: 19256275-19256275 on Build GRCh38
rs915206322 Chr.19: 19256276-19256276 on Build GRCh38
rs943866612 Chr.19: 19256297-19256297 on Build GRCh38
rs866437726 Chr.19: 19256197-19256197 on Build GRCh38
rs997993641 Chr.19: 19256074-19256074 on Build GRCh38
rs560731560 Chr.19: 19256358-19256358 on Build GRCh38
rs1013017730 Chr.19: 19256383-19256383 on Build GRCh38
rs970863791 Chr.19: 19256202-19256202 on Build GRCh38
rs1025488922 Chr.19: 19256089-19256089 on Build GRCh38
rs207477024 Chr.19: 19255989-19255989 on Build GRCh38
rs924177945 Chr.19: 19256254-19256254 on Build GRCh38
rs766576224 Chr.19: 19256352-19256352 on Build GRCh38
rs535551508 Chr.19: 19256385-19256385 on Build GRCh38
rs3752170 Chr.19: 19256369-19256369 on Build GRCh38
rs991222849 Chr.19: 19256164-19256164 on Build GRCh38
rs1053814930 Chr.19: 19255984-19255984 on Build GRCh38
rs924145487 Chr.19: 19256413-19256413 on Build GRCh38
rs935631616 Chr.19: 19256258-19256258 on Build GRCh38
rs145641503 Chr.19: 19256192-19256192 on Build GRCh38
rs900974751 Chr.19: 19256068-19256068 on Build GRCh38
rs773302295 Chr.19: 19256226-19256226 on Build GRCh38
rs56278919 Chr.19: 19256381-19256381 on Build GRCh38
rs550869714 Chr.19: 19256289-19256289 on Build GRCh38
rs899537268 Chr.19: 19256013-19256013 on Build GRCh38
rs546317050 Chr.19: 19256136-19256136 on Build GRCh38
rs538141439 Chr.19: 19256083-19256083 on Build GRCh38
rs774731005 Chr.19: 19256186-19256186 on Build GRCh38
rs901282417 Chr.19: 19256317-19256317 on Build GRCh38
rs577892615 Chr.19: 19256424-19256424 on Build GRCh38
rs568844765 Chr.19: 19256396-19256396 on Build GRCh38
rs531063295 Chr.19: 19256238-19256238 on Build GRCh38
rs540850998 Chr.19: 19256204-19256204 on Build GRCh38
rs1031238978 Chr.19: 19256398-19256398 on Build GRCh38
rs147752236 Chr.19: 19256222-19256222 on Build GRCh38
rs187408829 Chr.19: 19256389-19256389 on Build GRCh38
rs1039761124 Chr.19: 19256298-19256298 on Build GRCh38
rs541825992 Chr.19: 19256203-19256203 on Build GRCh38
rs575413457 Chr.19: 19256024-19256024 on Build GRCh38
rs919905899 Chr.19: 19256218-19256218 on Build GRCh38
rs756721106 Chr.19: 19256015-19256015 on Build GRCh38
rs1024044019 Chr.19: 19256388-19256388 on Build GRCh38
rs772376924 Chr.19: 19256199-19256199 on Build GRCh38
rs56044734 Chr.19: 19256118-19256118 on Build GRCh38
rs968290934 Chr.19: 19256071-19256071 on Build GRCh38
rs917989538 Chr.19: 19256166-19256166 on Build GRCh38
rs1040835111 Chr.19: 19256343-19256343 on Build GRCh38
rs867248667 Chr.19: 19256091-19256091 on Build GRCh38

View Full Product Details