Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40383398-40383936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIPK4
Entrez Gene ID
147746
Gene Name
homeodomain interacting protein kinase 4
Gene Aliases
-
UniGene
Hs.79363
Gene Chromosome Location
Chr.19: 40379271-40390221 on Build GRCh38
Transcripts
NM_144685.4 XM_006723036.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4556535 c.694G>A p.G232S Chr.19: 40383911-40383911 on Build GRCh38
1393801 c.786G>A p.Q262Q Chr.19: 40383819-40383819 on Build GRCh38
3225476 c.744C>T p.F248F Chr.19: 40383861-40383861 on Build GRCh38
6003621 c.673C>T p.R225C Chr.19: 40383932-40383932 on Build GRCh38
996662 c.814G>A p.E272K Chr.19: 40383791-40383791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757519442 Chr.19: 40383873-40383873 on Build GRCh38
rs776301359 Chr.19: 40383742-40383742 on Build GRCh38
rs754877298 Chr.19: 40383846-40383846 on Build GRCh38
rs762600610 Chr.19: 40383555-40383555 on Build GRCh38
rs772748974 Chr.19: 40383807-40383807 on Build GRCh38
rs530385981 Chr.19: 40383463-40383463 on Build GRCh38
rs148229742 Chr.19: 40383543-40383543 on Build GRCh38
rs888213578 Chr.19: 40383881-40383881 on Build GRCh38
rs781211143 Chr.19: 40383777-40383777 on Build GRCh38
rs888512312 Chr.19: 40383717-40383717 on Build GRCh38
rs200561773 Chr.19: 40383836-40383836 on Build GRCh38
rs1033675607 Chr.19: 40383700-40383700 on Build GRCh38
rs751396676 Chr.19: 40383767-40383767 on Build GRCh38
rs148038861 Chr.19: 40383793-40383793 on Build GRCh38
rs751404410 Chr.19: 40383840-40383840 on Build GRCh38
rs978819766 Chr.19: 40383584-40383585 on Build GRCh38
rs1057319350 Chr.19: 40383622-40383622 on Build GRCh38
rs1048181868 Chr.19: 40383613-40383613 on Build GRCh38
rs1040695786 Chr.19: 40383581-40383581 on Build GRCh38
rs989483325 Chr.19: 40383710-40383710 on Build GRCh38
rs761528252 Chr.19: 40383748-40383748 on Build GRCh38
rs570327200 Chr.19: 40383737-40383737 on Build GRCh38
rs80125643 Chr.19: 40383605-40383605 on Build GRCh38
rs888243108 Chr.19: 40383619-40383619 on Build GRCh38
rs1017465649 Chr.19: 40383911-40383911 on Build GRCh38
rs199630536 Chr.19: 40383849-40383849 on Build GRCh38
rs756252191 Chr.19: 40383854-40383854 on Build GRCh38
rs541357005 Chr.19: 40383541-40383541 on Build GRCh38
rs766201514 Chr.19: 40383762-40383762 on Build GRCh38
rs528831941 Chr.19: 40383777-40383778 on Build GRCh38
rs752760077 Chr.19: 40383853-40383853 on Build GRCh38
rs944975576 Chr.19: 40383815-40383815 on Build GRCh38
rs372266650 Chr.19: 40383770-40383770 on Build GRCh38
rs920899826 Chr.19: 40383847-40383847 on Build GRCh38
rs961944374 Chr.19: 40383423-40383423 on Build GRCh38
rs758198344 Chr.19: 40383754-40383754 on Build GRCh38
rs945549676 Chr.19: 40383445-40383445 on Build GRCh38
rs571026735 Chr.19: 40383808-40383808 on Build GRCh38
rs746309036 Chr.19: 40383800-40383800 on Build GRCh38
rs547458310 Chr.19: 40383664-40383664 on Build GRCh38
rs574611791 Chr.19: 40383879-40383879 on Build GRCh38
rs948903042 Chr.19: 40383668-40383668 on Build GRCh38
rs373462941 Chr.19: 40383791-40383791 on Build GRCh38
rs186210468 Chr.19: 40383671-40383671 on Build GRCh38
rs760089870 Chr.19: 40383736-40383736 on Build GRCh38
rs968519406 Chr.19: 40383572-40383572 on Build GRCh38
rs75602333 Chr.19: 40383850-40383850 on Build GRCh38
rs747527574 Chr.19: 40383893-40383893 on Build GRCh38
rs959371447 Chr.19: 40383706-40383706 on Build GRCh38
rs563575814 Chr.19: 40383446-40383446 on Build GRCh38
rs749410438 Chr.19: 40383867-40383867 on Build GRCh38
rs1003906686 Chr.19: 40383687-40383687 on Build GRCh38
rs970660441 Chr.19: 40383677-40383677 on Build GRCh38
rs200281362 Chr.19: 40383787-40383787 on Build GRCh38
rs868155846 Chr.19: 40383776-40383777 on Build GRCh38
rs771130701 Chr.19: 40383792-40383792 on Build GRCh38
rs530758379 Chr.19: 40383489-40383489 on Build GRCh38
rs770659093 Chr.19: 40383902-40383902 on Build GRCh38
rs377700394 Chr.19: 40383837-40383837 on Build GRCh38
rs912159673 Chr.19: 40383538-40383538 on Build GRCh38
rs772802613 Chr.19: 40383896-40383896 on Build GRCh38
rs1037433624 Chr.19: 40383768-40383768 on Build GRCh38
rs372625567 Chr.19: 40383882-40383882 on Build GRCh38
rs746295654 Chr.19: 40383880-40383880 on Build GRCh38
rs772674739 Chr.19: 40383809-40383809 on Build GRCh38
rs61730201 Chr.19: 40383711-40383711 on Build GRCh38
rs765034874 Chr.19: 40383749-40383749 on Build GRCh38
rs763824787 Chr.19: 40383596-40383596 on Build GRCh38
rs762474104 Chr.19: 40383899-40383899 on Build GRCh38
rs750222296 Chr.19: 40383753-40383753 on Build GRCh38
rs748264743 Chr.19: 40383783-40383783 on Build GRCh38
rs780777730 Chr.19: 40383884-40383884 on Build GRCh38
rs530090631 Chr.19: 40383688-40383688 on Build GRCh38
rs181487568 Chr.19: 40383562-40383562 on Build GRCh38
rs993450349 Chr.19: 40383479-40383479 on Build GRCh38
rs756305129 Chr.19: 40383786-40383786 on Build GRCh38

View Full Product Details