Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46124545-46125049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFL2
Entrez Gene ID
147920
Gene Name
IGF like family member 2
Gene Aliases
UNQ645, VPRI645
UniGene
Hs.99376
Gene Chromosome Location
Chr.19: 46078513-46195097 on Build GRCh38
Transcripts
NM_207393.1 XM_011526953.2 XM_017026796.1 XM_017026797.1

Gene Symbol
IGFL3
Entrez Gene ID
388555
Gene Name
IGF like family member 3
Gene Aliases
UNQ483
UniGene
Hs.365496
Gene Chromosome Location
Chr.19: 46119778-46127101 on Build GRCh38
Transcripts
NM_207393.1 XM_011526953.2 XM_017026796.1 XM_017026797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5966041 c.3G>A p.M1I Chr.19: 46124647-46124647 on Build GRCh38
1394731 c.15C>T p.C5C Chr.19: 46124635-46124635 on Build GRCh38
7077625 c.24G>A p.L8L Chr.19: 46124626-46124626 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749234146 Chr.19: 46124608-46124608 on Build GRCh38
rs747645118 Chr.19: 46124579-46124579 on Build GRCh38
rs1047549487 Chr.19: 46124985-46124985 on Build GRCh38
rs535275749 Chr.19: 46125024-46125024 on Build GRCh38
rs758214864 Chr.19: 46124631-46124631 on Build GRCh38
rs953742689 Chr.19: 46124867-46124867 on Build GRCh38
rs985073393 Chr.19: 46124880-46124880 on Build GRCh38
rs995307459 Chr.19: 46124862-46124862 on Build GRCh38
rs914065038 Chr.19: 46124815-46124815 on Build GRCh38
rs763329386 Chr.19: 46124594-46124594 on Build GRCh38
rs570141180 Chr.19: 46124734-46124734 on Build GRCh38
rs797016212 Chr.19: 46124674-46124674 on Build GRCh38
rs538086125 Chr.19: 46124863-46124863 on Build GRCh38
rs980075897 Chr.19: 46124840-46124840 on Build GRCh38
rs1033660203 Chr.19: 46124764-46124764 on Build GRCh38
rs771335305 Chr.19: 46124677-46124677 on Build GRCh38
rs747697563 Chr.19: 46124651-46124651 on Build GRCh38
rs749805786 Chr.19: 46124662-46124662 on Build GRCh38
rs190226328 Chr.19: 46124648-46124648 on Build GRCh38
rs1038790222 Chr.19: 46124663-46124663 on Build GRCh38
rs1019844862 Chr.19: 46124923-46124923 on Build GRCh38
rs368574851 Chr.19: 46124596-46124596 on Build GRCh38
rs755652766 Chr.19: 46124657-46124657 on Build GRCh38
rs376595190 Chr.19: 46124589-46124589 on Build GRCh38
rs997266982 Chr.19: 46124695-46124695 on Build GRCh38
rs554973329 Chr.19: 46125025-46125025 on Build GRCh38
rs963019921 Chr.19: 46124751-46124751 on Build GRCh38
rs570651866 Chr.19: 46124802-46124802 on Build GRCh38
rs759867309 Chr.19: 46124611-46124611 on Build GRCh38
rs151178962 Chr.19: 46124639-46124639 on Build GRCh38
rs965631349 Chr.19: 46124948-46124948 on Build GRCh38
rs1006082333 Chr.19: 46125014-46125014 on Build GRCh38
rs532686558 Chr.19: 46124736-46124736 on Build GRCh38
rs770336961 Chr.19: 46124580-46124580 on Build GRCh38
rs978675607 Chr.19: 46124959-46124959 on Build GRCh38
rs764516278 Chr.19: 46124792-46124792 on Build GRCh38
rs922938035 Chr.19: 46124960-46124960 on Build GRCh38
rs751310981 Chr.19: 46124640-46124640 on Build GRCh38
rs777206835 Chr.19: 46124660-46124660 on Build GRCh38
rs966945876 Chr.19: 46124821-46124821 on Build GRCh38
rs552569377 Chr.19: 46124769-46124769 on Build GRCh38
rs750393688 Chr.19: 46124624-46124624 on Build GRCh38
rs1002180468 Chr.19: 46124763-46124763 on Build GRCh38
rs1038678194 Chr.19: 46124882-46124882 on Build GRCh38
rs1047559483 Chr.19: 46124702-46124702 on Build GRCh38
rs763285429 Chr.19: 46124779-46124779 on Build GRCh38
rs561876864 Chr.19: 46124622-46124622 on Build GRCh38
rs774511759 Chr.19: 46124606-46124606 on Build GRCh38
rs775757576 Chr.19: 46124619-46124619 on Build GRCh38
rs895186369 Chr.19: 46124641-46124641 on Build GRCh38
rs181960116 Chr.19: 46124991-46124991 on Build GRCh38
rs933047902 Chr.19: 46124961-46124961 on Build GRCh38
rs936737520 Chr.19: 46124612-46124612 on Build GRCh38
rs1015397466 Chr.19: 46124776-46124776 on Build GRCh38
rs371832725 Chr.19: 46124705-46124705 on Build GRCh38
rs772461227 Chr.19: 46124698-46124698 on Build GRCh38
rs775958212 Chr.19: 46124757-46124757 on Build GRCh38
rs780802054 Chr.19: 46124650-46124650 on Build GRCh38
rs1016204443 Chr.19: 46124740-46124740 on Build GRCh38
rs1006549253 Chr.19: 46124731-46124731 on Build GRCh38
rs767837058 Chr.19: 46124615-46124615 on Build GRCh38
rs760803915 Chr.19: 46124620-46124620 on Build GRCh38
rs774835836 Chr.19: 46124686-46124686 on Build GRCh38
rs901568821 Chr.19: 46124678-46124678 on Build GRCh38
rs386809809 Chr.19: 46124763-46124764 on Build GRCh38
rs943633240 Chr.19: 46124878-46124878 on Build GRCh38

View Full Product Details