Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49154728-49155231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HRC
Entrez Gene ID
3270
Gene Name
histidine rich calcium binding protein
Gene Aliases
-
UniGene
Hs.436885
Gene Chromosome Location
Chr.19: 49151198-49155443 on Build GRCh38
Transcripts
NM_002152.2 XM_017026733.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
273647 c.397G>A p.G133R Chr.19: 49154841-49154841 on Build GRCh38
6525168 c.180C>T p.H60H Chr.19: 49155058-49155058 on Build GRCh38
1241281 c.71delC p.P24fs*3 Chr.19: 49155167-49155167 on Build GRCh38
4080007 c.273G>T p.E91D Chr.19: 49154965-49154965 on Build GRCh38
5716776 c.292G>A p.E98K Chr.19: 49154946-49154946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770726719 Chr.19: 49154917-49154918 on Build GRCh38
rs750915466 Chr.19: 49155162-49155162 on Build GRCh38
rs146528509 Chr.19: 49154878-49154878 on Build GRCh38
rs1005416189 Chr.19: 49154929-49154929 on Build GRCh38
rs769867452 Chr.19: 49154898-49154898 on Build GRCh38
rs765338813 Chr.19: 49155211-49155211 on Build GRCh38
rs763091274 Chr.19: 49155001-49155001 on Build GRCh38
rs61585939 Chr.19: 49155167-49155167 on Build GRCh38
rs759271855 Chr.19: 49155182-49155182 on Build GRCh38
rs145441002 Chr.19: 49155030-49155030 on Build GRCh38
rs138059420 Chr.19: 49155087-49155087 on Build GRCh38
rs749814407 Chr.19: 49154913-49154913 on Build GRCh38
rs754075548 Chr.19: 49155146-49155146 on Build GRCh38
rs567101396 Chr.19: 49155103-49155103 on Build GRCh38
rs779130783 Chr.19: 49155009-49155009 on Build GRCh38
rs761830700 Chr.19: 49155052-49155052 on Build GRCh38
rs771880201 Chr.19: 49155190-49155190 on Build GRCh38
rs762413705 Chr.19: 49154790-49154790 on Build GRCh38
rs781406128 Chr.19: 49154978-49154978 on Build GRCh38
rs892809953 Chr.19: 49155068-49155068 on Build GRCh38
rs61732827 Chr.19: 49155210-49155210 on Build GRCh38
rs774815749 Chr.19: 49154920-49154920 on Build GRCh38
rs755148910 Chr.19: 49154977-49154977 on Build GRCh38
rs200701712 Chr.19: 49154796-49154796 on Build GRCh38
rs746180918 Chr.19: 49155010-49155010 on Build GRCh38
rs368314141 Chr.19: 49154791-49154791 on Build GRCh38
rs757829023 Chr.19: 49155005-49155005 on Build GRCh38
rs746388493 Chr.19: 49155094-49155094 on Build GRCh38
rs770343124 Chr.19: 49154829-49154829 on Build GRCh38
rs777700679 Chr.19: 49154821-49154821 on Build GRCh38
rs754385123 Chr.19: 49154802-49154802 on Build GRCh38
rs748126883 Chr.19: 49154761-49154761 on Build GRCh38
rs150731204 Chr.19: 49155119-49155119 on Build GRCh38
rs766110453 Chr.19: 49155065-49155065 on Build GRCh38
rs3745299 Chr.19: 49155133-49155133 on Build GRCh38
rs553028431 Chr.19: 49154814-49154814 on Build GRCh38
rs771828053 Chr.19: 49154938-49154938 on Build GRCh38
rs749759363 Chr.19: 49155002-49155002 on Build GRCh38
rs780084646 Chr.19: 49154823-49154823 on Build GRCh38
rs767072386 Chr.19: 49154812-49154812 on Build GRCh38
rs759085109 Chr.19: 49154923-49154923 on Build GRCh38
rs746827859 Chr.19: 49154829-49154830 on Build GRCh38
rs775710183 Chr.19: 49154786-49154786 on Build GRCh38
rs776330249 Chr.19: 49155049-49155049 on Build GRCh38
rs768579628 Chr.19: 49154826-49154826 on Build GRCh38
rs951175417 Chr.19: 49154906-49154906 on Build GRCh38
rs117942355 Chr.19: 49155088-49155088 on Build GRCh38
rs557094516 Chr.19: 49155066-49155066 on Build GRCh38
rs770075079 Chr.19: 49155209-49155212 on Build GRCh38
rs762750640 Chr.19: 49155064-49155064 on Build GRCh38
rs773026216 Chr.19: 49155056-49155056 on Build GRCh38
rs550534373 Chr.19: 49155042-49155042 on Build GRCh38
rs773559151 Chr.19: 49154832-49154832 on Build GRCh38
rs768798109 Chr.19: 49154788-49154788 on Build GRCh38
rs1017233906 Chr.19: 49154943-49154943 on Build GRCh38
rs1028086185 Chr.19: 49154799-49154799 on Build GRCh38
rs200689421 Chr.19: 49154999-49154999 on Build GRCh38
rs776241878 Chr.19: 49155108-49155110 on Build GRCh38
rs756563869 Chr.19: 49155153-49155153 on Build GRCh38
rs200146692 Chr.19: 49155100-49155100 on Build GRCh38
rs755278652 Chr.19: 49154887-49154887 on Build GRCh38
rs749223234 Chr.19: 49155178-49155178 on Build GRCh38
rs768251645 Chr.19: 49155047-49155047 on Build GRCh38
rs746108572 Chr.19: 49155143-49155145 on Build GRCh38
rs757313452 Chr.19: 49155152-49155152 on Build GRCh38
rs540512844 Chr.19: 49154813-49154813 on Build GRCh38
rs558301743 Chr.19: 49155120-49155120 on Build GRCh38
rs532514821 Chr.19: 49155011-49155011 on Build GRCh38
rs776877200 Chr.19: 49155195-49155195 on Build GRCh38
rs1011259746 Chr.19: 49155069-49155069 on Build GRCh38
rs187889965 Chr.19: 49154983-49154983 on Build GRCh38
rs764367539 Chr.19: 49155175-49155175 on Build GRCh38
rs755812840 Chr.19: 49154766-49154766 on Build GRCh38
rs780923644 Chr.19: 49154755-49154755 on Build GRCh38
rs147750873 Chr.19: 49154946-49154946 on Build GRCh38
rs1053158883 Chr.19: 49155044-49155044 on Build GRCh38
rs199719753 Chr.19: 49154857-49154857 on Build GRCh38
rs764133314 Chr.19: 49155081-49155081 on Build GRCh38
rs762578736 Chr.19: 49154792-49154792 on Build GRCh38
rs765287843 Chr.19: 49155053-49155053 on Build GRCh38
rs777739525 Chr.19: 49154772-49154772 on Build GRCh38
rs771240704 Chr.19: 49154917-49154917 on Build GRCh38
rs755400580 Chr.19: 49155168-49155168 on Build GRCh38
rs74666147 Chr.19: 49154947-49154947 on Build GRCh38
rs749165705 Chr.19: 49154822-49154822 on Build GRCh38
rs763236818 Chr.19: 49154843-49154843 on Build GRCh38
rs148966785 Chr.19: 49154976-49154976 on Build GRCh38
rs3745297 Chr.19: 49154952-49154952 on Build GRCh38
rs375530211 Chr.19: 49155166-49155166 on Build GRCh38
rs778194640 Chr.19: 49154899-49154899 on Build GRCh38
rs868339772 Chr.19: 49154875-49154875 on Build GRCh38
rs57897109 Chr.19: 49154827-49154827 on Build GRCh38
rs3745298 Chr.19: 49155110-49155110 on Build GRCh38
rs781119935 Chr.19: 49154890-49154890 on Build GRCh38
rs780606171 Chr.19: 49155097-49155097 on Build GRCh38
rs1043250044 Chr.19: 49155084-49155084 on Build GRCh38
rs369761377 Chr.19: 49154987-49154987 on Build GRCh38
rs149712080 Chr.19: 49155096-49155096 on Build GRCh38
rs777184848 Chr.19: 49155177-49155177 on Build GRCh38
rs770909687 Chr.19: 49155179-49155179 on Build GRCh38
rs756364611 Chr.19: 49155072-49155072 on Build GRCh38
rs760689300 Chr.19: 49154869-49154869 on Build GRCh38
rs200253547 Chr.19: 49154783-49154783 on Build GRCh38
rs144139026 Chr.19: 49154752-49154752 on Build GRCh38
rs765267082 Chr.19: 49155160-49155160 on Build GRCh38
rs776955882 Chr.19: 49154789-49154789 on Build GRCh38
rs751700207 Chr.19: 49154883-49154883 on Build GRCh38
rs746053074 Chr.19: 49154782-49154782 on Build GRCh38
rs374958519 Chr.19: 49154975-49154975 on Build GRCh38
rs777753188 Chr.19: 49154762-49154770 on Build GRCh38
rs780932995 Chr.19: 49155169-49155169 on Build GRCh38
rs760870417 Chr.19: 49155130-49155130 on Build GRCh38
rs865942950 Chr.19: 49154808-49154808 on Build GRCh38
rs561843263 Chr.19: 49154779-49154779 on Build GRCh38
rs746997929 Chr.19: 49154787-49154787 on Build GRCh38
rs374745031 Chr.19: 49154862-49154862 on Build GRCh38
rs999154531 Chr.19: 49155025-49155025 on Build GRCh38
rs766087080 Chr.19: 49154793-49154793 on Build GRCh38
rs148276077 Chr.19: 49155099-49155099 on Build GRCh38
rs769199329 Chr.19: 49155104-49155104 on Build GRCh38
rs138754639 Chr.19: 49154958-49154958 on Build GRCh38
rs771297889 Chr.19: 49154844-49154844 on Build GRCh38
rs778800866 Chr.19: 49154880-49154880 on Build GRCh38
rs145567121 Chr.19: 49155181-49155181 on Build GRCh38
rs141840823 Chr.19: 49154864-49154864 on Build GRCh38
rs372492411 Chr.19: 49154847-49154847 on Build GRCh38
rs1028012565 Chr.19: 49155085-49155085 on Build GRCh38
rs748293010 Chr.19: 49154892-49154892 on Build GRCh38
rs747279134 Chr.19: 49154825-49154825 on Build GRCh38
rs779421982 Chr.19: 49155093-49155093 on Build GRCh38

View Full Product Details