Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55365426-55365924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL11
Entrez Gene ID
3589
Gene Name
interleukin 11
Gene Aliases
AGIF, IL-11
UniGene
Hs.467304
Gene Chromosome Location
Chr.19: 55364382-55370463 on Build GRCh38
Transcripts
NM_000641.3 NM_001267718.1

SNP Detail

SNP ID Chromosome Location
rs180741776 Chr.19: 55365784-55365784 on Build GRCh38
rs914944659 Chr.19: 55365654-55365654 on Build GRCh38
rs964341672 Chr.19: 55365843-55365843 on Build GRCh38
rs537030639 Chr.19: 55365446-55365446 on Build GRCh38
rs540747284 Chr.19: 55365468-55365468 on Build GRCh38
rs920526323 Chr.19: 55365860-55365860 on Build GRCh38
rs1017719667 Chr.19: 55365827-55365827 on Build GRCh38
rs111605504 Chr.19: 55365463-55365463 on Build GRCh38
rs1055273029 Chr.19: 55365746-55365746 on Build GRCh38
rs1047530087 Chr.19: 55365581-55365581 on Build GRCh38
rs983716184 Chr.19: 55365868-55365868 on Build GRCh38
rs895313026 Chr.19: 55365496-55365496 on Build GRCh38
rs929142877 Chr.19: 55365553-55365553 on Build GRCh38
rs4252558 Chr.19: 55365878-55365878 on Build GRCh38
rs572947521 Chr.19: 55365501-55365501 on Build GRCh38
rs532707375 Chr.19: 55365792-55365792 on Build GRCh38
rs989246425 Chr.19: 55365639-55365639 on Build GRCh38
rs943316715 Chr.19: 55365533-55365533 on Build GRCh38
rs530333352 Chr.19: 55365769-55365769 on Build GRCh38
rs10402868 Chr.19: 55365578-55365578 on Build GRCh38
rs1019395052 Chr.19: 55365858-55365858 on Build GRCh38
rs186526404 Chr.19: 55365754-55365754 on Build GRCh38
rs987224376 Chr.19: 55365826-55365826 on Build GRCh38
rs1018926908 Chr.19: 55365557-55365557 on Build GRCh38
rs938764456 Chr.19: 55365669-55365669 on Build GRCh38
rs879289625 Chr.19: 55365562-55365562 on Build GRCh38
rs950767961 Chr.19: 55365541-55365541 on Build GRCh38
rs921106762 Chr.19: 55365573-55365573 on Build GRCh38
rs894469514 Chr.19: 55365722-55365722 on Build GRCh38
rs1042506 Chr.19: 55365743-55365743 on Build GRCh38
rs190723249 Chr.19: 55365770-55365770 on Build GRCh38
rs965712066 Chr.19: 55365564-55365564 on Build GRCh38
rs4806475 Chr.19: 55365800-55365800 on Build GRCh38
rs533866922 Chr.19: 55365447-55365447 on Build GRCh38
rs998105529 Chr.19: 55365871-55365871 on Build GRCh38
rs4252559 Chr.19: 55365507-55365507 on Build GRCh38
rs780299709 Chr.19: 55365656-55365656 on Build GRCh38
rs375257734 Chr.19: 55365749-55365749 on Build GRCh38
rs1042471807 Chr.19: 55365703-55365703 on Build GRCh38
rs1031813995 Chr.19: 55365799-55365799 on Build GRCh38
rs994891749 Chr.19: 55365503-55365503 on Build GRCh38
rs565362807 Chr.19: 55365588-55365588 on Build GRCh38
rs4252560 Chr.19: 55365461-55365461 on Build GRCh38
rs1042505 Chr.19: 55365863-55365863 on Build GRCh38
rs781609515 Chr.19: 55365817-55365817 on Build GRCh38
rs983884794 Chr.19: 55365548-55365548 on Build GRCh38
rs927650319 Chr.19: 55365713-55365713 on Build GRCh38
rs867849030 Chr.19: 55365902-55365902 on Build GRCh38
rs953819179 Chr.19: 55365598-55365598 on Build GRCh38
rs4252578 Chr.19: 55365887-55365887 on Build GRCh38
rs530841159 Chr.19: 55365671-55365671 on Build GRCh38
rs10402867 Chr.19: 55365577-55365577 on Build GRCh38
rs906808914 Chr.19: 55365457-55365457 on Build GRCh38
rs1037571431 Chr.19: 55365538-55365538 on Build GRCh38
rs886544917 Chr.19: 55365798-55365798 on Build GRCh38
rs558624144 Chr.19: 55365455-55365455 on Build GRCh38
rs1049160008 Chr.19: 55365787-55365787 on Build GRCh38
rs186048072 Chr.19: 55365882-55365882 on Build GRCh38
rs1028219914 Chr.19: 55365877-55365877 on Build GRCh38
rs976654991 Chr.19: 55365565-55365565 on Build GRCh38

View Full Product Details