Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9961363-9961871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRHL1
Entrez Gene ID
29841
Gene Name
grainyhead like transcription factor 1
Gene Aliases
LBP32, MGR, NH32, TFCP2L2
UniGene
Hs.418493
Gene Chromosome Location
Chr.2: 9951663-10002284 on Build GRCh38
Transcripts
NM_198182.2 XM_005246159.3 XM_006711882.2 XM_006711884.3 XM_011510343.2 XM_017003900.1 XM_017003901.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1690191 c.142C>T p.R48* Chr.2: 9961399-9961399 on Build GRCh38
4083735 c.661G>A p.V221I Chr.2: 9961428-9961428 on Build GRCh38
4300935 c.607A>C p.N203H Chr.2: 9961374-9961374 on Build GRCh38
3564282 c.169G>A p.A57T Chr.2: 9961426-9961426 on Build GRCh38
3564281 c.659G>A p.G220D Chr.2: 9961426-9961426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs137999440 Chr.2: 9961671-9961671 on Build GRCh38
rs1014433260 Chr.2: 9961427-9961427 on Build GRCh38
rs757786464 Chr.2: 9961399-9961399 on Build GRCh38
rs4669512 Chr.2: 9961816-9961816 on Build GRCh38
rs554097110 Chr.2: 9961715-9961715 on Build GRCh38
rs16867257 Chr.2: 9961540-9961540 on Build GRCh38
rs528790803 Chr.2: 9961511-9961511 on Build GRCh38
rs753310015 Chr.2: 9961395-9961395 on Build GRCh38
rs950237688 Chr.2: 9961837-9961837 on Build GRCh38
rs376447054 Chr.2: 9961465-9961465 on Build GRCh38
rs1017774268 Chr.2: 9961846-9961846 on Build GRCh38
rs746495632 Chr.2: 9961624-9961624 on Build GRCh38
rs894131814 Chr.2: 9961416-9961416 on Build GRCh38
rs780528847 Chr.2: 9961722-9961722 on Build GRCh38
rs905893342 Chr.2: 9961615-9961615 on Build GRCh38
rs949705051 Chr.2: 9961748-9961748 on Build GRCh38
rs574522639 Chr.2: 9961654-9961654 on Build GRCh38
rs141699066 Chr.2: 9961678-9961678 on Build GRCh38
rs763692885 Chr.2: 9961480-9961480 on Build GRCh38
rs771743850 Chr.2: 9961471-9961471 on Build GRCh38
rs189930547 Chr.2: 9961451-9961451 on Build GRCh38
rs78264111 Chr.2: 9961400-9961400 on Build GRCh38
rs897989753 Chr.2: 9961450-9961450 on Build GRCh38
rs781452558 Chr.2: 9961453-9961453 on Build GRCh38
rs1007754853 Chr.2: 9961542-9961542 on Build GRCh38
rs1045896522 Chr.2: 9961768-9961768 on Build GRCh38
rs760279118 Chr.2: 9961474-9961474 on Build GRCh38
rs1007785747 Chr.2: 9961829-9961829 on Build GRCh38
rs539464106 Chr.2: 9961772-9961772 on Build GRCh38
rs566730606 Chr.2: 9961814-9961814 on Build GRCh38
rs1044483494 Chr.2: 9961608-9961608 on Build GRCh38
rs1017574310 Chr.2: 9961553-9961553 on Build GRCh38
rs758193786 Chr.2: 9961463-9961463 on Build GRCh38
rs181100464 Chr.2: 9961707-9961707 on Build GRCh38
rs966449222 Chr.2: 9961847-9961847 on Build GRCh38
rs759434226 Chr.2: 9961466-9961466 on Build GRCh38
rs754892625 Chr.2: 9961388-9961388 on Build GRCh38
rs750801214 Chr.2: 9961412-9961412 on Build GRCh38
rs112410023 Chr.2: 9961795-9961795 on Build GRCh38
rs138724700 Chr.2: 9961558-9961558 on Build GRCh38
rs748189809 Chr.2: 9961459-9961459 on Build GRCh38
rs758682916 Chr.2: 9961420-9961420 on Build GRCh38
rs775251158 Chr.2: 9961472-9961472 on Build GRCh38
rs976576695 Chr.2: 9961576-9961576 on Build GRCh38
rs140278187 Chr.2: 9961428-9961428 on Build GRCh38
rs951824805 Chr.2: 9961732-9961732 on Build GRCh38
rs910148133 Chr.2: 9961791-9961791 on Build GRCh38
rs776108583 Chr.2: 9961780-9961780 on Build GRCh38
rs778718467 Chr.2: 9961446-9961446 on Build GRCh38
rs1049250951 Chr.2: 9961797-9961797 on Build GRCh38
rs747173521 Chr.2: 9961441-9961441 on Build GRCh38
rs891692369 Chr.2: 9961664-9961664 on Build GRCh38
rs150557170 Chr.2: 9961769-9961769 on Build GRCh38
rs950137795 Chr.2: 9961558-9961558 on Build GRCh38
rs1055690404 Chr.2: 9961708-9961708 on Build GRCh38
rs972898656 Chr.2: 9961813-9961813 on Build GRCh38

View Full Product Details