Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26191393-26191898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAREM2
Entrez Gene ID
150946
Gene Name
GRB2 associated regulator of MAPK1 subtype 2
Gene Aliases
FAM59B, GAREML
UniGene
Hs.187912
Gene Chromosome Location
Chr.2: 26173091-26202405 on Build GRCh38
Transcripts
XM_011532567.2 NM_000182.4

Gene Symbol
HADHA
Entrez Gene ID
3030
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit
Gene Aliases
ECHA, GBP, HADH, LCEH, LCHAD, MTPA, TP-ALPHA
UniGene
Hs.516032
Gene Chromosome Location
Chr.2: 26190635-26244726 on Build GRCh38
Transcripts
XM_011532567.2 NM_000182.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6677464 c.2059A>G p.M687V Chr.2: 26191570-26191570 on Build GRCh38
1306643 c.2039G>C p.R680T Chr.2: 26191590-26191590 on Build GRCh38
4381683 c.2032G>T p.V678L Chr.2: 26191597-26191597 on Build GRCh38
6278302 c.2112C>T p.A704A Chr.2: 26191517-26191517 on Build GRCh38
5930896 c.2147-3C>T p.? Chr.2: 26191398-26191398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs267599302 Chr.2: 26191523-26191523 on Build GRCh38
rs767616996 Chr.2: 26191497-26191497 on Build GRCh38
rs200075452 Chr.2: 26191446-26191446 on Build GRCh38
rs557453199 Chr.2: 26191738-26191738 on Build GRCh38
rs200438844 Chr.2: 26191522-26191522 on Build GRCh38
rs775606155 Chr.2: 26191466-26191466 on Build GRCh38
rs535654673 Chr.2: 26191436-26191436 on Build GRCh38
rs766306913 Chr.2: 26191437-26191437 on Build GRCh38
rs950132581 Chr.2: 26191821-26191821 on Build GRCh38
rs868693884 Chr.2: 26191608-26191609 on Build GRCh38
rs749316549 Chr.2: 26191596-26191596 on Build GRCh38
rs186499456 Chr.2: 26191496-26191496 on Build GRCh38
rs202066629 Chr.2: 26191525-26191525 on Build GRCh38
rs765521978 Chr.2: 26191428-26191428 on Build GRCh38
rs372827257 Chr.2: 26191429-26191429 on Build GRCh38
rs587776501 Chr.2: 26191546-26191546 on Build GRCh38
rs1017968154 Chr.2: 26191552-26191552 on Build GRCh38
rs1027408812 Chr.2: 26191847-26191848 on Build GRCh38
rs1057517132 Chr.2: 26191558-26191558 on Build GRCh38
rs919223394 Chr.2: 26191839-26191839 on Build GRCh38
rs756433832 Chr.2: 26191611-26191611 on Build GRCh38
rs149347225 Chr.2: 26191655-26191657 on Build GRCh38
rs776491880 Chr.2: 26191474-26191474 on Build GRCh38
rs775551171 Chr.2: 26191632-26191632 on Build GRCh38
rs774667417 Chr.2: 26191747-26191747 on Build GRCh38
rs775407799 Chr.2: 26191419-26191419 on Build GRCh38
rs778281315 Chr.2: 26191464-26191464 on Build GRCh38
rs756431871 Chr.2: 26191448-26191448 on Build GRCh38
rs539850142 Chr.2: 26191741-26191741 on Build GRCh38
rs757938383 Chr.2: 26191535-26191535 on Build GRCh38
rs201929031 Chr.2: 26191447-26191447 on Build GRCh38
rs758130419 Chr.2: 26191671-26191671 on Build GRCh38
rs1008724130 Chr.2: 26191751-26191751 on Build GRCh38
rs753068811 Chr.2: 26191519-26191527 on Build GRCh38
rs1057516350 Chr.2: 26191481-26191481 on Build GRCh38
rs897765805 Chr.2: 26191784-26191785 on Build GRCh38
rs890316065 Chr.2: 26191749-26191749 on Build GRCh38
rs780054585 Chr.2: 26191500-26191500 on Build GRCh38
rs374503588 Chr.2: 26191508-26191508 on Build GRCh38
rs759778780 Chr.2: 26191620-26191620 on Build GRCh38
rs199857377 Chr.2: 26191483-26191483 on Build GRCh38
rs200961004 Chr.2: 26191465-26191465 on Build GRCh38
rs765104107 Chr.2: 26191598-26191598 on Build GRCh38
rs771028541 Chr.2: 26191603-26191603 on Build GRCh38
rs377688807 Chr.2: 26191619-26191619 on Build GRCh38
rs537486195 Chr.2: 26191423-26191423 on Build GRCh38
rs794727219 Chr.2: 26191482-26191482 on Build GRCh38
rs200486145 Chr.2: 26191421-26191421 on Build GRCh38
rs114021904 Chr.2: 26191766-26191766 on Build GRCh38
rs145930159 Chr.2: 26191467-26191467 on Build GRCh38
rs754125767 Chr.2: 26191658-26191658 on Build GRCh38
rs994726642 Chr.2: 26191801-26191801 on Build GRCh38
rs750040371 Chr.2: 26191666-26191666 on Build GRCh38
rs768516891 Chr.2: 26191473-26191473 on Build GRCh38
rs372669268 Chr.2: 26191460-26191460 on Build GRCh38
rs112966040 Chr.2: 26191567-26191567 on Build GRCh38
rs1055722662 Chr.2: 26191418-26191418 on Build GRCh38
rs1036860952 Chr.2: 26191828-26191828 on Build GRCh38
rs181778579 Chr.2: 26191417-26191417 on Build GRCh38
rs748024841 Chr.2: 26191597-26191597 on Build GRCh38
rs755428570 Chr.2: 26191444-26191444 on Build GRCh38
rs893435756 Chr.2: 26191737-26191737 on Build GRCh38
rs764719697 Chr.2: 26191652-26191652 on Build GRCh38
rs575328538 Chr.2: 26191664-26191664 on Build GRCh38
rs149632783 Chr.2: 26191569-26191569 on Build GRCh38
rs572555864 Chr.2: 26191739-26191739 on Build GRCh38
rs917365964 Chr.2: 26191452-26191452 on Build GRCh38
rs1053865333 Chr.2: 26191721-26191721 on Build GRCh38
rs761164879 Chr.2: 26191643-26191643 on Build GRCh38
rs779726875 Chr.2: 26191536-26191536 on Build GRCh38
rs979101095 Chr.2: 26191442-26191442 on Build GRCh38
rs754294789 Chr.2: 26191517-26191517 on Build GRCh38
rs925893560 Chr.2: 26191720-26191720 on Build GRCh38
rs756964298 Chr.2: 26191587-26191587 on Build GRCh38
rs201029031 Chr.2: 26191663-26191663 on Build GRCh38
rs763085690 Chr.2: 26191480-26191480 on Build GRCh38
rs529622107 Chr.2: 26191859-26191859 on Build GRCh38
rs768784732 Chr.2: 26191588-26191588 on Build GRCh38
rs751732102 Chr.2: 26191443-26191443 on Build GRCh38
rs778131571 Chr.2: 26191454-26191454 on Build GRCh38
rs138966725 Chr.2: 26191498-26191498 on Build GRCh38
rs368736585 Chr.2: 26191459-26191459 on Build GRCh38
rs771245886 Chr.2: 26191490-26191490 on Build GRCh38
rs770676727 Chr.2: 26191809-26191809 on Build GRCh38
rs545558100 Chr.2: 26191688-26191688 on Build GRCh38
rs772080441 Chr.2: 26191625-26191625 on Build GRCh38
rs371979389 Chr.2: 26191708-26191708 on Build GRCh38
rs146406360 Chr.2: 26191516-26191516 on Build GRCh38

View Full Product Details