Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26197607-26198105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAREM2
Entrez Gene ID
150946
Gene Name
GRB2 associated regulator of MAPK1 subtype 2
Gene Aliases
FAM59B, GAREML
UniGene
Hs.187912
Gene Chromosome Location
Chr.2: 26173091-26202405 on Build GRCh38
Transcripts
XM_011532567.2 NM_000182.4

Gene Symbol
HADHA
Entrez Gene ID
3030
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit
Gene Aliases
ECHA, GBP, HADH, LCEH, LCHAD, MTPA, TP-ALPHA
UniGene
Hs.516032
Gene Chromosome Location
Chr.2: 26190635-26244726 on Build GRCh38
Transcripts
XM_011532567.2 NM_000182.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3713753 c.1435C>T p.L479F Chr.2: 26197735-26197735 on Build GRCh38
1578865 c.1453G>A p.A485T Chr.2: 26197717-26197717 on Build GRCh38
281616 c.1452C>T p.I484I Chr.2: 26197718-26197718 on Build GRCh38
237491 c.1479+10G>C p.? Chr.2: 26197681-26197681 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774681333 Chr.2: 26197828-26197828 on Build GRCh38
rs2289020 Chr.2: 26197880-26197880 on Build GRCh38
rs572765453 Chr.2: 26197797-26197797 on Build GRCh38
rs762279071 Chr.2: 26197835-26197835 on Build GRCh38
rs866059606 Chr.2: 26198000-26198000 on Build GRCh38
rs567683820 Chr.2: 26197878-26197878 on Build GRCh38
rs1054727767 Chr.2: 26198032-26198032 on Build GRCh38
rs745990980 Chr.2: 26197750-26197750 on Build GRCh38
rs141875362 Chr.2: 26197912-26197913 on Build GRCh38
rs79286973 Chr.2: 26197971-26197971 on Build GRCh38
rs750009289 Chr.2: 26197711-26197711 on Build GRCh38
rs568307364 Chr.2: 26197941-26197941 on Build GRCh38
rs766002849 Chr.2: 26197822-26197822 on Build GRCh38
rs187094345 Chr.2: 26198055-26198055 on Build GRCh38
rs563051653 Chr.2: 26197864-26197864 on Build GRCh38
rs940189922 Chr.2: 26197993-26197993 on Build GRCh38
rs991588740 Chr.2: 26198012-26198012 on Build GRCh38
rs760026395 Chr.2: 26197667-26197667 on Build GRCh38
rs971404806 Chr.2: 26198065-26198065 on Build GRCh38
rs772166712 Chr.2: 26197752-26197752 on Build GRCh38
rs773436007 Chr.2: 26197646-26197646 on Build GRCh38
rs763098015 Chr.2: 26197652-26197652 on Build GRCh38
rs377123628 Chr.2: 26197730-26197730 on Build GRCh38
rs756864103 Chr.2: 26197687-26197687 on Build GRCh38
rs2289019 Chr.2: 26197940-26197940 on Build GRCh38
rs753943033 Chr.2: 26197925-26197925 on Build GRCh38
rs776013887 Chr.2: 26197763-26197763 on Build GRCh38
rs758130768 Chr.2: 26197739-26197739 on Build GRCh38
rs967192879 Chr.2: 26197943-26197943 on Build GRCh38
rs758386186 Chr.2: 26197718-26197718 on Build GRCh38
rs374046333 Chr.2: 26197808-26197808 on Build GRCh38
rs770931882 Chr.2: 26197661-26197661 on Build GRCh38
rs764385972 Chr.2: 26197683-26197683 on Build GRCh38
rs13387855 Chr.2: 26197839-26197839 on Build GRCh38
rs768065156 Chr.2: 26197677-26197677 on Build GRCh38
rs145422395 Chr.2: 26197705-26197705 on Build GRCh38
rs745946711 Chr.2: 26197797-26197797 on Build GRCh38
rs888168589 Chr.2: 26197897-26197897 on Build GRCh38
rs894807641 Chr.2: 26198051-26198051 on Build GRCh38
rs1020090937 Chr.2: 26197656-26197656 on Build GRCh38
rs930992571 Chr.2: 26197671-26197671 on Build GRCh38
rs13385152 Chr.2: 26197916-26197916 on Build GRCh38
rs1048124951 Chr.2: 26197881-26197881 on Build GRCh38
rs746944577 Chr.2: 26197729-26197729 on Build GRCh38
rs776925695 Chr.2: 26197792-26197792 on Build GRCh38
rs998100641 Chr.2: 26197939-26197939 on Build GRCh38
rs1000056377 Chr.2: 26197949-26197949 on Build GRCh38
rs560608592 Chr.2: 26197918-26197918 on Build GRCh38
rs184023842 Chr.2: 26197921-26197921 on Build GRCh38
rs920196141 Chr.2: 26198008-26198008 on Build GRCh38
rs894901125 Chr.2: 26197696-26197696 on Build GRCh38
rs764448963 Chr.2: 26197771-26197771 on Build GRCh38
rs780854216 Chr.2: 26197736-26197736 on Build GRCh38
rs771205629 Chr.2: 26197742-26197742 on Build GRCh38
rs573126129 Chr.2: 26197958-26197958 on Build GRCh38
rs538871015 Chr.2: 26198067-26198067 on Build GRCh38
rs550390870 Chr.2: 26198029-26198029 on Build GRCh38
rs958717402 Chr.2: 26197717-26197717 on Build GRCh38
rs764119897 Chr.2: 26197995-26197995 on Build GRCh38
rs757837042 Chr.2: 26197763-26197763 on Build GRCh38
rs991869018 Chr.2: 26197723-26197723 on Build GRCh38
rs201472602 Chr.2: 26197731-26197731 on Build GRCh38
rs375746236 Chr.2: 26197810-26197810 on Build GRCh38
rs761544248 Chr.2: 26197793-26197793 on Build GRCh38
rs555281625 Chr.2: 26197864-26197882 on Build GRCh38
rs544867098 Chr.2: 26197861-26197861 on Build GRCh38
rs761058783 Chr.2: 26197682-26197682 on Build GRCh38
rs779501172 Chr.2: 26197793-26197794 on Build GRCh38
rs545776378 Chr.2: 26197765-26197765 on Build GRCh38
rs774394602 Chr.2: 26197745-26197745 on Build GRCh38
rs77141401 Chr.2: 26197801-26197801 on Build GRCh38
rs774485832 Chr.2: 26197666-26197666 on Build GRCh38
rs779169526 Chr.2: 26198009-26198009 on Build GRCh38

View Full Product Details